Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 8867032-8867540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO1
Entrez Gene ID
2023
Gene Name
enolase 1
Gene Aliases
ENO1L1, HEL-S-17, MPB1, NNE, PPH
UniGene
Hs.517145
Gene Chromosome Location
Chr.1: 8861000-8879092 on Build GRCh38
Transcripts
NM_001201483.1 NM_001428.3 XM_006710433.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
912899 c.419A>G p.N140S Chr.1: 8867142-8867142 on Build GRCh38
5381648 c.385C>T p.P129S Chr.1: 8867176-8867176 on Build GRCh38
912901 c.323C>T p.A108V Chr.1: 8867238-8867238 on Build GRCh38
912900 c.402C>T p.I134I Chr.1: 8867159-8867159 on Build GRCh38
1251187 c.383T>C p.V128A Chr.1: 8867178-8867178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111654228 Chr.1: 8867251-8867251 on Build GRCh38
rs11544519 Chr.1: 8867191-8867191 on Build GRCh38
rs368745021 Chr.1: 8867340-8867351 on Build GRCh38
rs567042358 Chr.1: 8867280-8867280 on Build GRCh38
rs770003428 Chr.1: 8867351-8867351 on Build GRCh38
rs374589133 Chr.1: 8867290-8867290 on Build GRCh38
rs11544497 Chr.1: 8867162-8867162 on Build GRCh38
rs767370428 Chr.1: 8867278-8867279 on Build GRCh38
rs529227052 Chr.1: 8867166-8867166 on Build GRCh38
rs753057751 Chr.1: 8867268-8867268 on Build GRCh38
rs747773922 Chr.1: 8867116-8867116 on Build GRCh38
rs144538182 Chr.1: 8867166-8867167 on Build GRCh38
rs11544507 Chr.1: 8867204-8867204 on Build GRCh38
rs763807184 Chr.1: 8867320-8867320 on Build GRCh38
rs183139190 Chr.1: 8867269-8867269 on Build GRCh38
rs777321133 Chr.1: 8867179-8867179 on Build GRCh38
rs760635870 Chr.1: 8867139-8867139 on Build GRCh38
rs762074733 Chr.1: 8867232-8867232 on Build GRCh38
rs756633097 Chr.1: 8867163-8867163 on Build GRCh38
rs757303301 Chr.1: 8867069-8867069 on Build GRCh38
rs767692238 Chr.1: 8867233-8867233 on Build GRCh38
rs763006734 Chr.1: 8867239-8867240 on Build GRCh38
rs774328034 Chr.1: 8867248-8867271 on Build GRCh38
rs772632044 Chr.1: 8867133-8867133 on Build GRCh38
rs752580188 Chr.1: 8867267-8867267 on Build GRCh38
rs779444406 Chr.1: 8867173-8867173 on Build GRCh38
rs1015434005 Chr.1: 8867119-8867119 on Build GRCh38
rs758032766 Chr.1: 8867177-8867177 on Build GRCh38
rs1013476900 Chr.1: 8867426-8867426 on Build GRCh38
rs765668364 Chr.1: 8867263-8867263 on Build GRCh38
rs878958212 Chr.1: 8867115-8867115 on Build GRCh38
rs28999081 Chr.1: 8867517-8867517 on Build GRCh38
rs1003355179 Chr.1: 8867317-8867317 on Build GRCh38
rs756707613 Chr.1: 8867278-8867278 on Build GRCh38
rs746684013 Chr.1: 8867276-8867276 on Build GRCh38
rs1040788874 Chr.1: 8867238-8867238 on Build GRCh38
rs199529530 Chr.1: 8867209-8867209 on Build GRCh38
rs768419437 Chr.1: 8867110-8867110 on Build GRCh38
rs759557009 Chr.1: 8867210-8867210 on Build GRCh38
rs962155672 Chr.1: 8867142-8867142 on Build GRCh38
rs556055143 Chr.1: 8867346-8867346 on Build GRCh38
rs767059816 Chr.1: 8867164-8867164 on Build GRCh38
rs538198787 Chr.1: 8867446-8867446 on Build GRCh38
rs775376513 Chr.1: 8867179-8867179 on Build GRCh38
rs200899556 Chr.1: 8867296-8867296 on Build GRCh38
rs187859355 Chr.1: 8867476-8867476 on Build GRCh38
rs906347060 Chr.1: 8867316-8867316 on Build GRCh38
rs142256351 Chr.1: 8867144-8867144 on Build GRCh38
rs754352845 Chr.1: 8867245-8867245 on Build GRCh38
rs888326267 Chr.1: 8867063-8867063 on Build GRCh38
rs560859958 Chr.1: 8867147-8867147 on Build GRCh38
rs770911403 Chr.1: 8867141-8867141 on Build GRCh38
rs200935067 Chr.1: 8867237-8867237 on Build GRCh38
rs574276319 Chr.1: 8867402-8867402 on Build GRCh38
rs771564625 Chr.1: 8867130-8867130 on Build GRCh38
rs761970983 Chr.1: 8867158-8867158 on Build GRCh38
rs1033328628 Chr.1: 8867312-8867312 on Build GRCh38
rs747799544 Chr.1: 8867176-8867176 on Build GRCh38
rs148815455 Chr.1: 8867082-8867082 on Build GRCh38
rs534407998 Chr.1: 8867328-8867328 on Build GRCh38
rs1001859718 Chr.1: 8867118-8867118 on Build GRCh38
rs377003555 Chr.1: 8867098-8867098 on Build GRCh38
rs768503842 Chr.1: 8867302-8867302 on Build GRCh38
rs776492610 Chr.1: 8867195-8867195 on Build GRCh38
rs569194886 Chr.1: 8867234-8867234 on Build GRCh38
rs751731640 Chr.1: 8867154-8867154 on Build GRCh38
rs370977906 Chr.1: 8867279-8867279 on Build GRCh38
rs750901262 Chr.1: 8867089-8867089 on Build GRCh38
rs761677406 Chr.1: 8867250-8867251 on Build GRCh38
rs763933974 Chr.1: 8867244-8867246 on Build GRCh38
rs778645839 Chr.1: 8867113-8867113 on Build GRCh38
rs756621363 Chr.1: 8867095-8867095 on Build GRCh38
rs556109901 Chr.1: 8867455-8867455 on Build GRCh38
rs550948324 Chr.1: 8867222-8867222 on Build GRCh38
rs373738971 Chr.1: 8867469-8867469 on Build GRCh38
rs769535629 Chr.1: 8867221-8867221 on Build GRCh38
rs755341114 Chr.1: 8867167-8867167 on Build GRCh38
rs765029350 Chr.1: 8867153-8867153 on Build GRCh38
rs770428970 Chr.1: 8867192-8867192 on Build GRCh38
rs369611743 Chr.1: 8867159-8867159 on Build GRCh38
rs968968242 Chr.1: 8867488-8867488 on Build GRCh38
rs775343507 Chr.1: 8867294-8867294 on Build GRCh38
rs577594220 Chr.1: 8867461-8867461 on Build GRCh38
rs766584131 Chr.1: 8867239-8867239 on Build GRCh38
rs896144451 Chr.1: 8867371-8867371 on Build GRCh38
rs199776385 Chr.1: 8867254-8867254 on Build GRCh38

View Full Product Details