Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9883616-9884107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNBIP1
Entrez Gene ID
56998
Gene Name
catenin beta interacting protein 1
Gene Aliases
ICAT
UniGene
Hs.463759
Gene Chromosome Location
Chr.1: 9848276-9910322 on Build GRCh38
Transcripts
NM_001012329.1 NM_020248.2 XM_017001853.1 XM_017001854.1 XM_017001855.1

SNP Detail

SNP ID Chromosome Location
rs530975891 Chr.1: 9883940-9883940 on Build GRCh38
rs1010688656 Chr.1: 9883970-9883970 on Build GRCh38
rs945473850 Chr.1: 9883660-9883660 on Build GRCh38
rs894647286 Chr.1: 9883700-9883700 on Build GRCh38
rs549542011 Chr.1: 9883947-9883947 on Build GRCh38
rs78705524 Chr.1: 9883734-9883734 on Build GRCh38
rs998032783 Chr.1: 9883767-9883767 on Build GRCh38
rs528595905 Chr.1: 9883696-9883696 on Build GRCh38
rs182325555 Chr.1: 9883884-9883884 on Build GRCh38
rs962726179 Chr.1: 9883860-9883860 on Build GRCh38
rs781588885 Chr.1: 9883652-9883652 on Build GRCh38
rs747671369 Chr.1: 9883688-9883688 on Build GRCh38
rs906215918 Chr.1: 9883719-9883719 on Build GRCh38
rs930821345 Chr.1: 9883671-9883671 on Build GRCh38
rs1010657505 Chr.1: 9883760-9883760 on Build GRCh38
rs374527257 Chr.1: 9884063-9884063 on Build GRCh38
rs568468152 Chr.1: 9883718-9883718 on Build GRCh38
rs755647345 Chr.1: 9883702-9883702 on Build GRCh38
rs562417332 Chr.1: 9884081-9884081 on Build GRCh38
rs535757762 Chr.1: 9883733-9883733 on Build GRCh38
rs914097383 Chr.1: 9883923-9883923 on Build GRCh38
rs117622411 Chr.1: 9883847-9883847 on Build GRCh38
rs1016681067 Chr.1: 9883848-9883848 on Build GRCh38
rs201061134 Chr.1: 9883678-9883678 on Build GRCh38
rs573618721 Chr.1: 9883948-9883948 on Build GRCh38
rs1001884526 Chr.1: 9883862-9883862 on Build GRCh38
rs1009108305 Chr.1: 9884057-9884057 on Build GRCh38
rs147291986 Chr.1: 9883917-9883917 on Build GRCh38
rs953177689 Chr.1: 9883775-9883775 on Build GRCh38
rs539989473 Chr.1: 9883809-9883809 on Build GRCh38
rs1028717340 Chr.1: 9883984-9883984 on Build GRCh38
rs186894577 Chr.1: 9884011-9884011 on Build GRCh38
rs916110427 Chr.1: 9883914-9883914 on Build GRCh38
rs962073143 Chr.1: 9883819-9883819 on Build GRCh38
rs777343231 Chr.1: 9883872-9883872 on Build GRCh38
rs1045982991 Chr.1: 9883928-9883928 on Build GRCh38
rs190758949 Chr.1: 9883717-9883717 on Build GRCh38
rs966866402 Chr.1: 9883929-9883929 on Build GRCh38
rs34435692 Chr.1: 9883784-9883784 on Build GRCh38
rs930770669 Chr.1: 9883883-9883883 on Build GRCh38
rs569593606 Chr.1: 9883792-9883792 on Build GRCh38
rs923244282 Chr.1: 9884071-9884071 on Build GRCh38
rs893591190 Chr.1: 9883749-9883749 on Build GRCh38
rs992397796 Chr.1: 9883890-9883890 on Build GRCh38
rs950235963 Chr.1: 9883915-9883915 on Build GRCh38
rs888971995 Chr.1: 9884031-9884031 on Build GRCh38
rs925400650 Chr.1: 9883954-9883954 on Build GRCh38
rs999556346 Chr.1: 9883724-9883724 on Build GRCh38
rs934919042 Chr.1: 9883942-9883942 on Build GRCh38
rs1028854203 Chr.1: 9883770-9883770 on Build GRCh38

View Full Product Details