Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11195515-11196024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL7
Entrez Gene ID
10218
Gene Name
angiopoietin like 7
Gene Aliases
AngX, CDT6, dJ647M16.1
UniGene
Hs.146559
Gene Chromosome Location
Chr.1: 11189289-11195981 on Build GRCh38
Transcripts
NM_021146.3 NM_004958.3 XM_005263438.2 XM_011541166.2 XM_017000900.1 XM_017000901.1 XM_017000902.1

Gene Symbol
MTOR
Entrez Gene ID
2475
Gene Name
mechanistic target of rapamycin
Gene Aliases
FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS
UniGene
Hs.338207
Gene Chromosome Location
Chr.1: 11106531-11262551 on Build GRCh38
Transcripts
NM_021146.3 NM_004958.3 XM_005263438.2 XM_011541166.2 XM_017000900.1 XM_017000901.1 XM_017000902.1

SNP Detail

SNP ID Chromosome Location
rs530340225 Chr.1: 11195829-11195829 on Build GRCh38
rs950999517 Chr.1: 11195567-11195567 on Build GRCh38
rs928740542 Chr.1: 11195607-11195607 on Build GRCh38
rs28918371 Chr.1: 11195942-11195942 on Build GRCh38
rs771242634 Chr.1: 11195559-11195559 on Build GRCh38
rs768509483 Chr.1: 11195652-11195652 on Build GRCh38
rs988793564 Chr.1: 11195879-11195879 on Build GRCh38
rs1042882477 Chr.1: 11195673-11195673 on Build GRCh38
rs538680559 Chr.1: 11195609-11195609 on Build GRCh38
rs986215341 Chr.1: 11195960-11195960 on Build GRCh38
rs374380296 Chr.1: 11195759-11195759 on Build GRCh38
rs965393153 Chr.1: 11195546-11195546 on Build GRCh38
rs988575987 Chr.1: 11195617-11195617 on Build GRCh38
rs947311011 Chr.1: 11195652-11195652 on Build GRCh38
rs543616901 Chr.1: 11195938-11195938 on Build GRCh38
rs947381191 Chr.1: 11195892-11195892 on Build GRCh38
rs576198666 Chr.1: 11195873-11195873 on Build GRCh38
rs770483918 Chr.1: 11195646-11195646 on Build GRCh38
rs140746133 Chr.1: 11195726-11195726 on Build GRCh38
rs956724524 Chr.1: 11195595-11195595 on Build GRCh38
rs942494558 Chr.1: 11195940-11195940 on Build GRCh38
rs1043341108 Chr.1: 11195901-11195901 on Build GRCh38
rs913265292 Chr.1: 11195881-11195881 on Build GRCh38
rs368700250 Chr.1: 11195669-11195669 on Build GRCh38
rs771764670 Chr.1: 11195655-11195655 on Build GRCh38
rs1048741795 Chr.1: 11195754-11195754 on Build GRCh38
rs533913726 Chr.1: 11195978-11195981 on Build GRCh38
rs956174491 Chr.1: 11195865-11195865 on Build GRCh38
rs970594216 Chr.1: 11195837-11195837 on Build GRCh38
rs1040629330 Chr.1: 11195725-11195725 on Build GRCh38
rs1053143756 Chr.1: 11195625-11195625 on Build GRCh38
rs900926062 Chr.1: 11195954-11195954 on Build GRCh38
rs968088554 Chr.1: 11195959-11195959 on Build GRCh38
rs28918369 Chr.1: 11195563-11195563 on Build GRCh38
rs12117032 Chr.1: 11195645-11195645 on Build GRCh38
rs562907085 Chr.1: 11195824-11195824 on Build GRCh38
rs28918370 Chr.1: 11195596-11195596 on Build GRCh38
rs966231411 Chr.1: 11195993-11195993 on Build GRCh38
rs17036508 Chr.1: 11195977-11195977 on Build GRCh38
rs1012633017 Chr.1: 11195772-11195772 on Build GRCh38
rs1025302766 Chr.1: 11195739-11195739 on Build GRCh38
rs971693620 Chr.1: 11195561-11195561 on Build GRCh38
rs980305225 Chr.1: 11195863-11195863 on Build GRCh38
rs1041152243 Chr.1: 11195713-11195713 on Build GRCh38
rs960150587 Chr.1: 11195875-11195875 on Build GRCh38
rs567852782 Chr.1: 11195562-11195562 on Build GRCh38
rs984920269 Chr.1: 11195573-11195573 on Build GRCh38
rs946800795 Chr.1: 11195647-11195647 on Build GRCh38
rs773607263 Chr.1: 11195666-11195666 on Build GRCh38
rs547930906 Chr.1: 11195939-11195939 on Build GRCh38
rs532647913 Chr.1: 11195589-11195589 on Build GRCh38
rs993784437 Chr.1: 11195730-11195730 on Build GRCh38
rs370047912 Chr.1: 11195690-11195690 on Build GRCh38
rs907038685 Chr.1: 11195805-11195805 on Build GRCh38
rs774722951 Chr.1: 11195626-11195626 on Build GRCh38
rs938630865 Chr.1: 11195612-11195612 on Build GRCh38
rs1006742569 Chr.1: 11195707-11195708 on Build GRCh38
rs536751630 Chr.1: 11195700-11195700 on Build GRCh38
rs1003153055 Chr.1: 11195808-11195808 on Build GRCh38
rs774862855 Chr.1: 11195815-11195815 on Build GRCh38
rs1022720854 Chr.1: 11195801-11195801 on Build GRCh38
rs928976187 Chr.1: 11195752-11195752 on Build GRCh38

View Full Product Details