Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 51251710-51252210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM124A
Entrez Gene ID
220108
Gene Name
family with sequence similarity 124 member A
Gene Aliases
-
UniGene
Hs.71913
Gene Chromosome Location
Chr.13: 51222328-51284241 on Build GRCh38
Transcripts
NM_001242312.1 NM_145019.3 XM_011534978.2 XM_017020419.1 XM_017020420.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
432477 c.800C>T p.P267L Chr.13: 51252059-51252059 on Build GRCh38
7103479 c.595C>T p.R199W Chr.13: 51251854-51251854 on Build GRCh38
1322849 c.650A>G p.D217G Chr.13: 51251909-51251909 on Build GRCh38
4774802 c.832C>T p.R278* Chr.13: 51252091-51252091 on Build GRCh38
6658294 c.596G>A p.R199Q Chr.13: 51251855-51251855 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054359983 Chr.13: 51251950-51251950 on Build GRCh38
rs762657108 Chr.13: 51251931-51251931 on Build GRCh38
rs779427558 Chr.13: 51252001-51252001 on Build GRCh38
rs778046298 Chr.13: 51252145-51252145 on Build GRCh38
rs774644447 Chr.13: 51252042-51252043 on Build GRCh38
rs1036911564 Chr.13: 51251742-51251742 on Build GRCh38
rs147514046 Chr.13: 51251827-51251827 on Build GRCh38
rs532318094 Chr.13: 51252159-51252159 on Build GRCh38
rs749227482 Chr.13: 51252180-51252180 on Build GRCh38
rs753787657 Chr.13: 51251752-51251752 on Build GRCh38
rs757521370 Chr.13: 51251778-51251778 on Build GRCh38
rs767755117 Chr.13: 51252070-51252070 on Build GRCh38
rs865840030 Chr.13: 51251801-51251801 on Build GRCh38
rs781381339 Chr.13: 51251782-51251782 on Build GRCh38
rs776374066 Chr.13: 51252060-51252060 on Build GRCh38
rs747422390 Chr.13: 51252148-51252148 on Build GRCh38
rs754022683 Chr.13: 51252121-51252121 on Build GRCh38
rs762933048 Chr.13: 51252092-51252092 on Build GRCh38
rs779303170 Chr.13: 51251892-51251892 on Build GRCh38
rs541072385 Chr.13: 51251884-51251884 on Build GRCh38
rs1039496456 Chr.13: 51251881-51251881 on Build GRCh38
rs371907248 Chr.13: 51251957-51251957 on Build GRCh38
rs756130112 Chr.13: 51251736-51251736 on Build GRCh38
rs763860004 Chr.13: 51252103-51252103 on Build GRCh38
rs948468046 Chr.13: 51251938-51251938 on Build GRCh38
rs139061272 Chr.13: 51252064-51252064 on Build GRCh38
rs1022248860 Chr.13: 51251883-51251883 on Build GRCh38
rs933950921 Chr.13: 51252079-51252079 on Build GRCh38
rs998404432 Chr.13: 51251773-51251773 on Build GRCh38
rs543244580 Chr.13: 51252027-51252027 on Build GRCh38
rs371062746 Chr.13: 51252150-51252150 on Build GRCh38
rs750587172 Chr.13: 51251867-51251867 on Build GRCh38
rs758941298 Chr.13: 51252143-51252143 on Build GRCh38
rs746678937 Chr.13: 51252054-51252054 on Build GRCh38
rs775058541 Chr.13: 51252167-51252167 on Build GRCh38
rs765161763 Chr.13: 51251848-51251848 on Build GRCh38
rs769359173 Chr.13: 51252066-51252066 on Build GRCh38
rs200916097 Chr.13: 51251841-51251841 on Build GRCh38
rs750666641 Chr.13: 51251956-51251956 on Build GRCh38
rs765243905 Chr.13: 51252138-51252138 on Build GRCh38
rs766488733 Chr.13: 51251961-51251961 on Build GRCh38
rs759540288 Chr.13: 51251846-51251846 on Build GRCh38
rs17075482 Chr.13: 51251908-51251908 on Build GRCh38
rs781296689 Chr.13: 51251859-51251859 on Build GRCh38
rs910803588 Chr.13: 51252156-51252156 on Build GRCh38
rs774168466 Chr.13: 51252109-51252109 on Build GRCh38
rs774696456 Chr.13: 51251822-51251822 on Build GRCh38
rs752517632 Chr.13: 51251849-51251849 on Build GRCh38
rs749873706 Chr.13: 51251735-51251735 on Build GRCh38
rs772443360 Chr.13: 51251897-51251897 on Build GRCh38
rs909636297 Chr.13: 51252010-51252010 on Build GRCh38
rs201937112 Chr.13: 51251949-51251949 on Build GRCh38
rs767125700 Chr.13: 51251729-51251729 on Build GRCh38
rs763889748 Chr.13: 51251951-51251951 on Build GRCh38
rs202058194 Chr.13: 51252020-51252020 on Build GRCh38
rs151116075 Chr.13: 51251923-51251923 on Build GRCh38
rs769122655 Chr.13: 51251885-51251885 on Build GRCh38
rs140040852 Chr.13: 51251838-51251838 on Build GRCh38
rs149874206 Chr.13: 51252069-51252069 on Build GRCh38
rs778187669 Chr.13: 51252047-51252047 on Build GRCh38
rs961008632 Chr.13: 51251909-51251909 on Build GRCh38
rs756159316 Chr.13: 51251878-51251878 on Build GRCh38
rs770593683 Chr.13: 51252056-51252056 on Build GRCh38
rs868846047 Chr.13: 51252077-51252077 on Build GRCh38
rs755056126 Chr.13: 51252137-51252137 on Build GRCh38
rs769069526 Chr.13: 51251815-51251815 on Build GRCh38
rs538055450 Chr.13: 51251911-51251911 on Build GRCh38
rs971891532 Chr.13: 51252057-51252057 on Build GRCh38
rs749734714 Chr.13: 51251806-51251806 on Build GRCh38
rs760858484 Chr.13: 51252114-51252114 on Build GRCh38
rs780427047 Chr.13: 51251802-51251802 on Build GRCh38
rs751895993 Chr.13: 51251775-51251775 on Build GRCh38
rs374624217 Chr.13: 51252158-51252158 on Build GRCh38
rs776248339 Chr.13: 51251907-51251907 on Build GRCh38
rs778836299 Chr.13: 51251774-51251774 on Build GRCh38
rs765850527 Chr.13: 51251840-51251840 on Build GRCh38
rs769483830 Chr.13: 51252165-51252165 on Build GRCh38
rs766249863 Chr.13: 51251737-51251737 on Build GRCh38
rs370492598 Chr.13: 51251893-51251893 on Build GRCh38
rs34899851 Chr.13: 51251902-51251902 on Build GRCh38
rs897083351 Chr.13: 51251766-51251766 on Build GRCh38
rs753204739 Chr.13: 51252141-51252141 on Build GRCh38
rs560839719 Chr.13: 51251903-51251903 on Build GRCh38
rs745952960 Chr.13: 51251788-51251788 on Build GRCh38
rs769874180 Chr.13: 51251797-51251797 on Build GRCh38
rs754827186 Chr.13: 51251767-51251767 on Build GRCh38
rs774406822 Chr.13: 51251991-51251991 on Build GRCh38
rs758162138 Chr.13: 51251854-51251854 on Build GRCh38
rs771605063 Chr.13: 51251828-51251828 on Build GRCh38
rs902993948 Chr.13: 51251928-51251928 on Build GRCh38
rs146463367 Chr.13: 51252115-51252115 on Build GRCh38
rs771436460 Chr.13: 51251891-51251891 on Build GRCh38
rs762645741 Chr.13: 51251786-51251787 on Build GRCh38
rs773031638 Chr.13: 51251785-51251785 on Build GRCh38
rs772872428 Chr.13: 51251835-51251835 on Build GRCh38

View Full Product Details