Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 99255025-99255527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR18
Entrez Gene ID
2841
Gene Name
G protein-coupled receptor 18
Gene Aliases
-
UniGene
Hs.741589
Gene Chromosome Location
Chr.13: 99254713-99262498 on Build GRCh38
Transcripts
NM_001098200.1 NM_005292.3 XM_006719946.3 NM_001144072.1 NM_177967.3 NR_026644.1 XM_006719948.3 XM_011521082.2 XM_011521083.2 XM_011521084.2

Gene Symbol
UBAC2
Entrez Gene ID
337867
Gene Name
UBA domain containing 2
Gene Aliases
PHGDHL1
UniGene
Hs.508545
Gene Chromosome Location
Chr.13: 99200425-99386499 on Build GRCh38
Transcripts
NM_001098200.1 NM_005292.3 XM_006719946.3 NM_001144072.1 NM_177967.3 NR_026644.1 XM_006719948.3 XM_011521082.2 XM_011521083.2 XM_011521084.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6674003 c.727G>A p.V243I Chr.13: 99255146-99255146 on Build GRCh38
4614731 c.587delT p.F196fs*2 Chr.13: 99255286-99255286 on Build GRCh38
6674002 c.391G>T p.E131* Chr.13: 99255482-99255482 on Build GRCh38
6450574 c.556G>A p.V186M Chr.13: 99255317-99255317 on Build GRCh38
3704750 c.421G>A p.A141T Chr.13: 99255452-99255452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374331780 Chr.13: 99255266-99255266 on Build GRCh38
rs754746735 Chr.13: 99255216-99255216 on Build GRCh38
rs367651389 Chr.13: 99255489-99255489 on Build GRCh38
rs376476131 Chr.13: 99255418-99255418 on Build GRCh38
rs143171650 Chr.13: 99255468-99255468 on Build GRCh38
rs759661276 Chr.13: 99255473-99255473 on Build GRCh38
rs749743204 Chr.13: 99255357-99255357 on Build GRCh38
rs760766191 Chr.13: 99255395-99255395 on Build GRCh38
rs746564668 Chr.13: 99255334-99255334 on Build GRCh38
rs146690022 Chr.13: 99255236-99255236 on Build GRCh38
rs748201402 Chr.13: 99255286-99255286 on Build GRCh38
rs750387704 Chr.13: 99255172-99255172 on Build GRCh38
rs776554722 Chr.13: 99255412-99255412 on Build GRCh38
rs780958175 Chr.13: 99255059-99255059 on Build GRCh38
rs986477803 Chr.13: 99255308-99255308 on Build GRCh38
rs755502855 Chr.13: 99255452-99255452 on Build GRCh38
rs776777158 Chr.13: 99255451-99255451 on Build GRCh38
rs1026790669 Chr.13: 99255431-99255431 on Build GRCh38
rs752505629 Chr.13: 99255419-99255419 on Build GRCh38
rs1007690420 Chr.13: 99255056-99255056 on Build GRCh38
rs769968108 Chr.13: 99255408-99255409 on Build GRCh38
rs374546166 Chr.13: 99255504-99255504 on Build GRCh38
rs768365665 Chr.13: 99255112-99255112 on Build GRCh38
rs747716613 Chr.13: 99255222-99255222 on Build GRCh38
rs779442891 Chr.13: 99255456-99255456 on Build GRCh38
rs764458382 Chr.13: 99255147-99255147 on Build GRCh38
rs148795491 Chr.13: 99255104-99255104 on Build GRCh38
rs774635906 Chr.13: 99255269-99255269 on Build GRCh38
rs151309537 Chr.13: 99255474-99255474 on Build GRCh38
rs372413471 Chr.13: 99255132-99255132 on Build GRCh38
rs780790126 Chr.13: 99255449-99255449 on Build GRCh38
rs369995186 Chr.13: 99255182-99255182 on Build GRCh38
rs138108915 Chr.13: 99255365-99255365 on Build GRCh38
rs543149825 Chr.13: 99255354-99255354 on Build GRCh38
rs752412207 Chr.13: 99255317-99255317 on Build GRCh38
rs758638842 Chr.13: 99255092-99255092 on Build GRCh38
rs773462379 Chr.13: 99255393-99255393 on Build GRCh38
rs774634231 Chr.13: 99255366-99255366 on Build GRCh38
rs533174058 Chr.13: 99255294-99255294 on Build GRCh38
rs758186651 Chr.13: 99255426-99255426 on Build GRCh38
rs746424593 Chr.13: 99255295-99255295 on Build GRCh38
rs540457420 Chr.13: 99255077-99255077 on Build GRCh38
rs185252466 Chr.13: 99255442-99255442 on Build GRCh38
rs745721043 Chr.13: 99255063-99255063 on Build GRCh38
rs773963673 Chr.13: 99255116-99255116 on Build GRCh38
rs781694906 Chr.13: 99255074-99255075 on Build GRCh38
rs140769802 Chr.13: 99255146-99255146 on Build GRCh38
rs766408447 Chr.13: 99255405-99255405 on Build GRCh38
rs201584591 Chr.13: 99255082-99255082 on Build GRCh38
rs142437132 Chr.13: 99255106-99255106 on Build GRCh38
rs776752710 Chr.13: 99255469-99255469 on Build GRCh38
rs755906704 Chr.13: 99255103-99255103 on Build GRCh38
rs150932589 Chr.13: 99255136-99255136 on Build GRCh38
rs748873391 Chr.13: 99255071-99255071 on Build GRCh38
rs562575938 Chr.13: 99255284-99255284 on Build GRCh38
rs757027038 Chr.13: 99255498-99255498 on Build GRCh38
rs549443596 Chr.13: 99255443-99255443 on Build GRCh38
rs772222254 Chr.13: 99255386-99255386 on Build GRCh38
rs997590727 Chr.13: 99255430-99255430 on Build GRCh38
rs777481240 Chr.13: 99255333-99255333 on Build GRCh38
rs776115198 Chr.13: 99255234-99255234 on Build GRCh38
rs149113944 Chr.13: 99255458-99255458 on Build GRCh38
rs779593606 Chr.13: 99255228-99255228 on Build GRCh38
rs140120491 Chr.13: 99255061-99255061 on Build GRCh38
rs771435262 Chr.13: 99255125-99255125 on Build GRCh38
rs1038270775 Chr.13: 99255486-99255486 on Build GRCh38
rs1002833847 Chr.13: 99255153-99255153 on Build GRCh38
rs765661875 Chr.13: 99255084-99255084 on Build GRCh38
rs775154268 Chr.13: 99255070-99255070 on Build GRCh38
rs145449053 Chr.13: 99255318-99255318 on Build GRCh38
rs775445604 Chr.13: 99255477-99255477 on Build GRCh38
rs3742130 Chr.13: 99255087-99255087 on Build GRCh38
rs773906701 Chr.13: 99255078-99255078 on Build GRCh38
rs766951793 Chr.13: 99255079-99255079 on Build GRCh38
rs41279138 Chr.13: 99255314-99255314 on Build GRCh38
rs143283188 Chr.13: 99255224-99255224 on Build GRCh38
rs370331461 Chr.13: 99255446-99255446 on Build GRCh38
rs374299759 Chr.13: 99255459-99255459 on Build GRCh38
rs199762287 Chr.13: 99255140-99255140 on Build GRCh38
rs769423816 Chr.13: 99255067-99255067 on Build GRCh38
rs761980710 Chr.13: 99255166-99255166 on Build GRCh38
rs955035338 Chr.13: 99255302-99255302 on Build GRCh38
rs777229237 Chr.13: 99255127-99255127 on Build GRCh38
rs769097676 Chr.13: 99255246-99255246 on Build GRCh38
rs757646007 Chr.13: 99255117-99255117 on Build GRCh38
rs768167198 Chr.13: 99255075-99255075 on Build GRCh38
rs111938999 Chr.13: 99255500-99255500 on Build GRCh38
rs201170460 Chr.13: 99255225-99255225 on Build GRCh38
rs61970277 Chr.13: 99255465-99255465 on Build GRCh38
rs377064623 Chr.13: 99255301-99255301 on Build GRCh38
rs745356452 Chr.13: 99255450-99255450 on Build GRCh38
rs748541807 Chr.13: 99255380-99255380 on Build GRCh38
rs376966351 Chr.13: 99255165-99255165 on Build GRCh38
rs376159827 Chr.13: 99255467-99255467 on Build GRCh38
rs143074994 Chr.13: 99255096-99255096 on Build GRCh38
rs984822888 Chr.13: 99255121-99255121 on Build GRCh38
rs766355278 Chr.13: 99255298-99255298 on Build GRCh38
rs767428640 Chr.13: 99255171-99255171 on Build GRCh38
rs778649813 Chr.13: 99255217-99255217 on Build GRCh38
rs762683808 Chr.13: 99255488-99255488 on Build GRCh38
rs770514440 Chr.13: 99255344-99255344 on Build GRCh38
rs761883130 Chr.13: 99255271-99255271 on Build GRCh38
rs193218266 Chr.13: 99255097-99255097 on Build GRCh38

View Full Product Details