Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 105490767-105491281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAOA
Entrez Gene ID
267012
Gene Name
D-amino acid oxidase activator
Gene Aliases
LG72, SG72
UniGene
Hs.381382
Gene Chromosome Location
Chr.13: 105465867-105491034 on Build GRCh38
Transcripts
NM_001161812.1 NM_001161814.1 NM_172370.4 XM_005254042.1 NR_040247.1

Gene Symbol
DAOA-AS1
Entrez Gene ID
282706
Gene Name
DAOA antisense RNA 1
Gene Aliases
DAOA-AS, DAOAAS, G30
UniGene
Hs.558713
Gene Chromosome Location
Chr.13: 105459055-105505681 on Build GRCh38
Transcripts
NM_001161812.1 NM_001161814.1 NM_172370.4 XM_005254042.1 NR_040247.1

SNP Detail

SNP ID Chromosome Location
rs72549500 Chr.13: 105490883-105490883 on Build GRCh38
rs1004940341 Chr.13: 105491129-105491129 on Build GRCh38
rs983838900 Chr.13: 105490873-105490873 on Build GRCh38
rs142171574 Chr.13: 105491112-105491112 on Build GRCh38
rs1003754056 Chr.13: 105491069-105491069 on Build GRCh38
rs907818274 Chr.13: 105490877-105490877 on Build GRCh38
rs995039446 Chr.13: 105490885-105490885 on Build GRCh38
rs72549502 Chr.13: 105490905-105490905 on Build GRCh38
rs921846184 Chr.13: 105491220-105491220 on Build GRCh38
rs957214787 Chr.13: 105490968-105490968 on Build GRCh38
rs111698782 Chr.13: 105490867-105490867 on Build GRCh38
rs945408108 Chr.13: 105490860-105490860 on Build GRCh38
rs1012825261 Chr.13: 105491097-105491097 on Build GRCh38
rs895349768 Chr.13: 105491093-105491093 on Build GRCh38
rs181957413 Chr.13: 105490895-105490895 on Build GRCh38
rs41275046 Chr.13: 105490811-105490811 on Build GRCh38
rs879758302 Chr.13: 105490866-105490867 on Build GRCh38
rs890501353 Chr.13: 105490867-105490867 on Build GRCh38
rs747796855 Chr.13: 105491133-105491133 on Build GRCh38
rs965993967 Chr.13: 105491155-105491155 on Build GRCh38
rs1039155521 Chr.13: 105490880-105490882 on Build GRCh38
rs9558573 Chr.13: 105490825-105490825 on Build GRCh38
rs976003548 Chr.13: 105491166-105491166 on Build GRCh38
rs562662859 Chr.13: 105491211-105491211 on Build GRCh38
rs190672384 Chr.13: 105491148-105491148 on Build GRCh38
rs150759542 Chr.13: 105491219-105491219 on Build GRCh38
rs563929988 Chr.13: 105490981-105490981 on Build GRCh38
rs990926994 Chr.13: 105490829-105490829 on Build GRCh38
rs1056608210 Chr.13: 105491081-105491081 on Build GRCh38
rs375413207 Chr.13: 105491164-105491167 on Build GRCh38
rs886657719 Chr.13: 105491163-105491166 on Build GRCh38
rs56158118 Chr.13: 105490933-105490933 on Build GRCh38
rs76798244 Chr.13: 105491000-105491000 on Build GRCh38
rs72549499 Chr.13: 105490833-105490833 on Build GRCh38
rs1026549318 Chr.13: 105490888-105490888 on Build GRCh38
rs913884562 Chr.13: 105490849-105490849 on Build GRCh38
rs72549501 Chr.13: 105490890-105490890 on Build GRCh38
rs1039124505 Chr.13: 105491098-105491099 on Build GRCh38
rs952214517 Chr.13: 105491120-105491120 on Build GRCh38
rs944616932 Chr.13: 105490887-105490887 on Build GRCh38
rs1041377976 Chr.13: 105491032-105491032 on Build GRCh38
rs148681984 Chr.13: 105491007-105491007 on Build GRCh38
rs953262676 Chr.13: 105491254-105491254 on Build GRCh38
rs576722731 Chr.13: 105490891-105490891 on Build GRCh38
rs75311878 Chr.13: 105490912-105490912 on Build GRCh38
rs186147829 Chr.13: 105491055-105491055 on Build GRCh38
rs71432853 Chr.13: 105491161-105491161 on Build GRCh38
rs959766831 Chr.13: 105490826-105490826 on Build GRCh38
rs979892273 Chr.13: 105490796-105490796 on Build GRCh38
rs1024360556 Chr.13: 105491201-105491201 on Build GRCh38
rs529003637 Chr.13: 105491228-105491228 on Build GRCh38
rs151094102 Chr.13: 105490877-105490877 on Build GRCh38
rs911916688 Chr.13: 105491087-105491087 on Build GRCh38
rs879706884 Chr.13: 105490917-105490917 on Build GRCh38
rs970186812 Chr.13: 105490914-105490914 on Build GRCh38
rs1047970098 Chr.13: 105491160-105491162 on Build GRCh38
rs913173880 Chr.13: 105491025-105491025 on Build GRCh38

View Full Product Details