Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 22974688-22975207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AJUBA
Entrez Gene ID
84962
Gene Name
ajuba LIM protein
Gene Aliases
JUB
UniGene
Hs.655832
Gene Chromosome Location
Chr.14: 22971174-22982642 on Build GRCh38
Transcripts
NM_032876.5 NM_198086.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3793578 c.1270C>A p.P424T Chr.14: 22975074-22975074 on Build GRCh38
2030746 c.1400G>A p.G467D Chr.14: 22974861-22974861 on Build GRCh38
4878786 c.1298A>G p.N433S Chr.14: 22975046-22975046 on Build GRCh38
6055934 c.1298A>T p.N433I Chr.14: 22975046-22975046 on Build GRCh38
3401236 c.1256G>A p.G419E Chr.14: 22975088-22975088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760834742 Chr.14: 22975007-22975007 on Build GRCh38
rs1009956245 Chr.14: 22974792-22974792 on Build GRCh38
rs755857152 Chr.14: 22975182-22975182 on Build GRCh38
rs61737805 Chr.14: 22974875-22974875 on Build GRCh38
rs535973946 Chr.14: 22974728-22974728 on Build GRCh38
rs755860715 Chr.14: 22974966-22974966 on Build GRCh38
rs550809065 Chr.14: 22975168-22975168 on Build GRCh38
rs753388680 Chr.14: 22975094-22975094 on Build GRCh38
rs370899035 Chr.14: 22974853-22974853 on Build GRCh38
rs776943065 Chr.14: 22975137-22975137 on Build GRCh38
rs898240820 Chr.14: 22974733-22974733 on Build GRCh38
rs751432681 Chr.14: 22974924-22974924 on Build GRCh38
rs918132280 Chr.14: 22974713-22974713 on Build GRCh38
rs761023349 Chr.14: 22974885-22974885 on Build GRCh38
rs890824851 Chr.14: 22974761-22974761 on Build GRCh38
rs926701734 Chr.14: 22974731-22974731 on Build GRCh38
rs767437060 Chr.14: 22975062-22975062 on Build GRCh38
rs772775547 Chr.14: 22975131-22975131 on Build GRCh38
rs749609130 Chr.14: 22974820-22974820 on Build GRCh38
rs777424549 Chr.14: 22974807-22974807 on Build GRCh38
rs200247985 Chr.14: 22975046-22975046 on Build GRCh38
rs771132963 Chr.14: 22974985-22974985 on Build GRCh38
rs780696275 Chr.14: 22975133-22975133 on Build GRCh38
rs772233650 Chr.14: 22974868-22974868 on Build GRCh38
rs752261469 Chr.14: 22975074-22975074 on Build GRCh38
rs949595325 Chr.14: 22974721-22974721 on Build GRCh38
rs182964081 Chr.14: 22974810-22974810 on Build GRCh38
rs116945987 Chr.14: 22974950-22974950 on Build GRCh38
rs138877420 Chr.14: 22975054-22975054 on Build GRCh38
rs996701686 Chr.14: 22974923-22974923 on Build GRCh38
rs746323876 Chr.14: 22974997-22974997 on Build GRCh38
rs141770329 Chr.14: 22975092-22975092 on Build GRCh38
rs776663237 Chr.14: 22975021-22975021 on Build GRCh38
rs772681812 Chr.14: 22975006-22975006 on Build GRCh38
rs748655988 Chr.14: 22975135-22975135 on Build GRCh38
rs780850472 Chr.14: 22974965-22974965 on Build GRCh38
rs749676873 Chr.14: 22974984-22974984 on Build GRCh38
rs937626353 Chr.14: 22975176-22975176 on Build GRCh38
rs957424116 Chr.14: 22975050-22975050 on Build GRCh38
rs202181054 Chr.14: 22974846-22974846 on Build GRCh38
rs927513465 Chr.14: 22975173-22975173 on Build GRCh38
rs779502019 Chr.14: 22974990-22974990 on Build GRCh38
rs768984164 Chr.14: 22975019-22975019 on Build GRCh38
rs567218913 Chr.14: 22974712-22974712 on Build GRCh38
rs780826685 Chr.14: 22974788-22974788 on Build GRCh38
rs952504637 Chr.14: 22974938-22974938 on Build GRCh38
rs774173728 Chr.14: 22975150-22975150 on Build GRCh38
rs770167601 Chr.14: 22974814-22974814 on Build GRCh38
rs754514221 Chr.14: 22974932-22974932 on Build GRCh38
rs530065086 Chr.14: 22974964-22974964 on Build GRCh38
rs746201246 Chr.14: 22975126-22975126 on Build GRCh38
rs936730631 Chr.14: 22974740-22974740 on Build GRCh38
rs201332657 Chr.14: 22974974-22974974 on Build GRCh38
rs199808733 Chr.14: 22975138-22975138 on Build GRCh38
rs899752385 Chr.14: 22974899-22974899 on Build GRCh38
rs867632173 Chr.14: 22975026-22975026 on Build GRCh38
rs892459974 Chr.14: 22974779-22974779 on Build GRCh38
rs201314137 Chr.14: 22975136-22975136 on Build GRCh38
rs777485704 Chr.14: 22975093-22975093 on Build GRCh38
rs773664731 Chr.14: 22974818-22974818 on Build GRCh38
rs767253576 Chr.14: 22975084-22975084 on Build GRCh38
rs941721 Chr.14: 22974753-22974753 on Build GRCh38
rs146869528 Chr.14: 22975099-22975099 on Build GRCh38
rs750301889 Chr.14: 22974902-22974902 on Build GRCh38
rs544864542 Chr.14: 22974905-22974905 on Build GRCh38
rs771525807 Chr.14: 22974824-22974824 on Build GRCh38
rs1018180963 Chr.14: 22974879-22974879 on Build GRCh38
rs10130692 Chr.14: 22975038-22975038 on Build GRCh38
rs779466558 Chr.14: 22975107-22975107 on Build GRCh38
rs766356982 Chr.14: 22974907-22974907 on Build GRCh38
rs201186974 Chr.14: 22975145-22975145 on Build GRCh38
rs575989495 Chr.14: 22974897-22974897 on Build GRCh38
rs774662572 Chr.14: 22974829-22974829 on Build GRCh38

View Full Product Details