Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 28766766-28767301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXG1
Entrez Gene ID
2290
Gene Name
forkhead box G1
Gene Aliases
BF1, BF2, FHKL3, FKH2, FKHL1, FKHL2, FKHL3, FKHL4, FOXG1A, FOXG1B, FOXG1C, HBF-1, HBF-2, HBF-3, HBF-G2, HBF2, HFK1, HFK2, HFK3, KHL2, QIN
UniGene
Hs.632336
Gene Chromosome Location
Chr.14: 28767072-28770277 on Build GRCh38
Transcripts
NM_005249.4

SNP Detail

SNP ID Chromosome Location
rs769086783 Chr.14: 28767273-28767273 on Build GRCh38
rs895434226 Chr.14: 28766922-28766922 on Build GRCh38
rs1012325759 Chr.14: 28766945-28766945 on Build GRCh38
rs963193232 Chr.14: 28766994-28766994 on Build GRCh38
rs908571005 Chr.14: 28767081-28767081 on Build GRCh38
rs542787885 Chr.14: 28767056-28767056 on Build GRCh38
rs902667581 Chr.14: 28766878-28766878 on Build GRCh38
rs1049788545 Chr.14: 28766834-28766834 on Build GRCh38
rs1032996112 Chr.14: 28766915-28766915 on Build GRCh38
rs1042435752 Chr.14: 28767174-28767174 on Build GRCh38
rs775779501 Chr.14: 28767264-28767264 on Build GRCh38
rs924058079 Chr.14: 28767202-28767202 on Build GRCh38
rs1038604992 Chr.14: 28767201-28767201 on Build GRCh38
rs980839255 Chr.14: 28766961-28766961 on Build GRCh38
rs973709251 Chr.14: 28766804-28766804 on Build GRCh38
rs931878445 Chr.14: 28767056-28767056 on Build GRCh38
rs987801293 Chr.14: 28767126-28767127 on Build GRCh38
rs912533428 Chr.14: 28767136-28767136 on Build GRCh38
rs921900383 Chr.14: 28766824-28766824 on Build GRCh38
rs932636413 Chr.14: 28766826-28766826 on Build GRCh38
rs992554630 Chr.14: 28767071-28767071 on Build GRCh38
rs960854074 Chr.14: 28767038-28767038 on Build GRCh38
rs921839117 Chr.14: 28767035-28767035 on Build GRCh38
rs866160184 Chr.14: 28767245-28767245 on Build GRCh38
rs1014703835 Chr.14: 28766982-28766982 on Build GRCh38
rs772479996 Chr.14: 28767251-28767251 on Build GRCh38
rs531432688 Chr.14: 28766789-28766789 on Build GRCh38
rs940251243 Chr.14: 28767086-28767086 on Build GRCh38
rs970391112 Chr.14: 28766967-28766967 on Build GRCh38
rs1024607088 Chr.14: 28766962-28766962 on Build GRCh38
rs12892291 Chr.14: 28766943-28766943 on Build GRCh38
rs12893487 Chr.14: 28766862-28766862 on Build GRCh38
rs1001013377 Chr.14: 28766894-28766894 on Build GRCh38
rs944059796 Chr.14: 28767141-28767141 on Build GRCh38
rs891121117 Chr.14: 28766843-28766843 on Build GRCh38
rs944089610 Chr.14: 28766875-28766875 on Build GRCh38
rs936827884 Chr.14: 28767207-28767207 on Build GRCh38
rs563015547 Chr.14: 28767056-28767057 on Build GRCh38
rs746165270 Chr.14: 28767235-28767235 on Build GRCh38
rs926697406 Chr.14: 28767027-28767027 on Build GRCh38
rs1042463327 Chr.14: 28766876-28766876 on Build GRCh38
rs973292843 Chr.14: 28767011-28767011 on Build GRCh38
rs751050823 Chr.14: 28767267-28767267 on Build GRCh38
rs747532272 Chr.14: 28767266-28767266 on Build GRCh38
rs59954935 Chr.14: 28766942-28766943 on Build GRCh38
rs983171803 Chr.14: 28766969-28766969 on Build GRCh38

View Full Product Details