Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 39919754-39920255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR176
Entrez Gene ID
11245
Gene Name
G protein-coupled receptor 176
Gene Aliases
HB-954
UniGene
Hs.37196
Gene Chromosome Location
Chr.15: 39795049-39920933 on Build GRCh38
Transcripts
NM_001271854.1 NM_007223.2 XM_011521168.2 XM_017021874.1 XM_017021875.1 XM_017021878.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5520224 c.91C>A p.L31I Chr.15: 39919936-39919936 on Build GRCh38
1239014 c.148G>A p.V50I Chr.15: 39919879-39919879 on Build GRCh38
7115913 c.93C>T p.L31L Chr.15: 39919934-39919934 on Build GRCh38
6340185 c.50A>T p.N17I Chr.15: 39919977-39919977 on Build GRCh38
7421265 c.71C>A p.A24E Chr.15: 39919956-39919956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937453624 Chr.15: 39920077-39920077 on Build GRCh38
rs751993655 Chr.15: 39920026-39920026 on Build GRCh38
rs974855777 Chr.15: 39919957-39919957 on Build GRCh38
rs772002590 Chr.15: 39919835-39919835 on Build GRCh38
rs372442129 Chr.15: 39920003-39920003 on Build GRCh38
rs758785401 Chr.15: 39919815-39919815 on Build GRCh38
rs1047296907 Chr.15: 39920164-39920164 on Build GRCh38
rs573815936 Chr.15: 39919998-39919998 on Build GRCh38
rs77245879 Chr.15: 39919830-39919830 on Build GRCh38
rs746195881 Chr.15: 39919811-39919811 on Build GRCh38
rs778582940 Chr.15: 39919943-39919944 on Build GRCh38
rs34575072 Chr.15: 39919903-39919903 on Build GRCh38
rs769184865 Chr.15: 39919821-39919821 on Build GRCh38
rs758695723 Chr.15: 39919871-39919871 on Build GRCh38
rs951806109 Chr.15: 39919967-39919967 on Build GRCh38
rs867673240 Chr.15: 39919855-39919855 on Build GRCh38
rs141122559 Chr.15: 39919973-39919973 on Build GRCh38
rs903941884 Chr.15: 39919868-39919868 on Build GRCh38
rs907764044 Chr.15: 39919791-39919791 on Build GRCh38
rs80137374 Chr.15: 39919849-39919849 on Build GRCh38
rs114232197 Chr.15: 39920175-39920175 on Build GRCh38
rs761022184 Chr.15: 39919841-39919841 on Build GRCh38
rs555628598 Chr.15: 39919870-39919870 on Build GRCh38
rs748791306 Chr.15: 39919914-39919914 on Build GRCh38
rs1018527559 Chr.15: 39920160-39920160 on Build GRCh38
rs200109400 Chr.15: 39920001-39920001 on Build GRCh38
rs535515092 Chr.15: 39919861-39919861 on Build GRCh38
rs999627536 Chr.15: 39920085-39920085 on Build GRCh38
rs551779445 Chr.15: 39919788-39919788 on Build GRCh38
rs528676199 Chr.15: 39920091-39920091 on Build GRCh38
rs1019768608 Chr.15: 39919928-39919928 on Build GRCh38
rs928685032 Chr.15: 39920069-39920069 on Build GRCh38
rs1008460921 Chr.15: 39919890-39919890 on Build GRCh38
rs995766705 Chr.15: 39920182-39920182 on Build GRCh38
rs368646413 Chr.15: 39919936-39919936 on Build GRCh38
rs370363084 Chr.15: 39919807-39919807 on Build GRCh38
rs761894953 Chr.15: 39919990-39919990 on Build GRCh38
rs891283660 Chr.15: 39920097-39920097 on Build GRCh38
rs532793751 Chr.15: 39919820-39919820 on Build GRCh38
rs899552884 Chr.15: 39920200-39920200 on Build GRCh38
rs755837176 Chr.15: 39919885-39919885 on Build GRCh38
rs770941070 Chr.15: 39919826-39919826 on Build GRCh38
rs745626442 Chr.15: 39919945-39919945 on Build GRCh38
rs770370276 Chr.15: 39919924-39919924 on Build GRCh38
rs750941574 Chr.15: 39919863-39919863 on Build GRCh38
rs570476659 Chr.15: 39919816-39919816 on Build GRCh38
rs545538455 Chr.15: 39920108-39920108 on Build GRCh38
rs12912357 Chr.15: 39919784-39919784 on Build GRCh38
rs12912359 Chr.15: 39919789-39919789 on Build GRCh38
rs939296858 Chr.15: 39919792-39919792 on Build GRCh38
rs907733086 Chr.15: 39919985-39919985 on Build GRCh38
rs775785783 Chr.15: 39919839-39919839 on Build GRCh38
rs948040432 Chr.15: 39920018-39920018 on Build GRCh38
rs766705556 Chr.15: 39920020-39920020 on Build GRCh38
rs749157323 Chr.15: 39919825-39919825 on Build GRCh38
rs556899221 Chr.15: 39919976-39919976 on Build GRCh38
rs996617083 Chr.15: 39920217-39920217 on Build GRCh38
rs143700851 Chr.15: 39919883-39919883 on Build GRCh38
rs182145795 Chr.15: 39919959-39919959 on Build GRCh38
rs764060660 Chr.15: 39919845-39919845 on Build GRCh38
rs1027292322 Chr.15: 39920202-39920202 on Build GRCh38
rs778932190 Chr.15: 39919929-39919929 on Build GRCh38
rs559563401 Chr.15: 39920081-39920081 on Build GRCh38
rs771986026 Chr.15: 39919939-39919939 on Build GRCh38
rs761833974 Chr.15: 39919856-39919856 on Build GRCh38
rs763511693 Chr.15: 39920005-39920005 on Build GRCh38
rs951777012 Chr.15: 39920231-39920231 on Build GRCh38
rs148093550 Chr.15: 39919897-39919897 on Build GRCh38
rs377215097 Chr.15: 39919874-39919874 on Build GRCh38
rs1052543931 Chr.15: 39920092-39920092 on Build GRCh38
rs745754778 Chr.15: 39919817-39919817 on Build GRCh38

View Full Product Details