Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45136264-45136765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUOX1
Entrez Gene ID
53905
Gene Name
dual oxidase 1
Gene Aliases
LNOX1, NOXEF1, THOX1
UniGene
Hs.272813
Gene Chromosome Location
Chr.15: 45129994-45165578 on Build GRCh38
Transcripts
NM_017434.4 NM_175940.2 XM_011521681.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
108254 c.943G>A p.D315N Chr.15: 45136546-45136546 on Build GRCh38
4883177 c.933G>T p.R311R Chr.15: 45136536-45136536 on Build GRCh38
6898528 c.961G>A p.E321K Chr.15: 45136564-45136564 on Build GRCh38
5849261 c.973G>A p.A325T Chr.15: 45136576-45136576 on Build GRCh38
2189403 c.882G>C p.E294D Chr.15: 45136367-45136367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183712963 Chr.15: 45136660-45136660 on Build GRCh38
rs761963527 Chr.15: 45136543-45136543 on Build GRCh38
rs773764858 Chr.15: 45136321-45136321 on Build GRCh38
rs200255711 Chr.15: 45136404-45136404 on Build GRCh38
rs375568524 Chr.15: 45136421-45136421 on Build GRCh38
rs767305748 Chr.15: 45136615-45136615 on Build GRCh38
rs778890901 Chr.15: 45136303-45136303 on Build GRCh38
rs142038372 Chr.15: 45136573-45136573 on Build GRCh38
rs765702071 Chr.15: 45136319-45136319 on Build GRCh38
rs140943243 Chr.15: 45136363-45136363 on Build GRCh38
rs745694191 Chr.15: 45136304-45136304 on Build GRCh38
rs751450593 Chr.15: 45136450-45136450 on Build GRCh38
rs748214906 Chr.15: 45136300-45136300 on Build GRCh38
rs2912246 Chr.15: 45136301-45136301 on Build GRCh38
rs777077129 Chr.15: 45136666-45136666 on Build GRCh38
rs767675930 Chr.15: 45136403-45136403 on Build GRCh38
rs747850481 Chr.15: 45136490-45136490 on Build GRCh38
rs768445900 Chr.15: 45136367-45136367 on Build GRCh38
rs112464125 Chr.15: 45136318-45136318 on Build GRCh38
rs368973025 Chr.15: 45136668-45136668 on Build GRCh38
rs746675063 Chr.15: 45136308-45136308 on Build GRCh38
rs372507382 Chr.15: 45136610-45136610 on Build GRCh38
rs973434544 Chr.15: 45136460-45136460 on Build GRCh38
rs374076030 Chr.15: 45136736-45136736 on Build GRCh38
rs781016173 Chr.15: 45136514-45136516 on Build GRCh38
rs141086153 Chr.15: 45136467-45136467 on Build GRCh38
rs376769071 Chr.15: 45136515-45136515 on Build GRCh38
rs552352882 Chr.15: 45136599-45136599 on Build GRCh38
rs772473653 Chr.15: 45136524-45136524 on Build GRCh38
rs776963020 Chr.15: 45136608-45136608 on Build GRCh38
rs369783477 Chr.15: 45136456-45136456 on Build GRCh38
rs200968175 Chr.15: 45136315-45136315 on Build GRCh38
rs752581953 Chr.15: 45136618-45136618 on Build GRCh38
rs758441619 Chr.15: 45136424-45136424 on Build GRCh38
rs780093125 Chr.15: 45136445-45136445 on Build GRCh38
rs375799776 Chr.15: 45136637-45136637 on Build GRCh38
rs780764073 Chr.15: 45136458-45136458 on Build GRCh38
rs750363343 Chr.15: 45136347-45136347 on Build GRCh38
rs766460787 Chr.15: 45136545-45136545 on Build GRCh38
rs748742911 Chr.15: 45136513-45136513 on Build GRCh38
rs151146127 Chr.15: 45136596-45136596 on Build GRCh38
rs774571713 Chr.15: 45136584-45136584 on Build GRCh38
rs751528162 Chr.15: 45136548-45136548 on Build GRCh38
rs774718763 Chr.15: 45136401-45136401 on Build GRCh38
rs752962125 Chr.15: 45136407-45136407 on Build GRCh38
rs752765371 Chr.15: 45136342-45136342 on Build GRCh38
rs747442860 Chr.15: 45136658-45136658 on Build GRCh38
rs147597074 Chr.15: 45136537-45136537 on Build GRCh38
rs143051498 Chr.15: 45136578-45136578 on Build GRCh38
rs760729601 Chr.15: 45136419-45136419 on Build GRCh38
rs778787889 Chr.15: 45136345-45136345 on Build GRCh38
rs928166495 Chr.15: 45136519-45136519 on Build GRCh38
rs200172337 Chr.15: 45136305-45136305 on Build GRCh38
rs745334694 Chr.15: 45136645-45136645 on Build GRCh38
rs544840824 Chr.15: 45136609-45136609 on Build GRCh38
rs745753408 Chr.15: 45136529-45136529 on Build GRCh38
rs144786721 Chr.15: 45136400-45136400 on Build GRCh38
rs780404176 Chr.15: 45136651-45136651 on Build GRCh38
rs566622588 Chr.15: 45136589-45136589 on Build GRCh38
rs753993447 Chr.15: 45136539-45136539 on Build GRCh38
rs372501889 Chr.15: 45136331-45136331 on Build GRCh38
rs752393455 Chr.15: 45136574-45136574 on Build GRCh38
rs756191622 Chr.15: 45136343-45136343 on Build GRCh38
rs778403380 Chr.15: 45136641-45136641 on Build GRCh38
rs772537888 Chr.15: 45136378-45136378 on Build GRCh38
rs779972165 Chr.15: 45136640-45136640 on Build GRCh38
rs202050517 Chr.15: 45136307-45136307 on Build GRCh38
rs758967890 Chr.15: 45136647-45136647 on Build GRCh38
rs75150741 Chr.15: 45136566-45136566 on Build GRCh38
rs780876955 Chr.15: 45136372-45136372 on Build GRCh38
rs769765704 Chr.15: 45136634-45136634 on Build GRCh38
rs72724253 Chr.15: 45136514-45136514 on Build GRCh38
rs747950105 Chr.15: 45136606-45136606 on Build GRCh38
rs895438692 Chr.15: 45136570-45136570 on Build GRCh38
rs369560581 Chr.15: 45136626-45136626 on Build GRCh38
rs772395646 Chr.15: 45136595-45136595 on Build GRCh38
rs138184170 Chr.15: 45136728-45136728 on Build GRCh38
rs999245098 Chr.15: 45136587-45136587 on Build GRCh38
rs568932337 Chr.15: 45136607-45136607 on Build GRCh38
rs759548835 Chr.15: 45136614-45136614 on Build GRCh38
rs145858411 Chr.15: 45136546-45136546 on Build GRCh38
rs202146742 Chr.15: 45136536-45136536 on Build GRCh38
rs779457767 Chr.15: 45136593-45136593 on Build GRCh38
rs763454459 Chr.15: 45136398-45136398 on Build GRCh38
rs753795440 Chr.15: 45136422-45136422 on Build GRCh38
rs550963016 Chr.15: 45136516-45136516 on Build GRCh38
rs769333622 Chr.15: 45136375-45136375 on Build GRCh38
rs747678818 Chr.15: 45136374-45136374 on Build GRCh38
rs773137579 Chr.15: 45136633-45136634 on Build GRCh38
rs769262711 Chr.15: 45136491-45136491 on Build GRCh38
rs375449924 Chr.15: 45136621-45136621 on Build GRCh38
rs188427152 Chr.15: 45136677-45136677 on Build GRCh38
rs573527785 Chr.15: 45136360-45136360 on Build GRCh38
rs754814955 Chr.15: 45136457-45136457 on Build GRCh38
rs756693028 Chr.15: 45136293-45136293 on Build GRCh38
rs773359172 Chr.15: 45136613-45136613 on Build GRCh38
rs746859062 Chr.15: 45136700-45136700 on Build GRCh38
rs200305037 Chr.15: 45136395-45136395 on Build GRCh38
rs755221759 Chr.15: 45136332-45136332 on Build GRCh38
rs777369747 Chr.15: 45136511-45136511 on Build GRCh38
rs756758466 Chr.15: 45136590-45136590 on Build GRCh38
rs767674181 Chr.15: 45136340-45136340 on Build GRCh38

View Full Product Details