Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49644951-49645471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTWD1
Entrez Gene ID
56986
Gene Name
DTW domain containing 1
Gene Aliases
MDS009
UniGene
Hs.127432
Gene Chromosome Location
Chr.15: 49620980-49654448 on Build GRCh38
Transcripts
NM_001144955.1 NM_020234.5 XM_011521815.1 XM_017022419.1 XM_017022420.1 XM_017022421.1 XM_017022422.1 XM_017022425.1 XM_017022426.1 XM_017022427.1 XM_017022428.1 XM_017022429.1 XM_017022430.1 XM_017022431.1

SNP Detail

SNP ID Chromosome Location
rs1004955644 Chr.15: 49645368-49645368 on Build GRCh38
rs756256354 Chr.15: 49645393-49645393 on Build GRCh38
rs766720560 Chr.15: 49645052-49645052 on Build GRCh38
rs890028214 Chr.15: 49645149-49645149 on Build GRCh38
rs981331876 Chr.15: 49644982-49644982 on Build GRCh38
rs981785508 Chr.15: 49645131-49645131 on Build GRCh38
rs781758106 Chr.15: 49645218-49645218 on Build GRCh38
rs535033250 Chr.15: 49645214-49645214 on Build GRCh38
rs557527844 Chr.15: 49645402-49645402 on Build GRCh38
rs867023299 Chr.15: 49645203-49645203 on Build GRCh38
rs1030309411 Chr.15: 49645308-49645308 on Build GRCh38
rs1051319632 Chr.15: 49645148-49645148 on Build GRCh38
rs947775071 Chr.15: 49645057-49645057 on Build GRCh38
rs771750494 Chr.15: 49645406-49645406 on Build GRCh38
rs890418722 Chr.15: 49645342-49645342 on Build GRCh38
rs375622697 Chr.15: 49645269-49645269 on Build GRCh38
rs996358009 Chr.15: 49645295-49645295 on Build GRCh38
rs954807743 Chr.15: 49644987-49644987 on Build GRCh38
rs1025331996 Chr.15: 49645291-49645291 on Build GRCh38
rs983478389 Chr.15: 49645004-49645004 on Build GRCh38
rs761388554 Chr.15: 49645022-49645022 on Build GRCh38
rs753310538 Chr.15: 49645288-49645288 on Build GRCh38
rs570845015 Chr.15: 49645056-49645056 on Build GRCh38
rs566083565 Chr.15: 49645222-49645222 on Build GRCh38
rs1014958373 Chr.15: 49645422-49645422 on Build GRCh38
rs923922373 Chr.15: 49645087-49645087 on Build GRCh38
rs555590366 Chr.15: 49645406-49645406 on Build GRCh38
rs141827221 Chr.15: 49645360-49645360 on Build GRCh38
rs934259235 Chr.15: 49645100-49645100 on Build GRCh38
rs777753059 Chr.15: 49645038-49645038 on Build GRCh38
rs1002526890 Chr.15: 49645378-49645378 on Build GRCh38
rs935070391 Chr.15: 49645171-49645171 on Build GRCh38
rs1014716467 Chr.15: 49645008-49645008 on Build GRCh38
rs1052151428 Chr.15: 49645191-49645191 on Build GRCh38
rs751904864 Chr.15: 49645072-49645072 on Build GRCh38
rs748500024 Chr.15: 49645069-49645069 on Build GRCh38
rs915348997 Chr.15: 49645025-49645025 on Build GRCh38
rs927589718 Chr.15: 49645143-49645143 on Build GRCh38
rs553456814 Chr.15: 49645240-49645240 on Build GRCh38
rs755437481 Chr.15: 49645138-49645138 on Build GRCh38
rs182857092 Chr.15: 49645219-49645219 on Build GRCh38
rs1007510039 Chr.15: 49645169-49645169 on Build GRCh38
rs190296234 Chr.15: 49645031-49645031 on Build GRCh38
rs958815823 Chr.15: 49645014-49645014 on Build GRCh38
rs971022465 Chr.15: 49645314-49645314 on Build GRCh38
rs946812812 Chr.15: 49645048-49645048 on Build GRCh38
rs891999404 Chr.15: 49645204-49645204 on Build GRCh38
rs963264897 Chr.15: 49645428-49645428 on Build GRCh38

View Full Product Details