Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64381246-64381748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0101
Entrez Gene ID
9768
Gene Name
KIAA0101
Gene Aliases
L5, NS5ATP9, OEATC, OEATC-1, OEATC1, PAF, PAF15, p15(PAF), p15/PAF, p15PAF
UniGene
Hs.81892
Gene Chromosome Location
Chr.15: 64364994-64387687 on Build GRCh38
Transcripts
NM_001029989.2 NM_014736.5 NR_109934.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6692968 c.28C>A p.P10T Chr.15: 64381344-64381344 on Build GRCh38
4695133 c.7C>T p.R3W Chr.15: 64381365-64381365 on Build GRCh38
3503049 c.28C>T p.P10S Chr.15: 64381344-64381344 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs188268476 Chr.15: 64381318-64381318 on Build GRCh38
rs898492772 Chr.15: 64381436-64381436 on Build GRCh38
rs1056188678 Chr.15: 64381626-64381626 on Build GRCh38
rs540984263 Chr.15: 64381264-64381264 on Build GRCh38
rs995586926 Chr.15: 64381454-64381454 on Build GRCh38
rs895039750 Chr.15: 64381413-64381413 on Build GRCh38
rs11554314 Chr.15: 64381378-64381378 on Build GRCh38
rs537203416 Chr.15: 64381634-64381634 on Build GRCh38
rs774145325 Chr.15: 64381409-64381409 on Build GRCh38
rs987037573 Chr.15: 64381297-64381297 on Build GRCh38
rs749506910 Chr.15: 64381399-64381399 on Build GRCh38
rs549945260 Chr.15: 64381569-64381569 on Build GRCh38
rs956154853 Chr.15: 64381502-64381502 on Build GRCh38
rs1044161588 Chr.15: 64381320-64381320 on Build GRCh38
rs1028343204 Chr.15: 64381458-64381458 on Build GRCh38
rs947033077 Chr.15: 64381527-64381527 on Build GRCh38
rs372516219 Chr.15: 64381419-64381419 on Build GRCh38
rs938419661 Chr.15: 64381562-64381562 on Build GRCh38
rs755552882 Chr.15: 64381356-64381356 on Build GRCh38
rs927042309 Chr.15: 64381557-64381557 on Build GRCh38
rs750482897 Chr.15: 64381286-64381286 on Build GRCh38
rs748321101 Chr.15: 64381372-64381372 on Build GRCh38
rs117530341 Chr.15: 64381689-64381689 on Build GRCh38
rs547740889 Chr.15: 64381464-64381464 on Build GRCh38
rs879126183 Chr.15: 64381293-64381293 on Build GRCh38
rs967519358 Chr.15: 64381533-64381533 on Build GRCh38
rs541184581 Chr.15: 64381277-64381277 on Build GRCh38
rs938690837 Chr.15: 64381385-64381394 on Build GRCh38
rs938805495 Chr.15: 64381624-64381624 on Build GRCh38
rs763480597 Chr.15: 64381420-64381420 on Build GRCh38
rs375121530 Chr.15: 64381341-64381341 on Build GRCh38
rs898523776 Chr.15: 64381600-64381600 on Build GRCh38
rs11554318 Chr.15: 64381393-64381393 on Build GRCh38
rs1046969760 Chr.15: 64381703-64381703 on Build GRCh38
rs979877604 Chr.15: 64381542-64381542 on Build GRCh38
rs1041801482 Chr.15: 64381383-64381384 on Build GRCh38
rs201419596 Chr.15: 64381274-64381274 on Build GRCh38
rs763812337 Chr.15: 64381590-64381590 on Build GRCh38
rs11554308 Chr.15: 64381368-64381368 on Build GRCh38
rs758835769 Chr.15: 64381338-64381338 on Build GRCh38
rs376649666 Chr.15: 64381420-64381426 on Build GRCh38
rs573649374 Chr.15: 64381690-64381690 on Build GRCh38
rs1028253214 Chr.15: 64381631-64381631 on Build GRCh38
rs781658574 Chr.15: 64381384-64381385 on Build GRCh38
rs940298443 Chr.15: 64381586-64381586 on Build GRCh38
rs914275010 Chr.15: 64381513-64381513 on Build GRCh38
rs943004870 Chr.15: 64381305-64381305 on Build GRCh38
rs1038583655 Chr.15: 64381431-64381431 on Build GRCh38
rs770605302 Chr.15: 64381291-64381291 on Build GRCh38
rs890860393 Chr.15: 64381722-64381722 on Build GRCh38
rs898298774 Chr.15: 64381678-64381678 on Build GRCh38
rs1057036622 Chr.15: 64381565-64381565 on Build GRCh38
rs1015555479 Chr.15: 64381434-64381434 on Build GRCh38
rs552887085 Chr.15: 64381278-64381278 on Build GRCh38
rs987053529 Chr.15: 64381482-64381482 on Build GRCh38
rs928597808 Chr.15: 64381384-64381384 on Build GRCh38
rs563260971 Chr.15: 64381628-64381628 on Build GRCh38
rs946105199 Chr.15: 64381381-64381381 on Build GRCh38
rs780593782 Chr.15: 64381340-64381340 on Build GRCh38
rs889888429 Chr.15: 64381673-64381673 on Build GRCh38
rs1019874834 Chr.15: 64381721-64381721 on Build GRCh38
rs780306735 Chr.15: 64381289-64381289 on Build GRCh38
rs575977251 Chr.15: 64381618-64381618 on Build GRCh38
rs993879839 Chr.15: 64381449-64381449 on Build GRCh38
rs898351044 Chr.15: 64381437-64381437 on Build GRCh38
rs953934735 Chr.15: 64381465-64381465 on Build GRCh38
rs34325580 Chr.15: 64381412-64381413 on Build GRCh38
rs376862648 Chr.15: 64381353-64381353 on Build GRCh38
rs1056607258 Chr.15: 64381344-64381344 on Build GRCh38
rs928713775 Chr.15: 64381605-64381605 on Build GRCh38
rs202164043 Chr.15: 64381296-64381296 on Build GRCh38
rs116766191 Chr.15: 64381496-64381496 on Build GRCh38
rs11554317 Chr.15: 64381365-64381365 on Build GRCh38
rs369121099 Chr.15: 64381335-64381335 on Build GRCh38
rs1037240531 Chr.15: 64381591-64381591 on Build GRCh38
rs914716740 Chr.15: 64381529-64381529 on Build GRCh38
rs755528410 Chr.15: 64381435-64381435 on Build GRCh38
rs896748215 Chr.15: 64381403-64381403 on Build GRCh38
rs372935868 Chr.15: 64381337-64381337 on Build GRCh38
rs559409372 Chr.15: 64381272-64381272 on Build GRCh38
rs564807992 Chr.15: 64381281-64381281 on Build GRCh38
rs990429207 Chr.15: 64381508-64381508 on Build GRCh38
rs755251540 Chr.15: 64381385-64381389 on Build GRCh38
rs769742010 Chr.15: 64381389-64381389 on Build GRCh38
rs947942319 Chr.15: 64381656-64381656 on Build GRCh38
rs1012493132 Chr.15: 64381270-64381270 on Build GRCh38
rs11554310 Chr.15: 64381416-64381416 on Build GRCh38
rs770123662 Chr.15: 64381279-64381279 on Build GRCh38
rs201415383 Chr.15: 64381314-64381314 on Build GRCh38
rs759261855 Chr.15: 64381414-64381414 on Build GRCh38
rs141142230 Chr.15: 64381288-64381289 on Build GRCh38
rs759492613 Chr.15: 64381300-64381300 on Build GRCh38
rs11554319 Chr.15: 64381404-64381404 on Build GRCh38
rs778101576 Chr.15: 64381395-64381395 on Build GRCh38
rs539483320 Chr.15: 64381537-64381537 on Build GRCh38
rs760097397 Chr.15: 64381327-64381327 on Build GRCh38
rs781735423 Chr.15: 64381366-64381366 on Build GRCh38
rs529794721 Chr.15: 64381503-64381503 on Build GRCh38

View Full Product Details