Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 410350-410856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DECR2
Entrez Gene ID
26063
Gene Name
2,4-dienoyl-CoA reductase 2, peroxisomal
Gene Aliases
PDCR, SDR17C1
UniGene
Hs.628831
Gene Chromosome Location
Chr.16: 401858-412487 on Build GRCh38
Transcripts
NM_020664.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2920620 c.480C>T p.I160I Chr.16: 410708-410708 on Build GRCh38
7218695 c.453G>T p.K151N Chr.16: 410358-410358 on Build GRCh38
3361766 c.634T>G p.*212G Chr.16: 410352-410352 on Build GRCh38
5016997 c.536G>C p.G179A Chr.16: 410764-410764 on Build GRCh38
6642128 c.461G>A p.R154Q Chr.16: 410366-410366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561603584 Chr.16: 410599-410599 on Build GRCh38
rs9924749 Chr.16: 410587-410587 on Build GRCh38
rs9924748 Chr.16: 410583-410583 on Build GRCh38
rs998694469 Chr.16: 410496-410496 on Build GRCh38
rs374096745 Chr.16: 410687-410687 on Build GRCh38
rs776712367 Chr.16: 410801-410801 on Build GRCh38
rs200342533 Chr.16: 410543-410543 on Build GRCh38
rs3743891 Chr.16: 410415-410415 on Build GRCh38
rs771806477 Chr.16: 410419-410419 on Build GRCh38
rs188648786 Chr.16: 410425-410425 on Build GRCh38
rs749261930 Chr.16: 410440-410440 on Build GRCh38
rs749395910 Chr.16: 410656-410656 on Build GRCh38
rs373172839 Chr.16: 410650-410650 on Build GRCh38
rs377244160 Chr.16: 410386-410386 on Build GRCh38
rs375711492 Chr.16: 410723-410723 on Build GRCh38
rs774605806 Chr.16: 410392-410420 on Build GRCh38
rs948926774 Chr.16: 410777-410777 on Build GRCh38
rs547888369 Chr.16: 410697-410697 on Build GRCh38
rs3743894 Chr.16: 410543-410543 on Build GRCh38
rs3743892 Chr.16: 410448-410448 on Build GRCh38
rs201553914 Chr.16: 410383-410383 on Build GRCh38
rs373529957 Chr.16: 410702-410702 on Build GRCh38
rs770721092 Chr.16: 410672-410672 on Build GRCh38
rs1023283065 Chr.16: 410612-410612 on Build GRCh38
rs774308860 Chr.16: 410799-410799 on Build GRCh38
rs767867489 Chr.16: 410737-410737 on Build GRCh38
rs754135610 Chr.16: 410768-410768 on Build GRCh38
rs866218680 Chr.16: 410555-410555 on Build GRCh38
rs534724557 Chr.16: 410516-410516 on Build GRCh38
rs369241969 Chr.16: 410602-410602 on Build GRCh38
rs199661265 Chr.16: 410382-410382 on Build GRCh38
rs764491607 Chr.16: 410790-410791 on Build GRCh38
rs776626806 Chr.16: 410646-410646 on Build GRCh38
rs922870459 Chr.16: 410435-410435 on Build GRCh38
rs746724359 Chr.16: 410661-410661 on Build GRCh38
rs769206831 Chr.16: 410416-410416 on Build GRCh38
rs922556875 Chr.16: 410542-410542 on Build GRCh38
rs112026055 Chr.16: 410659-410659 on Build GRCh38
rs759622204 Chr.16: 410412-410413 on Build GRCh38
rs772473294 Chr.16: 410508-410508 on Build GRCh38
rs531767849 Chr.16: 410388-410388 on Build GRCh38
rs748095831 Chr.16: 410382-410384 on Build GRCh38
rs61645593 Chr.16: 410830-410830 on Build GRCh38
rs1004052524 Chr.16: 410503-410503 on Build GRCh38
rs576664155 Chr.16: 410819-410819 on Build GRCh38
rs1003934675 Chr.16: 410742-410742 on Build GRCh38
rs754528043 Chr.16: 410836-410836 on Build GRCh38
rs1045843099 Chr.16: 410485-410485 on Build GRCh38
rs766475255 Chr.16: 410763-410763 on Build GRCh38
rs565878000 Chr.16: 410417-410417 on Build GRCh38
rs369873189 Chr.16: 410676-410676 on Build GRCh38
rs765424319 Chr.16: 410775-410775 on Build GRCh38
rs780529988 Chr.16: 410408-410408 on Build GRCh38
rs1011385518 Chr.16: 410509-410509 on Build GRCh38
rs28398951 Chr.16: 410630-410630 on Build GRCh38
rs908677978 Chr.16: 410446-410446 on Build GRCh38
rs35431910 Chr.16: 410802-410803 on Build GRCh38
rs752172006 Chr.16: 410815-410815 on Build GRCh38
rs1039250947 Chr.16: 410427-410427 on Build GRCh38
rs767288280 Chr.16: 410378-410378 on Build GRCh38
rs542830909 Chr.16: 410591-410591 on Build GRCh38
rs369798941 Chr.16: 410696-410696 on Build GRCh38
rs983005097 Chr.16: 410544-410544 on Build GRCh38
rs748746167 Chr.16: 410728-410728 on Build GRCh38
rs3743893 Chr.16: 410504-410504 on Build GRCh38
rs527313474 Chr.16: 410376-410376 on Build GRCh38
rs536949084 Chr.16: 410755-410755 on Build GRCh38
rs766642876 Chr.16: 410709-410709 on Build GRCh38
rs550183591 Chr.16: 410397-410397 on Build GRCh38
rs370736528 Chr.16: 410737-410737 on Build GRCh38
rs752549486 Chr.16: 410395-410395 on Build GRCh38
rs191919628 Chr.16: 410574-410574 on Build GRCh38
rs748761351 Chr.16: 410833-410833 on Build GRCh38
rs144950917 Chr.16: 410746-410746 on Build GRCh38
rs750770772 Chr.16: 410823-410823 on Build GRCh38
rs975434549 Chr.16: 410533-410533 on Build GRCh38
rs774079592 Chr.16: 410754-410754 on Build GRCh38
rs894240795 Chr.16: 410476-410476 on Build GRCh38
rs1031562309 Chr.16: 410502-410502 on Build GRCh38
rs759736356 Chr.16: 410802-410802 on Build GRCh38
rs979161525 Chr.16: 410626-410626 on Build GRCh38
rs992771632 Chr.16: 410531-410531 on Build GRCh38
rs774205521 Chr.16: 410545-410546 on Build GRCh38
rs750704568 Chr.16: 410651-410651 on Build GRCh38
rs9932806 Chr.16: 410595-410595 on Build GRCh38
rs200588772 Chr.16: 410739-410739 on Build GRCh38
rs955337412 Chr.16: 410735-410735 on Build GRCh38
rs141003182 Chr.16: 410727-410727 on Build GRCh38
rs779503831 Chr.16: 410657-410657 on Build GRCh38
rs1022363803 Chr.16: 410517-410517 on Build GRCh38
rs908813499 Chr.16: 410578-410578 on Build GRCh38
rs3743895 Chr.16: 410673-410673 on Build GRCh38
rs572622836 Chr.16: 410547-410547 on Build GRCh38
rs998495170 Chr.16: 410680-410680 on Build GRCh38
rs970120641 Chr.16: 410621-410621 on Build GRCh38
rs11315123 Chr.16: 410579-410579 on Build GRCh38
rs761014616 Chr.16: 410789-410790 on Build GRCh38
rs115317717 Chr.16: 410796-410796 on Build GRCh38
rs200583305 Chr.16: 410403-410403 on Build GRCh38
rs765243014 Chr.16: 410649-410649 on Build GRCh38
rs367817507 Chr.16: 410769-410769 on Build GRCh38
rs772984866 Chr.16: 410660-410660 on Build GRCh38
rs769178682 Chr.16: 410497-410498 on Build GRCh38
rs569596237 Chr.16: 410456-410456 on Build GRCh38
rs372285003 Chr.16: 410787-410787 on Build GRCh38
rs778456055 Chr.16: 410792-410792 on Build GRCh38
rs375233826 Chr.16: 410798-410798 on Build GRCh38
rs751943184 Chr.16: 410793-410793 on Build GRCh38
rs576800459 Chr.16: 410603-410603 on Build GRCh38
rs771958925 Chr.16: 410800-410800 on Build GRCh38
rs377511084 Chr.16: 410730-410730 on Build GRCh38
rs1052945601 Chr.16: 410689-410689 on Build GRCh38
rs9936742 Chr.16: 410487-410487 on Build GRCh38
rs201367889 Chr.16: 410407-410407 on Build GRCh38
rs1047348456 Chr.16: 410445-410445 on Build GRCh38
rs779472210 Chr.16: 410420-410420 on Build GRCh38
rs755122943 Chr.16: 410726-410726 on Build GRCh38
rs9936889 Chr.16: 410622-410622 on Build GRCh38
rs760291085 Chr.16: 410380-410380 on Build GRCh38
rs759334874 Chr.16: 410648-410648 on Build GRCh38
rs181461221 Chr.16: 410797-410797 on Build GRCh38
rs200015511 Chr.16: 410794-410794 on Build GRCh38
rs150042425 Chr.16: 410618-410618 on Build GRCh38
rs893082477 Chr.16: 410701-410701 on Build GRCh38
rs775843178 Chr.16: 410767-410769 on Build GRCh38
rs555199808 Chr.16: 410818-410818 on Build GRCh38
rs779707992 Chr.16: 410832-410832 on Build GRCh38
rs60098912 Chr.16: 410795-410795 on Build GRCh38
rs149208666 Chr.16: 410708-410708 on Build GRCh38
rs374884091 Chr.16: 410655-410655 on Build GRCh38
rs964507695 Chr.16: 410521-410521 on Build GRCh38
rs147456438 Chr.16: 410747-410747 on Build GRCh38
rs765056268 Chr.16: 410681-410681 on Build GRCh38
rs769804320 Chr.16: 410383-410390 on Build GRCh38
rs768599023 Chr.16: 410753-410753 on Build GRCh38
rs778216557 Chr.16: 410731-410731 on Build GRCh38
rs552010933 Chr.16: 410778-410778 on Build GRCh38
rs974236470 Chr.16: 410764-410764 on Build GRCh38
rs754356484 Chr.16: 410793-410794 on Build GRCh38
rs752785156 Chr.16: 410803-410803 on Build GRCh38
rs9939384 Chr.16: 410567-410567 on Build GRCh38
rs371311313 Chr.16: 410379-410379 on Build GRCh38
rs747400979 Chr.16: 410736-410736 on Build GRCh38
rs764677277 Chr.16: 410813-410813 on Build GRCh38
rs536686090 Chr.16: 410477-410477 on Build GRCh38
rs369453909 Chr.16: 410402-410402 on Build GRCh38
rs532650826 Chr.16: 410412-410412 on Build GRCh38
rs757808522 Chr.16: 410794-410794 on Build GRCh38
rs906869888 Chr.16: 410495-410495 on Build GRCh38
rs745349366 Chr.16: 410741-410741 on Build GRCh38
rs548488938 Chr.16: 410452-410452 on Build GRCh38
rs536028257 Chr.16: 410426-410426 on Build GRCh38

View Full Product Details