Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1826904-1827438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAHD1
Entrez Gene ID
81889
Gene Name
fumarylacetoacetate hydrolase domain containing 1
Gene Aliases
C16orf36, YISKL
UniGene
Hs.513265
Gene Chromosome Location
Chr.16: 1827224-1840207 on Build GRCh38
Transcripts
NM_001018104.2 NM_001142398.1 NM_031208.3 NM_001040427.1 NM_001286249.1 NM_005326.4

Gene Symbol
HAGH
Entrez Gene ID
3029
Gene Name
hydroxyacylglutathione hydrolase
Gene Aliases
GLO2, GLX2, GLXII, HAGH1
UniGene
Hs.157394
Gene Chromosome Location
Chr.16: 1809099-1827194 on Build GRCh38
Transcripts
NM_001018104.2 NM_001142398.1 NM_031208.3 NM_001040427.1 NM_001286249.1 NM_005326.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7422627 c.25C>G p.P9A Chr.16: 1827254-1827254 on Build GRCh38
4169013 c.157G>T p.A53S Chr.16: 1827386-1827386 on Build GRCh38
3506581 c.4G>A p.G2R Chr.16: 1827233-1827233 on Build GRCh38
5170671 c.36C>T p.R12R Chr.16: 1827265-1827265 on Build GRCh38
4847907 c.170C>T p.S57L Chr.16: 1827399-1827399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374169944 Chr.16: 1827269-1827269 on Build GRCh38
rs912896729 Chr.16: 1827310-1827310 on Build GRCh38
rs746561454 Chr.16: 1827085-1827085 on Build GRCh38
rs765572027 Chr.16: 1827218-1827218 on Build GRCh38
rs559334672 Chr.16: 1827164-1827164 on Build GRCh38
rs769628893 Chr.16: 1827233-1827233 on Build GRCh38
rs74518158 Chr.16: 1827391-1827391 on Build GRCh38
rs745817340 Chr.16: 1827231-1827231 on Build GRCh38
rs201663677 Chr.16: 1827222-1827222 on Build GRCh38
rs985359469 Chr.16: 1827020-1827020 on Build GRCh38
rs778746548 Chr.16: 1827274-1827274 on Build GRCh38
rs965030710 Chr.16: 1826958-1826958 on Build GRCh38
rs1049276504 Chr.16: 1827139-1827139 on Build GRCh38
rs760027708 Chr.16: 1827217-1827217 on Build GRCh38
rs772713495 Chr.16: 1827252-1827252 on Build GRCh38
rs757623702 Chr.16: 1827261-1827261 on Build GRCh38
rs150059885 Chr.16: 1827271-1827271 on Build GRCh38
rs754752783 Chr.16: 1827272-1827272 on Build GRCh38
rs771725204 Chr.16: 1827289-1827289 on Build GRCh38
rs954131473 Chr.16: 1827017-1827017 on Build GRCh38
rs532501579 Chr.16: 1827095-1827095 on Build GRCh38
rs906033565 Chr.16: 1827123-1827123 on Build GRCh38
rs767879757 Chr.16: 1827326-1827326 on Build GRCh38
rs769621726 Chr.16: 1827188-1827188 on Build GRCh38
rs201966044 Chr.16: 1827392-1827392 on Build GRCh38
rs770138965 Chr.16: 1827239-1827239 on Build GRCh38
rs748721127 Chr.16: 1827416-1827416 on Build GRCh38
rs746726928 Chr.16: 1827356-1827356 on Build GRCh38
rs141621076 Chr.16: 1827384-1827384 on Build GRCh38
rs199742650 Chr.16: 1827417-1827417 on Build GRCh38
rs906148726 Chr.16: 1826954-1826954 on Build GRCh38
rs957502688 Chr.16: 1827250-1827250 on Build GRCh38
rs768476314 Chr.16: 1827240-1827240 on Build GRCh38
rs746533693 Chr.16: 1827295-1827295 on Build GRCh38
rs939088659 Chr.16: 1827022-1827022 on Build GRCh38
rs774313686 Chr.16: 1827356-1827356 on Build GRCh38
rs550938698 Chr.16: 1827375-1827375 on Build GRCh38
rs983946746 Chr.16: 1826985-1826985 on Build GRCh38
rs753713338 Chr.16: 1827344-1827344 on Build GRCh38
rs143728918 Chr.16: 1827415-1827415 on Build GRCh38
rs369066164 Chr.16: 1827207-1827207 on Build GRCh38
rs952046907 Chr.16: 1826974-1826974 on Build GRCh38
rs369724815 Chr.16: 1827161-1827161 on Build GRCh38
rs747732396 Chr.16: 1827277-1827277 on Build GRCh38
rs762605469 Chr.16: 1827192-1827192 on Build GRCh38
rs754329258 Chr.16: 1827211-1827211 on Build GRCh38
rs779845810 Chr.16: 1827238-1827238 on Build GRCh38
rs574585733 Chr.16: 1827061-1827061 on Build GRCh38
rs756942777 Chr.16: 1827401-1827401 on Build GRCh38
rs949785898 Chr.16: 1827104-1827104 on Build GRCh38
rs757654537 Chr.16: 1827169-1827169 on Build GRCh38
rs13330675 Chr.16: 1826943-1826943 on Build GRCh38
rs778621990 Chr.16: 1827350-1827350 on Build GRCh38
rs757392213 Chr.16: 1827228-1827228 on Build GRCh38
rs1027883144 Chr.16: 1827215-1827215 on Build GRCh38
rs776275467 Chr.16: 1827366-1827366 on Build GRCh38
rs994770476 Chr.16: 1827205-1827205 on Build GRCh38
rs780514775 Chr.16: 1827407-1827407 on Build GRCh38
rs751873738 Chr.16: 1827225-1827225 on Build GRCh38
rs749749641 Chr.16: 1827367-1827367 on Build GRCh38
rs752672591 Chr.16: 1827394-1827394 on Build GRCh38
rs539990790 Chr.16: 1826948-1826948 on Build GRCh38
rs781246581 Chr.16: 1827181-1827181 on Build GRCh38
rs745854469 Chr.16: 1827182-1827182 on Build GRCh38
rs973627936 Chr.16: 1826971-1826971 on Build GRCh38
rs1053291674 Chr.16: 1827092-1827092 on Build GRCh38
rs543294784 Chr.16: 1827062-1827062 on Build GRCh38
rs28364709 Chr.16: 1827174-1827174 on Build GRCh38
rs775149181 Chr.16: 1827189-1827189 on Build GRCh38
rs572036965 Chr.16: 1826950-1826950 on Build GRCh38
rs1004857996 Chr.16: 1827152-1827152 on Build GRCh38
rs949837941 Chr.16: 1827340-1827340 on Build GRCh38
rs145433996 Chr.16: 1827296-1827296 on Build GRCh38
rs76260324 Chr.16: 1827005-1827006 on Build GRCh38
rs961215462 Chr.16: 1827000-1827000 on Build GRCh38
rs1006690331 Chr.16: 1827388-1827388 on Build GRCh38
rs560980687 Chr.16: 1826991-1826991 on Build GRCh38
rs1029896119 Chr.16: 1827149-1827149 on Build GRCh38
rs751170172 Chr.16: 1827399-1827399 on Build GRCh38
rs976612401 Chr.16: 1827001-1827001 on Build GRCh38
rs929492834 Chr.16: 1826982-1826982 on Build GRCh38
rs890579601 Chr.16: 1827167-1827167 on Build GRCh38
rs769465409 Chr.16: 1827412-1827412 on Build GRCh38
rs999382158 Chr.16: 1827140-1827140 on Build GRCh38
rs370553546 Chr.16: 1827247-1827247 on Build GRCh38
rs7188401 Chr.16: 1826948-1826948 on Build GRCh38
rs28364708 Chr.16: 1826991-1826992 on Build GRCh38
rs538771887 Chr.16: 1827378-1827378 on Build GRCh38
rs765687388 Chr.16: 1827253-1827253 on Build GRCh38
rs758168909 Chr.16: 1827395-1827395 on Build GRCh38
rs937503012 Chr.16: 1827353-1827353 on Build GRCh38
rs779821974 Chr.16: 1827270-1827270 on Build GRCh38
rs766075445 Chr.16: 1827303-1827303 on Build GRCh38
rs1014923611 Chr.16: 1827156-1827156 on Build GRCh38
rs754914005 Chr.16: 1827349-1827349 on Build GRCh38
rs762235061 Chr.16: 1827373-1827373 on Build GRCh38
rs747960009 Chr.16: 1827352-1827352 on Build GRCh38
rs887795423 Chr.16: 1827145-1827145 on Build GRCh38
rs761280959 Chr.16: 1827206-1827206 on Build GRCh38
rs922425066 Chr.16: 1827010-1827010 on Build GRCh38
rs753016924 Chr.16: 1827219-1827219 on Build GRCh38
rs773969752 Chr.16: 1827243-1827243 on Build GRCh38
rs751822858 Chr.16: 1827258-1827258 on Build GRCh38
rs764552587 Chr.16: 1827313-1827313 on Build GRCh38
rs541359078 Chr.16: 1826970-1826970 on Build GRCh38
rs774863827 Chr.16: 1827314-1827314 on Build GRCh38
rs1045424436 Chr.16: 1827342-1827342 on Build GRCh38
rs1045917957 Chr.16: 1827112-1827112 on Build GRCh38
rs766536784 Chr.16: 1827343-1827343 on Build GRCh38
rs773808219 Chr.16: 1827204-1827204 on Build GRCh38
rs745399106 Chr.16: 1827410-1827410 on Build GRCh38
rs548865046 Chr.16: 1827199-1827199 on Build GRCh38
rs763615875 Chr.16: 1827308-1827308 on Build GRCh38
rs1043676815 Chr.16: 1827107-1827107 on Build GRCh38
rs142644041 Chr.16: 1827354-1827354 on Build GRCh38
rs1014994143 Chr.16: 1826987-1826987 on Build GRCh38
rs200115670 Chr.16: 1827329-1827329 on Build GRCh38
rs1002202691 Chr.16: 1827134-1827134 on Build GRCh38
rs773714274 Chr.16: 1827114-1827125 on Build GRCh38
rs753068298 Chr.16: 1827255-1827255 on Build GRCh38
rs1017650020 Chr.16: 1827176-1827176 on Build GRCh38
rs545879432 Chr.16: 1827150-1827150 on Build GRCh38
rs563496689 Chr.16: 1827064-1827064 on Build GRCh38
rs374700352 Chr.16: 1827397-1827397 on Build GRCh38
rs753965151 Chr.16: 1827390-1827390 on Build GRCh38
rs373607849 Chr.16: 1827246-1827246 on Build GRCh38
rs144580262 Chr.16: 1827372-1827372 on Build GRCh38
rs913011357 Chr.16: 1827102-1827102 on Build GRCh38
rs762073854 Chr.16: 1827316-1827316 on Build GRCh38
rs73488045 Chr.16: 1827376-1827376 on Build GRCh38
rs746163377 Chr.16: 1827201-1827201 on Build GRCh38
rs781484943 Chr.16: 1827230-1827230 on Build GRCh38
rs925609762 Chr.16: 1827060-1827060 on Build GRCh38
rs746614021 Chr.16: 1826960-1826960 on Build GRCh38
rs772730936 Chr.16: 1827294-1827294 on Build GRCh38
rs781358600 Chr.16: 1827265-1827265 on Build GRCh38
rs367923567 Chr.16: 1827266-1827266 on Build GRCh38
rs888066927 Chr.16: 1827355-1827355 on Build GRCh38
rs1054836573 Chr.16: 1827063-1827063 on Build GRCh38
rs770337528 Chr.16: 1827364-1827364 on Build GRCh38
rs756122765 Chr.16: 1827332-1827332 on Build GRCh38

View Full Product Details