Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3067239-3067747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL32
Entrez Gene ID
9235
Gene Name
interleukin 32
Gene Aliases
IL-32alpha, IL-32beta, IL-32delta, IL-32gamma, NK4, TAIF, TAIFa, TAIFb, TAIFc, TAIFd
UniGene
Hs.943
Gene Chromosome Location
Chr.16: 3065312-3069819 on Build GRCh38
Transcripts
NM_001012631.1 NM_001012632.1 NM_001012633.1 NM_001012634.1 NM_001012635.1 NM_001012636.1 NM_001012718.1 NM_001308078.1 NM_004221.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3509094 c.245G>T p.R82L Chr.16: 3067606-3067606 on Build GRCh38
3690959 c.16G>A p.V6I Chr.16: 3067377-3067377 on Build GRCh38
1708653 c.54G>A p.M18I Chr.16: 3067415-3067415 on Build GRCh38
3509091 c.113C>T p.T38I Chr.16: 3067474-3067474 on Build GRCh38
3509092 c.158C>T p.P53L Chr.16: 3067519-3067519 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772245098 Chr.16: 3067512-3067512 on Build GRCh38
rs543754704 Chr.16: 3067362-3067362 on Build GRCh38
rs774088325 Chr.16: 3067708-3067708 on Build GRCh38
rs752612594 Chr.16: 3067682-3067682 on Build GRCh38
rs764211929 Chr.16: 3067686-3067686 on Build GRCh38
rs753486240 Chr.16: 3067542-3067542 on Build GRCh38
rs985176934 Chr.16: 3067268-3067268 on Build GRCh38
rs777862930 Chr.16: 3067657-3067657 on Build GRCh38
rs201900317 Chr.16: 3067435-3067435 on Build GRCh38
rs909242927 Chr.16: 3067269-3067269 on Build GRCh38
rs375355403 Chr.16: 3067520-3067520 on Build GRCh38
rs575998875 Chr.16: 3067635-3067635 on Build GRCh38
rs763748487 Chr.16: 3067535-3067535 on Build GRCh38
rs573179124 Chr.16: 3067343-3067343 on Build GRCh38
rs771642503 Chr.16: 3067342-3067342 on Build GRCh38
rs769232002 Chr.16: 3067703-3067703 on Build GRCh38
rs747931658 Chr.16: 3067289-3067289 on Build GRCh38
rs750210356 Chr.16: 3067599-3067599 on Build GRCh38
rs566802763 Chr.16: 3067519-3067519 on Build GRCh38
rs866469249 Chr.16: 3067271-3067271 on Build GRCh38
rs149999866 Chr.16: 3067272-3067272 on Build GRCh38
rs865818341 Chr.16: 3067304-3067310 on Build GRCh38
rs894845199 Chr.16: 3067276-3067277 on Build GRCh38
rs969298612 Chr.16: 3067637-3067637 on Build GRCh38
rs774782082 Chr.16: 3067575-3067575 on Build GRCh38
rs761171126 Chr.16: 3067419-3067419 on Build GRCh38
rs763334955 Chr.16: 3067570-3067570 on Build GRCh38
rs1050261261 Chr.16: 3067305-3067305 on Build GRCh38
rs539684284 Chr.16: 3067476-3067476 on Build GRCh38
rs757668081 Chr.16: 3067660-3067660 on Build GRCh38
rs754580611 Chr.16: 3067482-3067482 on Build GRCh38
rs767794140 Chr.16: 3067413-3067413 on Build GRCh38
rs144171852 Chr.16: 3067272-3067289 on Build GRCh38
rs748418655 Chr.16: 3067609-3067609 on Build GRCh38
rs777364422 Chr.16: 3067488-3067488 on Build GRCh38
rs3223106 Chr.16: 3067266-3067267 on Build GRCh38
rs547829248 Chr.16: 3067406-3067406 on Build GRCh38
rs768786551 Chr.16: 3067560-3067560 on Build GRCh38
rs528198933 Chr.16: 3067401-3067401 on Build GRCh38
rs761965943 Chr.16: 3067668-3067668 on Build GRCh38
rs75809617 Chr.16: 3067335-3067335 on Build GRCh38
rs760631512 Chr.16: 3067721-3067721 on Build GRCh38
rs188420220 Chr.16: 3067718-3067718 on Build GRCh38
rs954767005 Chr.16: 3067559-3067559 on Build GRCh38
rs758302498 Chr.16: 3067546-3067546 on Build GRCh38
rs267604510 Chr.16: 3067624-3067624 on Build GRCh38
rs367909393 Chr.16: 3067436-3067436 on Build GRCh38
rs371706904 Chr.16: 3067449-3067449 on Build GRCh38
rs773196760 Chr.16: 3067408-3067408 on Build GRCh38
rs778376368 Chr.16: 3067339-3067339 on Build GRCh38
rs370139293 Chr.16: 3067551-3067551 on Build GRCh38
rs1006386862 Chr.16: 3067629-3067629 on Build GRCh38
rs140230160 Chr.16: 3067272-3067277 on Build GRCh38
rs750515606 Chr.16: 3067724-3067724 on Build GRCh38
rs755730296 Chr.16: 3067487-3067487 on Build GRCh38
rs539533027 Chr.16: 3067317-3067317 on Build GRCh38
rs140063808 Chr.16: 3067605-3067605 on Build GRCh38
rs762540486 Chr.16: 3067448-3067448 on Build GRCh38
rs1011998596 Chr.16: 3067277-3067278 on Build GRCh38
rs769602896 Chr.16: 3067422-3067423 on Build GRCh38
rs976845357 Chr.16: 3067265-3067265 on Build GRCh38
rs763899227 Chr.16: 3067722-3067722 on Build GRCh38
rs1017679453 Chr.16: 3067667-3067667 on Build GRCh38
rs761341059 Chr.16: 3067633-3067633 on Build GRCh38
rs933185044 Chr.16: 3067269-3067270 on Build GRCh38
rs114354531 Chr.16: 3067377-3067377 on Build GRCh38
rs1047618314 Chr.16: 3067270-3067273 on Build GRCh38
rs145687578 Chr.16: 3067606-3067606 on Build GRCh38
rs756612456 Chr.16: 3067337-3067337 on Build GRCh38
rs141094270 Chr.16: 3067643-3067643 on Build GRCh38
rs3063339 Chr.16: 3067310-3067311 on Build GRCh38
rs762726968 Chr.16: 3067638-3067638 on Build GRCh38
rs748704496 Chr.16: 3067705-3067705 on Build GRCh38
rs112498944 Chr.16: 3067273-3067273 on Build GRCh38
rs375602999 Chr.16: 3067357-3067357 on Build GRCh38
rs776523987 Chr.16: 3067287-3067287 on Build GRCh38
rs201732786 Chr.16: 3067308-3067308 on Build GRCh38
rs754385064 Chr.16: 3067428-3067428 on Build GRCh38
rs146964710 Chr.16: 3067654-3067654 on Build GRCh38
rs936632496 Chr.16: 3067311-3067311 on Build GRCh38
rs778976584 Chr.16: 3067505-3067505 on Build GRCh38
rs537370763 Chr.16: 3067478-3067478 on Build GRCh38
rs754932618 Chr.16: 3067651-3067651 on Build GRCh38
rs751803269 Chr.16: 3067611-3067611 on Build GRCh38
rs377148978 Chr.16: 3067424-3067424 on Build GRCh38
rs764404714 Chr.16: 3067608-3067608 on Build GRCh38
rs772745333 Chr.16: 3067704-3067704 on Build GRCh38
rs745965931 Chr.16: 3067452-3067452 on Build GRCh38
rs758158107 Chr.16: 3067602-3067602 on Build GRCh38
rs139622637 Chr.16: 3067356-3067356 on Build GRCh38
rs951472884 Chr.16: 3067458-3067458 on Build GRCh38
rs774105849 Chr.16: 3067451-3067451 on Build GRCh38
rs993441075 Chr.16: 3067544-3067544 on Build GRCh38
rs202247796 Chr.16: 3067626-3067626 on Build GRCh38
rs115923319 Chr.16: 3067563-3067563 on Build GRCh38
rs753539663 Chr.16: 3067578-3067578 on Build GRCh38
rs190241571 Chr.16: 3067373-3067373 on Build GRCh38
rs763343691 Chr.16: 3067528-3067528 on Build GRCh38
rs747430360 Chr.16: 3067605-3067606 on Build GRCh38
rs55756224 Chr.16: 3067425-3067425 on Build GRCh38
rs762223099 Chr.16: 3067469-3067469 on Build GRCh38
rs760959135 Chr.16: 3067465-3067465 on Build GRCh38
rs144473418 Chr.16: 3067569-3067569 on Build GRCh38
rs972126450 Chr.16: 3067291-3067291 on Build GRCh38
rs766919795 Chr.16: 3067485-3067485 on Build GRCh38
rs116296862 Chr.16: 3067664-3067664 on Build GRCh38
rs768197879 Chr.16: 3067622-3067622 on Build GRCh38
rs746762861 Chr.16: 3067284-3067289 on Build GRCh38
rs761826780 Chr.16: 3067726-3067726 on Build GRCh38
rs1018273489 Chr.16: 3067309-3067309 on Build GRCh38
rs555441503 Chr.16: 3067545-3067545 on Build GRCh38
rs748365535 Chr.16: 3067564-3067564 on Build GRCh38
rs746247110 Chr.16: 3067695-3067695 on Build GRCh38
rs768389623 Chr.16: 3067348-3067348 on Build GRCh38
rs757503745 Chr.16: 3067689-3067689 on Build GRCh38
rs770728868 Chr.16: 3067706-3067706 on Build GRCh38
rs570727825 Chr.16: 3067295-3067295 on Build GRCh38
rs1555001 Chr.16: 3067353-3067353 on Build GRCh38
rs773073934 Chr.16: 3067355-3067355 on Build GRCh38
rs776458480 Chr.16: 3067350-3067350 on Build GRCh38
rs780656000 Chr.16: 3067397-3067397 on Build GRCh38
rs761341688 Chr.16: 3067587-3067587 on Build GRCh38
rs941662114 Chr.16: 3067270-3067287 on Build GRCh38
rs752796018 Chr.16: 3067416-3067416 on Build GRCh38
rs933147432 Chr.16: 3067303-3067303 on Build GRCh38
rs774345723 Chr.16: 3067682-3067682 on Build GRCh38
rs767588584 Chr.16: 3067678-3067678 on Build GRCh38
rs774458382 Chr.16: 3067410-3067410 on Build GRCh38
rs752233130 Chr.16: 3067531-3067531 on Build GRCh38
rs752442725 Chr.16: 3067486-3067486 on Build GRCh38
rs372974931 Chr.16: 3067628-3067628 on Build GRCh38
rs753199544 Chr.16: 3067309-3067310 on Build GRCh38
rs1018972379 Chr.16: 3067431-3067431 on Build GRCh38
rs959199113 Chr.16: 3067354-3067354 on Build GRCh38
rs746682981 Chr.16: 3067390-3067390 on Build GRCh38
rs755957701 Chr.16: 3067440-3067440 on Build GRCh38
rs879136389 Chr.16: 3067272-3067273 on Build GRCh38
rs369943340 Chr.16: 3067571-3067571 on Build GRCh38
rs778552874 Chr.16: 3067543-3067543 on Build GRCh38
rs781606389 Chr.16: 3067331-3067331 on Build GRCh38
rs749103415 Chr.16: 3067498-3067498 on Build GRCh38
rs778001461 Chr.16: 3067610-3067610 on Build GRCh38
rs772859719 Chr.16: 3067636-3067636 on Build GRCh38
rs372604790 Chr.16: 3067392-3067392 on Build GRCh38
rs759437113 Chr.16: 3067717-3067718 on Build GRCh38
rs751594072 Chr.16: 3067650-3067650 on Build GRCh38
rs147586298 Chr.16: 3067684-3067684 on Build GRCh38
rs756980976 Chr.16: 3067503-3067503 on Build GRCh38
rs535438991 Chr.16: 3067345-3067345 on Build GRCh38
rs753135810 Chr.16: 3067526-3067526 on Build GRCh38
rs1054890147 Chr.16: 3067620-3067620 on Build GRCh38
rs923074327 Chr.16: 3067299-3067299 on Build GRCh38
rs962309213 Chr.16: 3067536-3067536 on Build GRCh38
rs370302447 Chr.16: 3067715-3067715 on Build GRCh38
rs202133237 Chr.16: 3067445-3067445 on Build GRCh38
rs762269785 Chr.16: 3067522-3067522 on Build GRCh38
rs369495267 Chr.16: 3067466-3067466 on Build GRCh38
rs771259952 Chr.16: 3067616-3067616 on Build GRCh38
rs764542270 Chr.16: 3067420-3067420 on Build GRCh38
rs773824307 Chr.16: 3067524-3067524 on Build GRCh38
rs913522456 Chr.16: 3067441-3067441 on Build GRCh38
rs779059587 Chr.16: 3067692-3067692 on Build GRCh38
rs758697810 Chr.16: 3067697-3067697 on Build GRCh38
rs147775375 Chr.16: 3067576-3067576 on Build GRCh38
rs60859102 Chr.16: 3067307-3067310 on Build GRCh38
rs550736863 Chr.16: 3067293-3067293 on Build GRCh38
rs537354219 Chr.16: 3067277-3067277 on Build GRCh38
rs747102738 Chr.16: 3067555-3067555 on Build GRCh38
rs1003039477 Chr.16: 3067352-3067352 on Build GRCh38
rs138027970 Chr.16: 3067508-3067508 on Build GRCh38
rs201114894 Chr.16: 3067515-3067515 on Build GRCh38
rs373456514 Chr.16: 3067372-3067372 on Build GRCh38
rs796776821 Chr.16: 3067289-3067290 on Build GRCh38
rs762640573 Chr.16: 3067677-3067677 on Build GRCh38
rs753135817 Chr.16: 3067367-3067367 on Build GRCh38
rs998107980 Chr.16: 3067573-3067573 on Build GRCh38
rs377020176 Chr.16: 3067364-3067364 on Build GRCh38
rs910339811 Chr.16: 3067566-3067566 on Build GRCh38
rs764907070 Chr.16: 3067596-3067596 on Build GRCh38
rs774315939 Chr.16: 3067679-3067679 on Build GRCh38
rs1031232824 Chr.16: 3067291-3067292 on Build GRCh38
rs760014118 Chr.16: 3067621-3067621 on Build GRCh38
rs374739811 Chr.16: 3067267-3067268 on Build GRCh38
rs772802450 Chr.16: 3067430-3067431 on Build GRCh38
rs886375489 Chr.16: 3067270-3067275 on Build GRCh38
rs937719935 Chr.16: 3067614-3067614 on Build GRCh38
rs561371637 Chr.16: 3067411-3067411 on Build GRCh38
rs767521322 Chr.16: 3067653-3067653 on Build GRCh38
rs936563185 Chr.16: 3067434-3067434 on Build GRCh38
rs376323814 Chr.16: 3067461-3067461 on Build GRCh38
rs776398270 Chr.16: 3067294-3067298 on Build GRCh38
rs868507983 Chr.16: 3067471-3067471 on Build GRCh38
rs777598202 Chr.16: 3067283-3067283 on Build GRCh38
rs756377965 Chr.16: 3067368-3067368 on Build GRCh38
rs75543532 Chr.16: 3067702-3067702 on Build GRCh38
rs749627881 Chr.16: 3067613-3067613 on Build GRCh38
rs117585555 Chr.16: 3067674-3067674 on Build GRCh38
rs774897010 Chr.16: 3067618-3067618 on Build GRCh38
rs767093013 Chr.16: 3067529-3067529 on Build GRCh38
rs368756741 Chr.16: 3067680-3067680 on Build GRCh38
rs139541431 Chr.16: 3067567-3067567 on Build GRCh38
rs113075507 Chr.16: 3067421-3067421 on Build GRCh38
rs760128612 Chr.16: 3067533-3067533 on Build GRCh38
rs746607963 Chr.16: 3067285-3067285 on Build GRCh38
rs750824317 Chr.16: 3067480-3067480 on Build GRCh38
rs765511176 Chr.16: 3067523-3067523 on Build GRCh38
rs541927223 Chr.16: 3067666-3067666 on Build GRCh38
rs775405103 Chr.16: 3067514-3067514 on Build GRCh38
rs779022695 Chr.16: 3067294-3067311 on Build GRCh38
rs1009799724 Chr.16: 3067264-3067291 on Build GRCh38
rs563456560 Chr.16: 3067363-3067363 on Build GRCh38
rs757809713 Chr.16: 3067423-3067423 on Build GRCh38
rs770094831 Chr.16: 3067275-3067276 on Build GRCh38
rs745782002 Chr.16: 3067511-3067511 on Build GRCh38
rs916507315 Chr.16: 3067510-3067510 on Build GRCh38

View Full Product Details