Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4739738-4740271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf71
Entrez Gene ID
146562
Gene Name
chromosome 16 open reading frame 71
Gene Aliases
-
UniGene
Hs.602738
Gene Chromosome Location
Chr.16: 4734288-4749396 on Build GRCh38
Transcripts
NM_139170.2 XM_005255144.3 XM_017022975.1 XM_017022976.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667006 c.281T>A p.V94D Chr.16: 4740157-4740157 on Build GRCh38
6614564 c.283C>A p.L95M Chr.16: 4740159-4740159 on Build GRCh38
3667923 c.308A>C p.E103A Chr.16: 4740184-4740184 on Build GRCh38
4060873 c.358G>A p.G120R Chr.16: 4740234-4740234 on Build GRCh38
2832790 c.372C>A p.G124G Chr.16: 4740248-4740248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995568073 Chr.16: 4739970-4739970 on Build GRCh38
rs553744194 Chr.16: 4740046-4740046 on Build GRCh38
rs912283211 Chr.16: 4740075-4740075 on Build GRCh38
rs368037408 Chr.16: 4739974-4739974 on Build GRCh38
rs914653012 Chr.16: 4740190-4740190 on Build GRCh38
rs546475599 Chr.16: 4740045-4740045 on Build GRCh38
rs969910253 Chr.16: 4740123-4740123 on Build GRCh38
rs555019596 Chr.16: 4739902-4739902 on Build GRCh38
rs372967863 Chr.16: 4740218-4740218 on Build GRCh38
rs751572839 Chr.16: 4740231-4740231 on Build GRCh38
rs749965261 Chr.16: 4740208-4740208 on Build GRCh38
rs947390396 Chr.16: 4739764-4739764 on Build GRCh38
rs199709647 Chr.16: 4739801-4739801 on Build GRCh38
rs867747991 Chr.16: 4740077-4740077 on Build GRCh38
rs753755250 Chr.16: 4740129-4740130 on Build GRCh38
rs746889459 Chr.16: 4740122-4740122 on Build GRCh38
rs1044015157 Chr.16: 4739881-4739882 on Build GRCh38
rs959758753 Chr.16: 4739770-4739770 on Build GRCh38
rs376548562 Chr.16: 4740234-4740234 on Build GRCh38
rs912920665 Chr.16: 4739796-4739796 on Build GRCh38
rs773441195 Chr.16: 4740152-4740152 on Build GRCh38
rs748470606 Chr.16: 4740146-4740146 on Build GRCh38
rs912697152 Chr.16: 4740078-4740078 on Build GRCh38
rs575015308 Chr.16: 4739981-4739981 on Build GRCh38
rs1043684890 Chr.16: 4739785-4739785 on Build GRCh38
rs756430216 Chr.16: 4740247-4740247 on Build GRCh38
rs763565640 Chr.16: 4740008-4740008 on Build GRCh38
rs144534429 Chr.16: 4739896-4739896 on Build GRCh38
rs537215516 Chr.16: 4739982-4739982 on Build GRCh38
rs985535110 Chr.16: 4740058-4740058 on Build GRCh38
rs1026912023 Chr.16: 4740027-4740027 on Build GRCh38
rs771600065 Chr.16: 4740168-4740168 on Build GRCh38
rs1021703727 Chr.16: 4739781-4739781 on Build GRCh38
rs902408860 Chr.16: 4739838-4739838 on Build GRCh38
rs962404299 Chr.16: 4739943-4739943 on Build GRCh38
rs111697257 Chr.16: 4740059-4740060 on Build GRCh38
rs1049896989 Chr.16: 4739930-4739930 on Build GRCh38
rs573260997 Chr.16: 4740134-4740134 on Build GRCh38
rs767988691 Chr.16: 4740195-4740195 on Build GRCh38
rs368696679 Chr.16: 4740193-4740193 on Build GRCh38
rs372515817 Chr.16: 4740024-4740024 on Build GRCh38
rs975510782 Chr.16: 4740186-4740186 on Build GRCh38
rs374310136 Chr.16: 4740150-4740150 on Build GRCh38
rs935209277 Chr.16: 4740230-4740230 on Build GRCh38
rs577418907 Chr.16: 4740014-4740014 on Build GRCh38
rs999383178 Chr.16: 4739874-4739874 on Build GRCh38
rs781046898 Chr.16: 4740243-4740243 on Build GRCh38
rs768546358 Chr.16: 4740130-4740130 on Build GRCh38
rs61731839 Chr.16: 4740203-4740203 on Build GRCh38
rs368134916 Chr.16: 4740110-4740110 on Build GRCh38
rs964100330 Chr.16: 4740095-4740095 on Build GRCh38
rs1023386661 Chr.16: 4739926-4739926 on Build GRCh38
rs112164430 Chr.16: 4739997-4739997 on Build GRCh38
rs1037508538 Chr.16: 4739989-4739989 on Build GRCh38
rs1056670620 Chr.16: 4739890-4739890 on Build GRCh38
rs953777888 Chr.16: 4740040-4740040 on Build GRCh38
rs988953504 Chr.16: 4739788-4739788 on Build GRCh38
rs765229464 Chr.16: 4740167-4740168 on Build GRCh38
rs529907872 Chr.16: 4740058-4740059 on Build GRCh38
rs779517240 Chr.16: 4740220-4740220 on Build GRCh38
rs779926441 Chr.16: 4740112-4740112 on Build GRCh38
rs569448019 Chr.16: 4740207-4740207 on Build GRCh38
rs774966816 Chr.16: 4740181-4740181 on Build GRCh38
rs757177276 Chr.16: 4740139-4740139 on Build GRCh38
rs367806863 Chr.16: 4740244-4740244 on Build GRCh38
rs763081018 Chr.16: 4740153-4740153 on Build GRCh38

View Full Product Details