Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 8853776-8854317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARHSP1
Entrez Gene ID
23589
Gene Name
calcium regulated heat stable protein 1
Gene Aliases
CRHSP-24, CRHSP24, CSDC1
UniGene
Hs.632184
Gene Chromosome Location
Chr.16: 8852942-8869012 on Build GRCh38
Transcripts
NM_001042476.2 NM_001278260.1 NM_001278261.1 NM_001278262.1 NM_001278263.1 NM_001278264.1 NM_001278265.1 NM_001278266.1 NM_014316.3 XM_005255228.4 XM_005255229.4 XM_011522444.2

SNP Detail

SNP ID Chromosome Location
rs947309768 Chr.16: 8854166-8854166 on Build GRCh38
rs1677479 Chr.16: 8853867-8853867 on Build GRCh38
rs936536947 Chr.16: 8853840-8853840 on Build GRCh38
rs977981048 Chr.16: 8853962-8853962 on Build GRCh38
rs545564869 Chr.16: 8854041-8854041 on Build GRCh38
rs912381383 Chr.16: 8854147-8854147 on Build GRCh38
rs1016439131 Chr.16: 8854207-8854207 on Build GRCh38
rs964490371 Chr.16: 8853922-8853922 on Build GRCh38
rs913392503 Chr.16: 8853849-8853849 on Build GRCh38
rs1040064455 Chr.16: 8854052-8854052 on Build GRCh38
rs993668981 Chr.16: 8853864-8853864 on Build GRCh38
rs997235090 Chr.16: 8854076-8854076 on Build GRCh38
rs767933669 Chr.16: 8854141-8854141 on Build GRCh38
rs751899123 Chr.16: 8854215-8854215 on Build GRCh38
rs966339317 Chr.16: 8854102-8854102 on Build GRCh38
rs907186685 Chr.16: 8854010-8854010 on Build GRCh38
rs929414671 Chr.16: 8854211-8854211 on Build GRCh38
rs1006922888 Chr.16: 8854061-8854061 on Build GRCh38
rs888010908 Chr.16: 8854046-8854046 on Build GRCh38
rs967077220 Chr.16: 8854081-8854081 on Build GRCh38
rs546877361 Chr.16: 8853848-8853848 on Build GRCh38
rs752765930 Chr.16: 8854157-8854157 on Build GRCh38
rs987553324 Chr.16: 8854220-8854220 on Build GRCh38
rs528465803 Chr.16: 8854080-8854080 on Build GRCh38
rs1054950198 Chr.16: 8853986-8853986 on Build GRCh38
rs772996952 Chr.16: 8853945-8853945 on Build GRCh38
rs536305154 Chr.16: 8854205-8854205 on Build GRCh38
rs35101709 Chr.16: 8854094-8854094 on Build GRCh38
rs556904653 Chr.16: 8853947-8853947 on Build GRCh38
rs1006440106 Chr.16: 8854219-8854219 on Build GRCh38
rs554832186 Chr.16: 8854168-8854169 on Build GRCh38
rs903650632 Chr.16: 8853823-8853823 on Build GRCh38
rs972022935 Chr.16: 8854117-8854117 on Build GRCh38
rs987585065 Chr.16: 8854140-8854140 on Build GRCh38
rs187137444 Chr.16: 8854276-8854276 on Build GRCh38
rs999316813 Chr.16: 8854260-8854260 on Build GRCh38
rs896989653 Chr.16: 8853893-8853893 on Build GRCh38
rs1039696545 Chr.16: 8854221-8854221 on Build GRCh38
rs572877051 Chr.16: 8853996-8853996 on Build GRCh38
rs370150824 Chr.16: 8853992-8853992 on Build GRCh38
rs767626866 Chr.16: 8854246-8854246 on Build GRCh38
rs142250560 Chr.16: 8854038-8854038 on Build GRCh38
rs766680945 Chr.16: 8853901-8853901 on Build GRCh38
rs560902175 Chr.16: 8854070-8854070 on Build GRCh38
rs554698881 Chr.16: 8854222-8854222 on Build GRCh38
rs993470611 Chr.16: 8854016-8854016 on Build GRCh38
rs945267721 Chr.16: 8853811-8853811 on Build GRCh38
rs192462211 Chr.16: 8854021-8854021 on Build GRCh38
rs936615293 Chr.16: 8853979-8853979 on Build GRCh38
rs916374480 Chr.16: 8854142-8854142 on Build GRCh38
rs548940318 Chr.16: 8853808-8853808 on Build GRCh38
rs550716793 Chr.16: 8854186-8854186 on Build GRCh38
rs1042336532 Chr.16: 8853814-8853814 on Build GRCh38
rs1041341828 Chr.16: 8854169-8854169 on Build GRCh38
rs1011320870 Chr.16: 8853959-8853959 on Build GRCh38
rs933365577 Chr.16: 8854131-8854131 on Build GRCh38
rs549077171 Chr.16: 8854067-8854067 on Build GRCh38
rs141826586 Chr.16: 8853843-8853843 on Build GRCh38
rs957981026 Chr.16: 8854130-8854130 on Build GRCh38
rs369494018 Chr.16: 8854178-8854179 on Build GRCh38
rs575382109 Chr.16: 8853953-8853953 on Build GRCh38
rs1024309481 Chr.16: 8853856-8853856 on Build GRCh38
rs182781507 Chr.16: 8854261-8854261 on Build GRCh38
rs1002580262 Chr.16: 8854083-8854083 on Build GRCh38
rs573013047 Chr.16: 8854239-8854239 on Build GRCh38
rs902247402 Chr.16: 8854258-8854258 on Build GRCh38
rs919371781 Chr.16: 8854036-8854036 on Build GRCh38
rs569080442 Chr.16: 8854192-8854192 on Build GRCh38
rs891021044 Chr.16: 8853924-8853924 on Build GRCh38
rs548074802 Chr.16: 8853876-8853877 on Build GRCh38
rs1036430305 Chr.16: 8854035-8854038 on Build GRCh38
rs762755846 Chr.16: 8853978-8853978 on Build GRCh38
rs1013443985 Chr.16: 8853851-8853851 on Build GRCh38
rs909208212 Chr.16: 8853865-8853865 on Build GRCh38
rs571503062 Chr.16: 8854133-8854133 on Build GRCh38
rs190659799 Chr.16: 8854252-8854252 on Build GRCh38
rs567339400 Chr.16: 8853812-8853812 on Build GRCh38
rs1045792633 Chr.16: 8854009-8854009 on Build GRCh38
rs563564605 Chr.16: 8854045-8854045 on Build GRCh38
rs928708904 Chr.16: 8854181-8854181 on Build GRCh38
rs1018372892 Chr.16: 8853844-8853844 on Build GRCh38
rs1041646388 Chr.16: 8854286-8854286 on Build GRCh38
rs909208519 Chr.16: 8854006-8854006 on Build GRCh38
rs947277297 Chr.16: 8854282-8854282 on Build GRCh38
rs530941964 Chr.16: 8853807-8853807 on Build GRCh38
rs139944823 Chr.16: 8853879-8853879 on Build GRCh38
rs374103741 Chr.16: 8853852-8853852 on Build GRCh38
rs756155219 Chr.16: 8854269-8854269 on Build GRCh38
rs928918424 Chr.16: 8853854-8853854 on Build GRCh38
rs1019920488 Chr.16: 8854078-8854078 on Build GRCh38
rs1006375292 Chr.16: 8853905-8853905 on Build GRCh38
rs578210872 Chr.16: 8854039-8854039 on Build GRCh38
rs376874872 Chr.16: 8853967-8853967 on Build GRCh38
rs914430999 Chr.16: 8854272-8854272 on Build GRCh38
rs1032005599 Chr.16: 8854267-8854267 on Build GRCh38
rs977741179 Chr.16: 8854106-8854106 on Build GRCh38
rs1013535417 Chr.16: 8854199-8854199 on Build GRCh38
rs894639735 Chr.16: 8854193-8854193 on Build GRCh38
rs764882927 Chr.16: 8853869-8853869 on Build GRCh38
rs369365316 Chr.16: 8853931-8853931 on Build GRCh38
rs919260510 Chr.16: 8854128-8854129 on Build GRCh38
rs1035870697 Chr.16: 8854189-8854189 on Build GRCh38
rs187869321 Chr.16: 8854158-8854158 on Build GRCh38
rs1030534196 Chr.16: 8854243-8854243 on Build GRCh38
rs890959831 Chr.16: 8854043-8854043 on Build GRCh38
rs183666666 Chr.16: 8854064-8854064 on Build GRCh38
rs372909120 Chr.16: 8853942-8853942 on Build GRCh38
rs972325256 Chr.16: 8854214-8854215 on Build GRCh38
rs907219124 Chr.16: 8853860-8853860 on Build GRCh38
rs759840759 Chr.16: 8854216-8854216 on Build GRCh38
rs925204775 Chr.16: 8853977-8853977 on Build GRCh38
rs536368304 Chr.16: 8853991-8853991 on Build GRCh38
rs942568906 Chr.16: 8854154-8854154 on Build GRCh38
rs544351494 Chr.16: 8854111-8854111 on Build GRCh38
rs1018212780 Chr.16: 8853919-8853919 on Build GRCh38
rs994077903 Chr.16: 8853806-8853806 on Build GRCh38
rs1035786161 Chr.16: 8854095-8854095 on Build GRCh38
rs1007240215 Chr.16: 8853929-8853929 on Build GRCh38
rs900645633 Chr.16: 8854232-8854232 on Build GRCh38

View Full Product Details