Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 22244543-22244983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF2K
Entrez Gene ID
29904
Gene Name
eukaryotic elongation factor 2 kinase
Gene Aliases
HSU93850, eEF-2K
UniGene
Hs.498892
Gene Chromosome Location
Chr.16: 22206264-22288745 on Build GRCh38
Transcripts
NM_013302.3 XM_017023193.1 XM_017023194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4596041 c.285G>T p.M95I Chr.16: 22244668-22244668 on Build GRCh38
968573 c.341G>A p.R114Q Chr.16: 22244724-22244724 on Build GRCh38
3507626 c.268C>T p.Q90* Chr.16: 22244651-22244651 on Build GRCh38
3420850 c.261C>T p.H87H Chr.16: 22244644-22244644 on Build GRCh38
968569 c.330A>G p.E110E Chr.16: 22244713-22244713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897867178 Chr.16: 22244893-22244893 on Build GRCh38
rs931055938 Chr.16: 22244657-22244657 on Build GRCh38
rs766359281 Chr.16: 22244609-22244609 on Build GRCh38
rs758824815 Chr.16: 22244673-22244673 on Build GRCh38
rs754116940 Chr.16: 22244656-22244656 on Build GRCh38
rs748801468 Chr.16: 22244582-22244588 on Build GRCh38
rs780231295 Chr.16: 22244752-22244752 on Build GRCh38
rs770737903 Chr.16: 22244771-22244771 on Build GRCh38
rs976615955 Chr.16: 22244795-22244795 on Build GRCh38
rs1053101457 Chr.16: 22244793-22244793 on Build GRCh38
rs775556478 Chr.16: 22244812-22244812 on Build GRCh38
rs753905865 Chr.16: 22244747-22244747 on Build GRCh38
rs575330838 Chr.16: 22244801-22244801 on Build GRCh38
rs773027044 Chr.16: 22244599-22244599 on Build GRCh38
rs772854122 Chr.16: 22244768-22244768 on Build GRCh38
rs769482109 Chr.16: 22244715-22244715 on Build GRCh38
rs766097584 Chr.16: 22244723-22244723 on Build GRCh38
rs779246947 Chr.16: 22244666-22244666 on Build GRCh38
rs1035615639 Chr.16: 22244694-22244694 on Build GRCh38
rs961570014 Chr.16: 22244907-22244907 on Build GRCh38
rs894437445 Chr.16: 22244925-22244925 on Build GRCh38
rs942009517 Chr.16: 22244890-22244890 on Build GRCh38
rs747658944 Chr.16: 22244714-22244714 on Build GRCh38
rs765047759 Chr.16: 22244586-22244586 on Build GRCh38
rs770395107 Chr.16: 22244763-22244764 on Build GRCh38
rs560299371 Chr.16: 22244889-22244889 on Build GRCh38
rs538079675 Chr.16: 22244843-22244843 on Build GRCh38
rs137985480 Chr.16: 22244651-22244651 on Build GRCh38
rs764269260 Chr.16: 22244732-22244732 on Build GRCh38
rs773981065 Chr.16: 22244772-22244772 on Build GRCh38
rs762801243 Chr.16: 22244721-22244721 on Build GRCh38
rs1039242736 Chr.16: 22244882-22244882 on Build GRCh38
rs747581471 Chr.16: 22244753-22244753 on Build GRCh38
rs893223548 Chr.16: 22244802-22244802 on Build GRCh38
rs758845310 Chr.16: 22244750-22244750 on Build GRCh38
rs373963789 Chr.16: 22244775-22244775 on Build GRCh38
rs1006246737 Chr.16: 22244569-22244569 on Build GRCh38
rs780708612 Chr.16: 22244696-22244696 on Build GRCh38
rs558082428 Chr.16: 22244868-22244868 on Build GRCh38
rs776358573 Chr.16: 22244579-22244579 on Build GRCh38
rs1021329524 Chr.16: 22244943-22244943 on Build GRCh38
rs1036468866 Chr.16: 22244891-22244891 on Build GRCh38
rs961018839 Chr.16: 22244607-22244607 on Build GRCh38
rs571646847 Chr.16: 22244583-22244583 on Build GRCh38
rs574050430 Chr.16: 22244945-22244945 on Build GRCh38
rs759346914 Chr.16: 22244728-22244728 on Build GRCh38
rs2303188 Chr.16: 22244577-22244577 on Build GRCh38
rs539041939 Chr.16: 22244641-22244641 on Build GRCh38
rs750410203 Chr.16: 22244756-22244756 on Build GRCh38
rs539865580 Chr.16: 22244831-22244831 on Build GRCh38
rs540591000 Chr.16: 22244884-22244884 on Build GRCh38
rs765347856 Chr.16: 22244779-22244779 on Build GRCh38
rs4783451 Chr.16: 22244872-22244872 on Build GRCh38
rs773524297 Chr.16: 22244818-22244818 on Build GRCh38
rs750671490 Chr.16: 22244749-22244749 on Build GRCh38
rs1002718732 Chr.16: 22244894-22244894 on Build GRCh38
rs567224940 Chr.16: 22244821-22244821 on Build GRCh38
rs569171030 Chr.16: 22244671-22244671 on Build GRCh38
rs775963419 Chr.16: 22244711-22244711 on Build GRCh38
rs1008466303 Chr.16: 22244883-22244883 on Build GRCh38
rs757391871 Chr.16: 22244748-22244748 on Build GRCh38
rs765930672 Chr.16: 22244804-22244804 on Build GRCh38
rs747666172 Chr.16: 22244758-22244758 on Build GRCh38
rs887894764 Chr.16: 22244568-22244568 on Build GRCh38
rs757382136 Chr.16: 22244664-22244664 on Build GRCh38
rs115593975 Chr.16: 22244760-22244760 on Build GRCh38
rs768234039 Chr.16: 22244710-22244710 on Build GRCh38
rs1012821218 Chr.16: 22244941-22244941 on Build GRCh38
rs1050350546 Chr.16: 22244566-22244566 on Build GRCh38
rs1050423536 Chr.16: 22244871-22244871 on Build GRCh38
rs747414948 Chr.16: 22244702-22244702 on Build GRCh38
rs762711344 Chr.16: 22244600-22244600 on Build GRCh38
rs184797021 Chr.16: 22244774-22244774 on Build GRCh38
rs200566569 Chr.16: 22244644-22244644 on Build GRCh38
rs1028931395 Chr.16: 22244857-22244857 on Build GRCh38
rs774362191 Chr.16: 22244724-22244724 on Build GRCh38
rs372042259 Chr.16: 22244754-22244754 on Build GRCh38
rs557860899 Chr.16: 22244832-22244832 on Build GRCh38
rs954323140 Chr.16: 22244574-22244574 on Build GRCh38

View Full Product Details