Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 55820294-55820796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES1
Entrez Gene ID
1066
Gene Name
carboxylesterase 1
Gene Aliases
ACAT, CE-1, CEH, CES2, HMSE, HMSE1, PCE-1, REH, SES1, TGH, hCE-1
UniGene
Hs.558865
Gene Chromosome Location
Chr.16: 55802851-55833186 on Build GRCh38
Transcripts
NM_001025194.1 NM_001025195.1 NM_001266.4 XM_005255774.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
971604 c.737C>A p.S246Y Chr.16: 55820436-55820436 on Build GRCh38
266233 c.789G>C p.K263N Chr.16: 55820384-55820384 on Build GRCh38
971603 c.747C>T p.G249G Chr.16: 55820426-55820426 on Build GRCh38
2835505 c.726C>A p.H242Q Chr.16: 55820447-55820447 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs573500192 Chr.16: 55820369-55820369 on Build GRCh38
rs556015993 Chr.16: 55820498-55820498 on Build GRCh38
rs376173751 Chr.16: 55820446-55820446 on Build GRCh38
rs370512106 Chr.16: 55820748-55820748 on Build GRCh38
rs767685613 Chr.16: 55820514-55820514 on Build GRCh38
rs2307233 Chr.16: 55820540-55820540 on Build GRCh38
rs750549715 Chr.16: 55820664-55820664 on Build GRCh38
rs185574998 Chr.16: 55820637-55820637 on Build GRCh38
rs374207113 Chr.16: 55820361-55820361 on Build GRCh38
rs758070562 Chr.16: 55820491-55820491 on Build GRCh38
rs531927363 Chr.16: 55820588-55820588 on Build GRCh38
rs751000766 Chr.16: 55820461-55820461 on Build GRCh38
rs754202870 Chr.16: 55820522-55820522 on Build GRCh38
rs752167947 Chr.16: 55820324-55820324 on Build GRCh38
rs577829610 Chr.16: 55820501-55820501 on Build GRCh38
rs539001917 Chr.16: 55820337-55820337 on Build GRCh38
rs965546922 Chr.16: 55820673-55820673 on Build GRCh38
rs766770710 Chr.16: 55820484-55820484 on Build GRCh38
rs776311502 Chr.16: 55820359-55820359 on Build GRCh38
rs2307235 Chr.16: 55820543-55820543 on Build GRCh38
rs116480059 Chr.16: 55820580-55820580 on Build GRCh38
rs554649848 Chr.16: 55820365-55820365 on Build GRCh38
rs778068631 Chr.16: 55820390-55820390 on Build GRCh38
rs2307229 Chr.16: 55820500-55820500 on Build GRCh38
rs761789239 Chr.16: 55820512-55820512 on Build GRCh38
rs368338616 Chr.16: 55820353-55820353 on Build GRCh38
rs377420862 Chr.16: 55820354-55820354 on Build GRCh38
rs746914825 Chr.16: 55820427-55820427 on Build GRCh38
rs374934271 Chr.16: 55820708-55820708 on Build GRCh38
rs183136362 Chr.16: 55820682-55820682 on Build GRCh38
rs764313649 Chr.16: 55820381-55820381 on Build GRCh38
rs776419792 Chr.16: 55820440-55820440 on Build GRCh38
rs775454603 Chr.16: 55820499-55820499 on Build GRCh38
rs756857207 Chr.16: 55820493-55820493 on Build GRCh38
rs8192940 Chr.16: 55820602-55820602 on Build GRCh38
rs544043476 Chr.16: 55820495-55820495 on Build GRCh38
rs17850910 Chr.16: 55820456-55820456 on Build GRCh38
rs752078237 Chr.16: 55820382-55820382 on Build GRCh38
rs564310220 Chr.16: 55820630-55820630 on Build GRCh38
rs113867465 Chr.16: 55820725-55820725 on Build GRCh38
rs4784587 Chr.16: 55820561-55820561 on Build GRCh38
rs202228585 Chr.16: 55820426-55820426 on Build GRCh38
rs750762584 Chr.16: 55820417-55820417 on Build GRCh38
rs769765497 Chr.16: 55820443-55820443 on Build GRCh38
rs745645081 Chr.16: 55820442-55820442 on Build GRCh38
rs114124874 Chr.16: 55820574-55820574 on Build GRCh38
rs190722337 Chr.16: 55820649-55820649 on Build GRCh38
rs765381272 Chr.16: 55820362-55820362 on Build GRCh38
rs201251252 Chr.16: 55820389-55820389 on Build GRCh38
rs879762768 Chr.16: 55820773-55820773 on Build GRCh38
rs746505320 Chr.16: 55820492-55820492 on Build GRCh38
rs536699744 Chr.16: 55820761-55820761 on Build GRCh38
rs752308493 Chr.16: 55820489-55820489 on Build GRCh38
rs753032475 Chr.16: 55820367-55820367 on Build GRCh38
rs774705380 Chr.16: 55820506-55820506 on Build GRCh38
rs781565246 Chr.16: 55820383-55820383 on Build GRCh38
rs771630744 Chr.16: 55820346-55820346 on Build GRCh38
rs2307242 Chr.16: 55820334-55820334 on Build GRCh38
rs756635235 Chr.16: 55820333-55820333 on Build GRCh38
rs2307237 Chr.16: 55820347-55820347 on Build GRCh38
rs778331496 Chr.16: 55820344-55820344 on Build GRCh38
rs775828820 Chr.16: 55820445-55820445 on Build GRCh38
rs756412911 Chr.16: 55820479-55820479 on Build GRCh38
rs111399497 Chr.16: 55820683-55820683 on Build GRCh38
rs774589607 Chr.16: 55820452-55820452 on Build GRCh38
rs762317279 Chr.16: 55820455-55820455 on Build GRCh38
rs553853003 Chr.16: 55820345-55820345 on Build GRCh38
rs745862457 Chr.16: 55820497-55820497 on Build GRCh38
rs756571951 Chr.16: 55820422-55820422 on Build GRCh38
rs778524104 Chr.16: 55820425-55820425 on Build GRCh38
rs757827444 Chr.16: 55820635-55820635 on Build GRCh38
rs752065224 Chr.16: 55820632-55820632 on Build GRCh38
rs757616088 Chr.16: 55820329-55820329 on Build GRCh38
rs3214389 Chr.16: 55820626-55820627 on Build GRCh38
rs770785242 Chr.16: 55820428-55820428 on Build GRCh38
rs112115572 Chr.16: 55820539-55820539 on Build GRCh38
rs111651683 Chr.16: 55820736-55820736 on Build GRCh38

View Full Product Details