Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66725726-66726232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1LI2
Entrez Gene ID
1783
Gene Name
dynein cytoplasmic 1 light intermediate chain 2
Gene Aliases
DNCLI2, LIC2
UniGene
Hs.369068
Gene Chromosome Location
Chr.16: 66720893-66751828 on Build GRCh38
Transcripts
NM_001286157.1 NM_001323955.1 NM_006141.2 NR_136658.1 XM_017023008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5851181 c.1363A>T p.T455S Chr.16: 66725843-66725843 on Build GRCh38
4607649 c.1362C>A p.S454R Chr.16: 66725844-66725844 on Build GRCh38
4061683 c.1339C>T p.P447S Chr.16: 66725867-66725867 on Build GRCh38
703928 c.1330C>T p.P444S Chr.16: 66725876-66725876 on Build GRCh38
972197 c.1321A>G p.T441A Chr.16: 66725885-66725885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776129108 Chr.16: 66726033-66726033 on Build GRCh38
rs746190108 Chr.16: 66725949-66725949 on Build GRCh38
rs1057128120 Chr.16: 66726198-66726198 on Build GRCh38
rs2305310 Chr.16: 66726073-66726073 on Build GRCh38
rs757233087 Chr.16: 66725964-66725964 on Build GRCh38
rs143707837 Chr.16: 66725900-66725900 on Build GRCh38
rs760622533 Chr.16: 66725910-66725910 on Build GRCh38
rs201662578 Chr.16: 66725783-66725783 on Build GRCh38
rs758909886 Chr.16: 66725981-66725993 on Build GRCh38
rs755508824 Chr.16: 66725980-66725980 on Build GRCh38
rs762283465 Chr.16: 66725878-66725878 on Build GRCh38
rs946018463 Chr.16: 66726159-66726159 on Build GRCh38
rs768910291 Chr.16: 66725957-66725960 on Build GRCh38
rs942722967 Chr.16: 66726079-66726079 on Build GRCh38
rs17852314 Chr.16: 66725832-66725832 on Build GRCh38
rs543659167 Chr.16: 66726167-66726167 on Build GRCh38
rs927385562 Chr.16: 66726170-66726170 on Build GRCh38
rs559136731 Chr.16: 66725945-66725945 on Build GRCh38
rs768051013 Chr.16: 66725894-66725894 on Build GRCh38
rs368490019 Chr.16: 66725960-66725960 on Build GRCh38
rs529958953 Chr.16: 66726078-66726078 on Build GRCh38
rs199572341 Chr.16: 66725959-66725959 on Build GRCh38
rs769380675 Chr.16: 66725780-66725780 on Build GRCh38
rs547330156 Chr.16: 66726182-66726182 on Build GRCh38
rs768894802 Chr.16: 66725867-66725867 on Build GRCh38
rs907084001 Chr.16: 66726178-66726178 on Build GRCh38
rs762050045 Chr.16: 66726196-66726196 on Build GRCh38
rs1028719521 Chr.16: 66726060-66726060 on Build GRCh38
rs755575801 Chr.16: 66726157-66726157 on Build GRCh38
rs972987241 Chr.16: 66725944-66725944 on Build GRCh38
rs2305311 Chr.16: 66725779-66725779 on Build GRCh38
rs748971013 Chr.16: 66725858-66725858 on Build GRCh38
rs541097200 Chr.16: 66726046-66726046 on Build GRCh38
rs1000045237 Chr.16: 66726047-66726047 on Build GRCh38
rs1003094282 Chr.16: 66725755-66725755 on Build GRCh38
rs781238558 Chr.16: 66725975-66725975 on Build GRCh38
rs953273568 Chr.16: 66726066-66726066 on Build GRCh38
rs1055993258 Chr.16: 66726197-66726197 on Build GRCh38
rs1054544043 Chr.16: 66726119-66726119 on Build GRCh38
rs1001522809 Chr.16: 66725797-66725797 on Build GRCh38
rs780334698 Chr.16: 66725992-66725997 on Build GRCh38
rs536917080 Chr.16: 66725773-66725773 on Build GRCh38
rs139479968 Chr.16: 66725871-66725871 on Build GRCh38
rs779423180 Chr.16: 66725963-66725963 on Build GRCh38
rs931489660 Chr.16: 66725752-66725752 on Build GRCh38
rs745827809 Chr.16: 66725979-66725979 on Build GRCh38
rs1015922794 Chr.16: 66725845-66725845 on Build GRCh38
rs770260608 Chr.16: 66726005-66726005 on Build GRCh38
rs1034115194 Chr.16: 66725759-66725759 on Build GRCh38
rs753942143 Chr.16: 66725957-66725957 on Build GRCh38
rs572489612 Chr.16: 66726041-66726041 on Build GRCh38
rs527254718 Chr.16: 66726135-66726135 on Build GRCh38
rs774961072 Chr.16: 66725795-66725795 on Build GRCh38
rs750892737 Chr.16: 66725962-66725963 on Build GRCh38
rs931458521 Chr.16: 66726160-66726160 on Build GRCh38
rs918851459 Chr.16: 66726138-66726138 on Build GRCh38

View Full Product Details