Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28759723-28760268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM222B
Entrez Gene ID
55731
Gene Name
family with sequence similarity 222 member B
Gene Aliases
C17orf63
UniGene
Hs.564533
Gene Chromosome Location
Chr.17: 28755978-28855036 on Build GRCh38
Transcripts
NM_001077498.2 NM_001288631.1 NM_001288632.1 NM_001288633.1 NM_001288634.1 NM_001288635.1 NM_001288636.1 NM_001288637.1 NM_001288638.1 NM_001288639.1 NM_001288640.1 NM_018182.3 XM_011524986.2 XM_011524987.1 XM_011524988.2 XM_017024831.1 XM_017024832.1 XM_017024833.1 XM_017024834.1 XM_017024835.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198254 c.128C>T p.A43V Chr.17: 28759831-28759831 on Build GRCh38
5795258 c.225T>C p.R75R Chr.17: 28759734-28759734 on Build GRCh38
5740924 c.130G>A p.E44K Chr.17: 28759829-28759829 on Build GRCh38
6464723 c.131A>C p.E44A Chr.17: 28759828-28759828 on Build GRCh38
7332840 c.219C>G p.H73Q Chr.17: 28759740-28759740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541156622 Chr.17: 28760211-28760211 on Build GRCh38
rs920078247 Chr.17: 28760094-28760094 on Build GRCh38
rs555230704 Chr.17: 28759818-28759818 on Build GRCh38
rs998752870 Chr.17: 28759955-28759955 on Build GRCh38
rs751701400 Chr.17: 28760017-28760017 on Build GRCh38
rs376744803 Chr.17: 28759749-28759749 on Build GRCh38
rs547863108 Chr.17: 28760092-28760092 on Build GRCh38
rs184296739 Chr.17: 28760106-28760106 on Build GRCh38
rs908807682 Chr.17: 28760134-28760134 on Build GRCh38
rs756237672 Chr.17: 28759823-28759823 on Build GRCh38
rs191869506 Chr.17: 28760063-28760063 on Build GRCh38
rs745936897 Chr.17: 28759819-28759819 on Build GRCh38
rs757434071 Chr.17: 28759807-28759807 on Build GRCh38
rs992572538 Chr.17: 28760018-28760018 on Build GRCh38
rs375690040 Chr.17: 28759913-28759913 on Build GRCh38
rs969870103 Chr.17: 28760053-28760053 on Build GRCh38
rs544156777 Chr.17: 28759970-28759970 on Build GRCh38
rs374617928 Chr.17: 28759803-28759803 on Build GRCh38
rs565571561 Chr.17: 28759944-28759944 on Build GRCh38
rs766545315 Chr.17: 28759940-28759940 on Build GRCh38
rs979952637 Chr.17: 28759976-28759976 on Build GRCh38
rs899529273 Chr.17: 28760163-28760163 on Build GRCh38
rs960238712 Chr.17: 28760215-28760215 on Build GRCh38
rs1041114892 Chr.17: 28760152-28760152 on Build GRCh38
rs930725478 Chr.17: 28759757-28759757 on Build GRCh38
rs956701911 Chr.17: 28760001-28760001 on Build GRCh38
rs971287306 Chr.17: 28759975-28759975 on Build GRCh38
rs1050636288 Chr.17: 28760180-28760180 on Build GRCh38
rs368944553 Chr.17: 28759794-28759794 on Build GRCh38
rs532017770 Chr.17: 28759954-28759954 on Build GRCh38
rs377055691 Chr.17: 28759785-28759785 on Build GRCh38
rs543611183 Chr.17: 28760114-28760114 on Build GRCh38
rs1021458591 Chr.17: 28759945-28759945 on Build GRCh38
rs749759030 Chr.17: 28759859-28759859 on Build GRCh38
rs765251483 Chr.17: 28759791-28759791 on Build GRCh38
rs1032737024 Chr.17: 28759936-28759936 on Build GRCh38
rs375228705 Chr.17: 28759748-28759748 on Build GRCh38
rs1034154283 Chr.17: 28759976-28759976 on Build GRCh38
rs996656229 Chr.17: 28759874-28759874 on Build GRCh38
rs377722416 Chr.17: 28759805-28759805 on Build GRCh38
rs117329750 Chr.17: 28760200-28760200 on Build GRCh38
rs879023578 Chr.17: 28759962-28759962 on Build GRCh38
rs1042851822 Chr.17: 28760179-28760179 on Build GRCh38
rs943677197 Chr.17: 28760138-28760138 on Build GRCh38
rs562263244 Chr.17: 28759982-28759982 on Build GRCh38
rs186922550 Chr.17: 28759872-28759872 on Build GRCh38
rs758096239 Chr.17: 28759802-28759802 on Build GRCh38
rs907934804 Chr.17: 28760098-28760098 on Build GRCh38
rs903985178 Chr.17: 28759870-28759870 on Build GRCh38
rs866526557 Chr.17: 28759847-28759847 on Build GRCh38
rs996386080 Chr.17: 28759827-28759827 on Build GRCh38
rs780509836 Chr.17: 28759831-28759831 on Build GRCh38
rs748405926 Chr.17: 28760223-28760223 on Build GRCh38
rs540612554 Chr.17: 28759938-28759938 on Build GRCh38
rs917975544 Chr.17: 28760049-28760049 on Build GRCh38
rs951885280 Chr.17: 28760010-28760010 on Build GRCh38
rs904035779 Chr.17: 28760184-28760185 on Build GRCh38
rs955920432 Chr.17: 28759941-28759941 on Build GRCh38
rs140820891 Chr.17: 28760171-28760171 on Build GRCh38
rs923227762 Chr.17: 28760132-28760132 on Build GRCh38
rs998700300 Chr.17: 28760207-28760207 on Build GRCh38
rs1000364363 Chr.17: 28759881-28759881 on Build GRCh38

View Full Product Details