Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32207790-32208301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCC
Entrez Gene ID
55721
Gene Name
IQ motif containing C
Gene Aliases
-
UniGene
Hs.274356
Gene Chromosome Location
Chr.1: 32205635-32208687 on Build GRCh38
Transcripts
NM_001160042.1 NM_018134.2 XM_011541795.2 XM_017001814.1 XM_017001815.1 XM_017001817.1 XM_017001818.1 XM_017001819.1 XM_017001821.1 XM_017001827.1 XM_017001828.1

Gene Symbol
TMEM234
Entrez Gene ID
56063
Gene Name
transmembrane protein 234
Gene Aliases
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7
UniGene
Hs.272299
Gene Chromosome Location
Chr.1: 32204807-32222371 on Build GRCh38
Transcripts
NM_001160042.1 NM_018134.2 XM_011541795.2 XM_017001814.1 XM_017001815.1 XM_017001817.1 XM_017001818.1 XM_017001819.1 XM_017001821.1 XM_017001827.1 XM_017001828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5596878 c.1513G>A p.G505R Chr.1: 32207954-32207954 on Build GRCh38
7372387 c.1370C>T p.S457L Chr.1: 32207811-32207811 on Build GRCh38
5619528 c.1438A>G p.T480A Chr.1: 32207879-32207879 on Build GRCh38
2077765 c.1401delG p.P470fs*>77 Chr.1: 32207842-32207842 on Build GRCh38
7323998 c.1431C>T p.L477L Chr.1: 32207872-32207872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930750015 Chr.1: 32208144-32208144 on Build GRCh38
rs374159176 Chr.1: 32207997-32207997 on Build GRCh38
rs770229433 Chr.1: 32207853-32207853 on Build GRCh38
rs113657884 Chr.1: 32208024-32208024 on Build GRCh38
rs886310808 Chr.1: 32208200-32208200 on Build GRCh38
rs41306593 Chr.1: 32208072-32208072 on Build GRCh38
rs138692176 Chr.1: 32207983-32207983 on Build GRCh38
rs748819826 Chr.1: 32207925-32207925 on Build GRCh38
rs775071567 Chr.1: 32208043-32208043 on Build GRCh38
rs1051160195 Chr.1: 32208075-32208075 on Build GRCh38
rs747607918 Chr.1: 32207849-32207849 on Build GRCh38
rs374618392 Chr.1: 32207840-32207840 on Build GRCh38
rs960377422 Chr.1: 32207989-32207990 on Build GRCh38
rs41302746 Chr.1: 32208265-32208265 on Build GRCh38
rs994716142 Chr.1: 32208253-32208253 on Build GRCh38
rs753105271 Chr.1: 32208001-32208001 on Build GRCh38
rs4628547 Chr.1: 32208089-32208089 on Build GRCh38
rs968824308 Chr.1: 32207832-32207832 on Build GRCh38
rs201819910 Chr.1: 32207833-32207833 on Build GRCh38
rs767640917 Chr.1: 32208000-32208000 on Build GRCh38
rs766482440 Chr.1: 32207990-32207990 on Build GRCh38
rs756496848 Chr.1: 32208005-32208005 on Build GRCh38
rs939275480 Chr.1: 32208211-32208211 on Build GRCh38
rs920648632 Chr.1: 32208114-32208114 on Build GRCh38
rs36068145 Chr.1: 32207911-32207911 on Build GRCh38
rs765176429 Chr.1: 32208079-32208079 on Build GRCh38
rs369036950 Chr.1: 32207813-32207813 on Build GRCh38
rs150341307 Chr.1: 32207913-32207913 on Build GRCh38
rs770706512 Chr.1: 32207863-32207863 on Build GRCh38
rs200765541 Chr.1: 32207915-32207915 on Build GRCh38
rs924074431 Chr.1: 32208052-32208052 on Build GRCh38
rs760611364 Chr.1: 32207886-32207886 on Build GRCh38
rs746804829 Chr.1: 32208044-32208044 on Build GRCh38
rs779093097 Chr.1: 32207956-32207956 on Build GRCh38
rs368769892 Chr.1: 32207843-32207843 on Build GRCh38
rs900416176 Chr.1: 32208201-32208201 on Build GRCh38
rs757314607 Chr.1: 32207842-32207842 on Build GRCh38
rs762996059 Chr.1: 32207988-32207988 on Build GRCh38
rs751666356 Chr.1: 32207992-32207992 on Build GRCh38
rs373839136 Chr.1: 32207867-32207867 on Build GRCh38
rs779009416 Chr.1: 32207836-32207836 on Build GRCh38
rs894848735 Chr.1: 32207971-32207971 on Build GRCh38
rs755301388 Chr.1: 32207912-32207912 on Build GRCh38
rs772725001 Chr.1: 32207856-32207856 on Build GRCh38
rs764034440 Chr.1: 32207841-32207842 on Build GRCh38
rs866842577 Chr.1: 32208012-32208012 on Build GRCh38
rs139414487 Chr.1: 32208068-32208068 on Build GRCh38
rs767407546 Chr.1: 32208109-32208109 on Build GRCh38
rs760541130 Chr.1: 32207840-32207840 on Build GRCh38
rs770616565 Chr.1: 32207926-32207926 on Build GRCh38
rs1056277299 Chr.1: 32208214-32208214 on Build GRCh38
rs943424273 Chr.1: 32207860-32207860 on Build GRCh38
rs752466982 Chr.1: 32207875-32207875 on Build GRCh38
rs149524712 Chr.1: 32207939-32207939 on Build GRCh38
rs1021931860 Chr.1: 32208236-32208236 on Build GRCh38
rs778213627 Chr.1: 32207922-32207922 on Build GRCh38
rs368933605 Chr.1: 32208190-32208190 on Build GRCh38
rs775296573 Chr.1: 32207978-32207978 on Build GRCh38
rs757254303 Chr.1: 32207834-32207834 on Build GRCh38
rs201603864 Chr.1: 32207859-32207859 on Build GRCh38
rs767771177 Chr.1: 32208086-32208086 on Build GRCh38
rs763952071 Chr.1: 32207887-32207887 on Build GRCh38
rs750448981 Chr.1: 32207895-32207895 on Build GRCh38
rs760389575 Chr.1: 32207979-32207979 on Build GRCh38
rs768381465 Chr.1: 32208058-32208058 on Build GRCh38
rs748726030 Chr.1: 32207851-32207851 on Build GRCh38
rs942702708 Chr.1: 32208149-32208149 on Build GRCh38
rs758708906 Chr.1: 32208076-32208077 on Build GRCh38
rs1047538248 Chr.1: 32208171-32208171 on Build GRCh38
rs766572463 Chr.1: 32207909-32207909 on Build GRCh38
rs756579957 Chr.1: 32207916-32207916 on Build GRCh38
rs752730373 Chr.1: 32207823-32207823 on Build GRCh38
rs377231837 Chr.1: 32208027-32208027 on Build GRCh38
rs1007316986 Chr.1: 32207876-32207876 on Build GRCh38
rs370022140 Chr.1: 32208032-32208032 on Build GRCh38
rs199929719 Chr.1: 32208035-32208035 on Build GRCh38
rs750590316 Chr.1: 32208014-32208014 on Build GRCh38
rs987479317 Chr.1: 32207845-32207845 on Build GRCh38
rs771873493 Chr.1: 32207950-32207950 on Build GRCh38
rs978926223 Chr.1: 32207999-32207999 on Build GRCh38
rs895069755 Chr.1: 32208226-32208226 on Build GRCh38
rs745478986 Chr.1: 32207949-32207949 on Build GRCh38
rs1011845284 Chr.1: 32208232-32208232 on Build GRCh38
rs759683010 Chr.1: 32208084-32208084 on Build GRCh38
rs758470830 Chr.1: 32207905-32207905 on Build GRCh38
rs1039760138 Chr.1: 32208191-32208191 on Build GRCh38
rs776598316 Chr.1: 32208059-32208059 on Build GRCh38
rs750490141 Chr.1: 32207839-32207839 on Build GRCh38
rs764086303 Chr.1: 32207981-32207981 on Build GRCh38
rs538795732 Chr.1: 32207866-32207866 on Build GRCh38
rs757872694 Chr.1: 32208025-32208025 on Build GRCh38
rs146534863 Chr.1: 32207844-32207844 on Build GRCh38
rs753907631 Chr.1: 32207842-32207844 on Build GRCh38
rs903590783 Chr.1: 32208255-32208255 on Build GRCh38
rs147386897 Chr.1: 32208017-32208017 on Build GRCh38
rs3887970 Chr.1: 32207848-32207848 on Build GRCh38

View Full Product Details