Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49043342-49043841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2BP1
Entrez Gene ID
10642
Gene Name
insulin like growth factor 2 mRNA binding protein 1
Gene Aliases
CRD-BP, CRDBP, IMP-1, IMP1, VICKZ1, ZBP1
UniGene
Hs.144936
Gene Chromosome Location
Chr.17: 48997412-49056145 on Build GRCh38
Transcripts
NM_001160423.1 NM_006546.3 XM_005256955.3 XM_011524201.2 XM_017024022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6538773 c.1130C>G p.P377R Chr.17: 49043480-49043480 on Build GRCh38
6523659 c.1136C>A p.S379* Chr.17: 49043486-49043486 on Build GRCh38
6481083 c.1143C>T p.S381S Chr.17: 49043493-49043493 on Build GRCh38
6081568 c.1175G>A p.G392E Chr.17: 49043525-49043525 on Build GRCh38
6226568 c.1151C>T p.P384L Chr.17: 49043501-49043501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905825794 Chr.17: 49043611-49043611 on Build GRCh38
rs942372706 Chr.17: 49043393-49043393 on Build GRCh38
rs959830352 Chr.17: 49043799-49043799 on Build GRCh38
rs572651803 Chr.17: 49043458-49043458 on Build GRCh38
rs112793864 Chr.17: 49043708-49043708 on Build GRCh38
rs532334217 Chr.17: 49043707-49043707 on Build GRCh38
rs759335300 Chr.17: 49043384-49043384 on Build GRCh38
rs182261413 Chr.17: 49043730-49043730 on Build GRCh38
rs369774252 Chr.17: 49043505-49043505 on Build GRCh38
rs547556690 Chr.17: 49043767-49043767 on Build GRCh38
rs755385570 Chr.17: 49043585-49043585 on Build GRCh38
rs766558932 Chr.17: 49043811-49043811 on Build GRCh38
rs1016367614 Chr.17: 49043597-49043597 on Build GRCh38
rs1001577874 Chr.17: 49043654-49043654 on Build GRCh38
rs780532469 Chr.17: 49043573-49043573 on Build GRCh38
rs745695988 Chr.17: 49043366-49043366 on Build GRCh38
rs371972898 Chr.17: 49043695-49043695 on Build GRCh38
rs936745394 Chr.17: 49043612-49043612 on Build GRCh38
rs773962841 Chr.17: 49043383-49043383 on Build GRCh38
rs764113447 Chr.17: 49043535-49043535 on Build GRCh38
rs976415151 Chr.17: 49043751-49043751 on Build GRCh38
rs923505956 Chr.17: 49043792-49043792 on Build GRCh38
rs146124273 Chr.17: 49043518-49043518 on Build GRCh38
rs769324443 Chr.17: 49043501-49043501 on Build GRCh38
rs142951853 Chr.17: 49043493-49043493 on Build GRCh38
rs373963827 Chr.17: 49043600-49043600 on Build GRCh38
rs779208625 Chr.17: 49043445-49043445 on Build GRCh38
rs747493051 Chr.17: 49043496-49043496 on Build GRCh38
rs773284383 Chr.17: 49043621-49043621 on Build GRCh38
rs1033850654 Chr.17: 49043797-49043797 on Build GRCh38
rs879156967 Chr.17: 49043549-49043549 on Build GRCh38
rs756094518 Chr.17: 49043407-49043407 on Build GRCh38
rs567378518 Chr.17: 49043800-49043800 on Build GRCh38
rs556174067 Chr.17: 49043367-49043367 on Build GRCh38
rs879657888 Chr.17: 49043710-49043710 on Build GRCh38
rs775305931 Chr.17: 49043517-49043517 on Build GRCh38
rs759245152 Chr.17: 49043513-49043513 on Build GRCh38
rs528375225 Chr.17: 49043700-49043700 on Build GRCh38
rs372768083 Chr.17: 49043412-49043412 on Build GRCh38
rs760882569 Chr.17: 49043764-49043764 on Build GRCh38
rs149283508 Chr.17: 49043558-49043558 on Build GRCh38
rs778332623 Chr.17: 49043405-49043407 on Build GRCh38
rs1054280201 Chr.17: 49043624-49043624 on Build GRCh38
rs752619053 Chr.17: 49043394-49043394 on Build GRCh38
rs978878732 Chr.17: 49043721-49043721 on Build GRCh38
rs750656941 Chr.17: 49043567-49043567 on Build GRCh38
rs992352209 Chr.17: 49043802-49043802 on Build GRCh38
rs2289636 Chr.17: 49043414-49043414 on Build GRCh38
rs772080581 Chr.17: 49043568-49043568 on Build GRCh38
rs978553444 Chr.17: 49043365-49043365 on Build GRCh38
rs781023978 Chr.17: 49043364-49043364 on Build GRCh38
rs995251998 Chr.17: 49043769-49043769 on Build GRCh38
rs899700353 Chr.17: 49043686-49043686 on Build GRCh38
rs754371094 Chr.17: 49043590-49043613 on Build GRCh38
rs201439087 Chr.17: 49043562-49043562 on Build GRCh38
rs748846744 Chr.17: 49043504-49043504 on Build GRCh38
rs576078717 Chr.17: 49043369-49043369 on Build GRCh38
rs767277963 Chr.17: 49043515-49043515 on Build GRCh38
rs144490638 Chr.17: 49043746-49043746 on Build GRCh38
rs753801728 Chr.17: 49043556-49043556 on Build GRCh38
rs968594961 Chr.17: 49043718-49043718 on Build GRCh38
rs767367618 Chr.17: 49043389-49043389 on Build GRCh38
rs377397969 Chr.17: 49043494-49043494 on Build GRCh38
rs140483006 Chr.17: 49043424-49043424 on Build GRCh38

View Full Product Details