Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82037536-82038048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCXR
Entrez Gene ID
51181
Gene Name
dicarbonyl and L-xylulose reductase
Gene Aliases
DCR, HCR2, HCRII, KIDCR, P34H, PNTSU, SDR20C1, XR
UniGene
Hs.9857
Gene Chromosome Location
Chr.17: 82035881-82037697 on Build GRCh38
Transcripts
NM_001195218.1 NM_016286.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3691949 c.52+1G>A p.? Chr.17: 82037630-82037630 on Build GRCh38
5797925 c.21C>T p.G7G Chr.17: 82037662-82037662 on Build GRCh38
6334754 c.19G>T p.G7C Chr.17: 82037664-82037664 on Build GRCh38
1680163 c.47G>A p.G16D Chr.17: 82037636-82037636 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs918730632 Chr.17: 82037640-82037640 on Build GRCh38
rs539904551 Chr.17: 82037900-82037900 on Build GRCh38
rs759349750 Chr.17: 82037691-82037691 on Build GRCh38
rs532681628 Chr.17: 82037726-82037726 on Build GRCh38
rs758559682 Chr.17: 82037581-82037581 on Build GRCh38
rs766790508 Chr.17: 82037919-82037919 on Build GRCh38
rs370735627 Chr.17: 82037712-82037712 on Build GRCh38
rs978001696 Chr.17: 82037708-82037708 on Build GRCh38
rs957421378 Chr.17: 82037785-82037785 on Build GRCh38
rs752727330 Chr.17: 82037661-82037661 on Build GRCh38
rs746709672 Chr.17: 82037685-82037685 on Build GRCh38
rs542452079 Chr.17: 82037848-82037848 on Build GRCh38
rs770424700 Chr.17: 82037618-82037618 on Build GRCh38
rs146030042 Chr.17: 82037960-82037960 on Build GRCh38
rs570468659 Chr.17: 82037982-82037982 on Build GRCh38
rs781714947 Chr.17: 82037611-82037611 on Build GRCh38
rs577283977 Chr.17: 82037795-82037795 on Build GRCh38
rs902356674 Chr.17: 82037563-82037563 on Build GRCh38
rs757636650 Chr.17: 82037668-82037668 on Build GRCh38
rs908943914 Chr.17: 82037776-82037776 on Build GRCh38
rs1046520992 Chr.17: 82037856-82037856 on Build GRCh38
rs906216002 Chr.17: 82037600-82037600 on Build GRCh38
rs569461463 Chr.17: 82037734-82037734 on Build GRCh38
rs561089120 Chr.17: 82037850-82037850 on Build GRCh38
rs1025215611 Chr.17: 82037621-82037621 on Build GRCh38
rs566974993 Chr.17: 82037743-82037743 on Build GRCh38
rs1024952516 Chr.17: 82037737-82037737 on Build GRCh38
rs757924992 Chr.17: 82037728-82037728 on Build GRCh38
rs776627238 Chr.17: 82037655-82037655 on Build GRCh38
rs528179712 Chr.17: 82037855-82037855 on Build GRCh38
rs1006850601 Chr.17: 82037753-82037753 on Build GRCh38
rs113364103 Chr.17: 82037614-82037615 on Build GRCh38
rs375243154 Chr.17: 82037630-82037630 on Build GRCh38
rs1018698597 Chr.17: 82037771-82037771 on Build GRCh38
rs191302793 Chr.17: 82037662-82037662 on Build GRCh38
rs555093314 Chr.17: 82037741-82037741 on Build GRCh38
rs957685876 Chr.17: 82037676-82037676 on Build GRCh38
rs570865890 Chr.17: 82037723-82037723 on Build GRCh38
rs994405448 Chr.17: 82037738-82037738 on Build GRCh38
rs750826734 Chr.17: 82037671-82037671 on Build GRCh38
rs375983320 Chr.17: 82037589-82037589 on Build GRCh38
rs755040105 Chr.17: 82037553-82037553 on Build GRCh38
rs771814237 Chr.17: 82037631-82037631 on Build GRCh38
rs559008750 Chr.17: 82037788-82037788 on Build GRCh38
rs770693304 Chr.17: 82037652-82037652 on Build GRCh38
rs906531258 Chr.17: 82037858-82037858 on Build GRCh38
rs550404024 Chr.17: 82037854-82037854 on Build GRCh38
rs769074396 Chr.17: 82037670-82037670 on Build GRCh38
rs751786056 Chr.17: 82037665-82037665 on Build GRCh38
rs545318038 Chr.17: 82037817-82037817 on Build GRCh38
rs764352986 Chr.17: 82037664-82037664 on Build GRCh38
rs546188954 Chr.17: 82037704-82037704 on Build GRCh38
rs1018829887 Chr.17: 82037951-82037951 on Build GRCh38
rs770622305 Chr.17: 82037688-82037688 on Build GRCh38
rs767800066 Chr.17: 82037718-82037718 on Build GRCh38
rs12946976 Chr.17: 82037554-82037554 on Build GRCh38
rs1051701423 Chr.17: 82037948-82037948 on Build GRCh38
rs775453313 Chr.17: 82037694-82037694 on Build GRCh38
rs761879059 Chr.17: 82037714-82037714 on Build GRCh38
rs979206825 Chr.17: 82037596-82037619 on Build GRCh38
rs953099760 Chr.17: 82037930-82037930 on Build GRCh38
rs930164906 Chr.17: 82037941-82037941 on Build GRCh38
rs764146677 Chr.17: 82037705-82037705 on Build GRCh38
rs57552134 Chr.17: 82037740-82037740 on Build GRCh38
rs984724429 Chr.17: 82037774-82037774 on Build GRCh38
rs61744795 Chr.17: 82037659-82037659 on Build GRCh38
rs116228945 Chr.17: 82037590-82037590 on Build GRCh38
rs773595537 Chr.17: 82037770-82037770 on Build GRCh38
rs200252005 Chr.17: 82037583-82037583 on Build GRCh38
rs113116886 Chr.17: 82037598-82037598 on Build GRCh38
rs941067237 Chr.17: 82037909-82037909 on Build GRCh38
rs747633916 Chr.17: 82037681-82037681 on Build GRCh38
rs939080286 Chr.17: 82037610-82037610 on Build GRCh38
rs759164616 Chr.17: 82037657-82037657 on Build GRCh38
rs556961899 Chr.17: 82037843-82037843 on Build GRCh38
rs746542885 Chr.17: 82037733-82037733 on Build GRCh38
rs750773700 Chr.17: 82037719-82037719 on Build GRCh38
rs759402588 Chr.17: 82037658-82037658 on Build GRCh38
rs534245399 Chr.17: 82037780-82037780 on Build GRCh38
rs1007564306 Chr.17: 82038012-82038012 on Build GRCh38
rs778506852 Chr.17: 82037622-82037622 on Build GRCh38
rs772783198 Chr.17: 82037643-82037643 on Build GRCh38
rs374705885 Chr.17: 82037730-82037730 on Build GRCh38
rs776354255 Chr.17: 82037689-82037689 on Build GRCh38
rs757611604 Chr.17: 82037595-82037595 on Build GRCh38
rs966544121 Chr.17: 82037798-82037798 on Build GRCh38
rs746440343 Chr.17: 82037617-82037617 on Build GRCh38
rs563522623 Chr.17: 82037578-82037578 on Build GRCh38
rs974423991 Chr.17: 82038013-82038013 on Build GRCh38
rs916246744 Chr.17: 82037883-82037883 on Build GRCh38
rs918747456 Chr.17: 82037937-82037937 on Build GRCh38
rs575438573 Chr.17: 82037846-82037846 on Build GRCh38
rs76351910 Chr.17: 82037932-82037932 on Build GRCh38
rs769687447 Chr.17: 82037692-82037692 on Build GRCh38
rs955427289 Chr.17: 82037666-82037666 on Build GRCh38
rs781341587 Chr.17: 82037670-82037670 on Build GRCh38
rs964187369 Chr.17: 82037979-82037979 on Build GRCh38
rs771533492 Chr.17: 82037682-82037682 on Build GRCh38
rs762303638 Chr.17: 82037673-82037673 on Build GRCh38
rs941098289 Chr.17: 82037729-82037729 on Build GRCh38
rs994203985 Chr.17: 82037896-82037896 on Build GRCh38
rs1026356224 Chr.17: 82037765-82037765 on Build GRCh38
rs778541900 Chr.17: 82037586-82037612 on Build GRCh38
rs780734986 Chr.17: 82037678-82037678 on Build GRCh38
rs984554771 Chr.17: 82037646-82037646 on Build GRCh38
rs12946979 Chr.17: 82037555-82037555 on Build GRCh38
rs895296966 Chr.17: 82037641-82037641 on Build GRCh38

View Full Product Details