Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 10454489-10455025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APCDD1
Entrez Gene ID
147495
Gene Name
APC down-regulated 1
Gene Aliases
B7323, DRAPC1, FP7019, HHS, HTS, HYPT1
UniGene
Hs.293274
Gene Chromosome Location
Chr.18: 10454628-10488701 on Build GRCh38
Transcripts
NM_153000.4 XM_011525603.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6438522 c.5_6CC>GT p.S2C Chr.18: 10454986-10454987 on Build GRCh38
1283446 c.44delC p.L16fs*24 Chr.18: 10455025-10455025 on Build GRCh38
6036841 c.27C>G p.L9L Chr.18: 10455008-10455008 on Build GRCh38
986492 c.4T>C p.S2P Chr.18: 10454985-10454985 on Build GRCh38
5706680 c.1A>G p.M1V Chr.18: 10454982-10454982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767678347 Chr.18: 10454985-10454985 on Build GRCh38
rs746186583 Chr.18: 10454958-10454958 on Build GRCh38
rs934554772 Chr.18: 10454550-10454550 on Build GRCh38
rs1014380215 Chr.18: 10454681-10454681 on Build GRCh38
rs763362220 Chr.18: 10454970-10454970 on Build GRCh38
rs545022643 Chr.18: 10454576-10454576 on Build GRCh38
rs979137051 Chr.18: 10454535-10454535 on Build GRCh38
rs941081145 Chr.18: 10454844-10454844 on Build GRCh38
rs28769876 Chr.18: 10454963-10454963 on Build GRCh38
rs899429835 Chr.18: 10454631-10454631 on Build GRCh38
rs573061567 Chr.18: 10454727-10454727 on Build GRCh38
rs1000670615 Chr.18: 10454925-10454925 on Build GRCh38
rs925917419 Chr.18: 10454767-10454767 on Build GRCh38
rs746528058 Chr.18: 10454954-10454954 on Build GRCh38
rs1008260230 Chr.18: 10454519-10454519 on Build GRCh38
rs1038066894 Chr.18: 10454872-10454872 on Build GRCh38
rs563991960 Chr.18: 10454715-10454715 on Build GRCh38
rs933781558 Chr.18: 10454568-10454568 on Build GRCh38
rs1039117467 Chr.18: 10454880-10454889 on Build GRCh38
rs754793645 Chr.18: 10454923-10454923 on Build GRCh38
rs956034092 Chr.18: 10454943-10454943 on Build GRCh38
rs933834935 Chr.18: 10454831-10454831 on Build GRCh38
rs750625563 Chr.18: 10454991-10454991 on Build GRCh38
rs948740541 Chr.18: 10454530-10454530 on Build GRCh38
rs770032384 Chr.18: 10454961-10454961 on Build GRCh38
rs751703029 Chr.18: 10454726-10454726 on Build GRCh38
rs992891356 Chr.18: 10454521-10454521 on Build GRCh38
rs896035778 Chr.18: 10454904-10454904 on Build GRCh38
rs373981457 Chr.18: 10454944-10454944 on Build GRCh38
rs776449924 Chr.18: 10454957-10454957 on Build GRCh38
rs752598200 Chr.18: 10455003-10455003 on Build GRCh38
rs972245538 Chr.18: 10454520-10454521 on Build GRCh38
rs988686457 Chr.18: 10454799-10454799 on Build GRCh38
rs764939671 Chr.18: 10455001-10455001 on Build GRCh38
rs544311114 Chr.18: 10454908-10454908 on Build GRCh38
rs556273092 Chr.18: 10454520-10454520 on Build GRCh38
rs529111767 Chr.18: 10454873-10454873 on Build GRCh38
rs922474630 Chr.18: 10454801-10454801 on Build GRCh38
rs370904903 Chr.18: 10454934-10454934 on Build GRCh38
rs764048233 Chr.18: 10454666-10454666 on Build GRCh38
rs763563825 Chr.18: 10454660-10454660 on Build GRCh38
rs540559360 Chr.18: 10454772-10454772 on Build GRCh38
rs766241107 Chr.18: 10454959-10454960 on Build GRCh38
rs1047998527 Chr.18: 10454894-10454894 on Build GRCh38
rs770637139 Chr.18: 10454956-10454956 on Build GRCh38
rs577581097 Chr.18: 10454564-10454564 on Build GRCh38
rs764676829 Chr.18: 10454971-10454971 on Build GRCh38
rs1026505515 Chr.18: 10454670-10454670 on Build GRCh38
rs1016988907 Chr.18: 10454611-10454611 on Build GRCh38
rs777131912 Chr.18: 10454953-10454953 on Build GRCh38
rs955980284 Chr.18: 10454797-10454797 on Build GRCh38
rs778821223 Chr.18: 10454931-10454931 on Build GRCh38
rs762887783 Chr.18: 10454937-10454946 on Build GRCh38
rs774349561 Chr.18: 10454973-10454973 on Build GRCh38
rs1052913591 Chr.18: 10454556-10454556 on Build GRCh38
rs745428289 Chr.18: 10454922-10454922 on Build GRCh38
rs910994878 Chr.18: 10454840-10454840 on Build GRCh38
rs761001688 Chr.18: 10455000-10455000 on Build GRCh38
rs561727777 Chr.18: 10454791-10454791 on Build GRCh38
rs960191646 Chr.18: 10454671-10454671 on Build GRCh38
rs978803634 Chr.18: 10454751-10454751 on Build GRCh38
rs866702634 Chr.18: 10454995-10454995 on Build GRCh38
rs1005223103 Chr.18: 10454590-10454590 on Build GRCh38
rs758430408 Chr.18: 10454950-10454950 on Build GRCh38
rs925893921 Chr.18: 10454536-10454536 on Build GRCh38
rs1049989265 Chr.18: 10454839-10454839 on Build GRCh38
rs775883527 Chr.18: 10454602-10454602 on Build GRCh38
rs1049461987 Chr.18: 10454573-10454573 on Build GRCh38
rs761913014 Chr.18: 10454979-10454979 on Build GRCh38
rs867308094 Chr.18: 10454998-10454998 on Build GRCh38

View Full Product Details