Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 30989987-30990489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSC3
Entrez Gene ID
1825
Gene Name
desmocollin 3
Gene Aliases
CDHF3, DSC, DSC1, DSC2, DSC4, HT-CP
UniGene
Hs.41690
Gene Chromosome Location
Chr.18: 30989365-31042815 on Build GRCh38
Transcripts
NM_001941.4 NM_024423.3

SNP Detail

SNP ID Chromosome Location
rs386802081 Chr.18: 30990163-30990169 on Build GRCh38
rs937401250 Chr.18: 30990133-30990133 on Build GRCh38
rs553914931 Chr.18: 30990227-30990227 on Build GRCh38
rs905864650 Chr.18: 30990125-30990125 on Build GRCh38
rs906280962 Chr.18: 30990433-30990433 on Build GRCh38
rs947675610 Chr.18: 30990101-30990101 on Build GRCh38
rs950864222 Chr.18: 30990324-30990324 on Build GRCh38
rs973662952 Chr.18: 30990265-30990265 on Build GRCh38
rs184490554 Chr.18: 30990152-30990152 on Build GRCh38
rs149167450 Chr.18: 30990236-30990236 on Build GRCh38
rs549195664 Chr.18: 30990301-30990301 on Build GRCh38
rs944716791 Chr.18: 30990363-30990363 on Build GRCh38
rs537407948 Chr.18: 30990137-30990137 on Build GRCh38
rs930086274 Chr.18: 30990411-30990411 on Build GRCh38
rs913450583 Chr.18: 30990078-30990078 on Build GRCh38
rs1003187080 Chr.18: 30990461-30990461 on Build GRCh38
rs900738808 Chr.18: 30990071-30990071 on Build GRCh38
rs114470574 Chr.18: 30990337-30990337 on Build GRCh38
rs146467836 Chr.18: 30990373-30990373 on Build GRCh38
rs1003155995 Chr.18: 30990230-30990230 on Build GRCh38
rs765138581 Chr.18: 30990214-30990214 on Build GRCh38
rs920700680 Chr.18: 30990016-30990016 on Build GRCh38
rs1057266814 Chr.18: 30990138-30990138 on Build GRCh38
rs1020582495 Chr.18: 30990190-30990190 on Build GRCh38
rs535004489 Chr.18: 30990394-30990394 on Build GRCh38
rs370193469 Chr.18: 30990033-30990033 on Build GRCh38
rs139613305 Chr.18: 30990065-30990065 on Build GRCh38
rs1047768149 Chr.18: 30990056-30990056 on Build GRCh38
rs192980195 Chr.18: 30990390-30990390 on Build GRCh38
rs976511703 Chr.18: 30990044-30990044 on Build GRCh38
rs774378254 Chr.18: 30990062-30990062 on Build GRCh38
rs187588119 Chr.18: 30990385-30990385 on Build GRCh38
rs1260720 Chr.18: 30990163-30990163 on Build GRCh38
rs1005472224 Chr.18: 30990223-30990223 on Build GRCh38
rs563847058 Chr.18: 30990166-30990166 on Build GRCh38
rs976456109 Chr.18: 30990295-30990295 on Build GRCh38
rs145167573 Chr.18: 30990116-30990116 on Build GRCh38
rs1260721 Chr.18: 30990169-30990169 on Build GRCh38
rs1010360010 Chr.18: 30990403-30990403 on Build GRCh38
rs559503147 Chr.18: 30990023-30990023 on Build GRCh38
rs1260719 Chr.18: 30990059-30990059 on Build GRCh38
rs530406498 Chr.18: 30990407-30990407 on Build GRCh38
rs149990660 Chr.18: 30990070-30990070 on Build GRCh38
rs549791808 Chr.18: 30990264-30990264 on Build GRCh38
rs535398738 Chr.18: 30990202-30990202 on Build GRCh38
rs936235755 Chr.18: 30990386-30990386 on Build GRCh38
rs920407331 Chr.18: 30990306-30990306 on Build GRCh38
rs911969999 Chr.18: 30990342-30990342 on Build GRCh38

View Full Product Details