Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 59431180-59431723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCBE1
Entrez Gene ID
147372
Gene Name
collagen and calcium binding EGF domains 1
Gene Aliases
HKLLS1
UniGene
Hs.34333
Gene Chromosome Location
Chr.18: 59430939-59698181 on Build GRCh38
Transcripts
NM_133459.3

SNP Detail

SNP ID Chromosome Location
rs921529780 Chr.18: 59431239-59431239 on Build GRCh38
rs1016219877 Chr.18: 59431256-59431259 on Build GRCh38
rs761212243 Chr.18: 59431586-59431586 on Build GRCh38
rs757909965 Chr.18: 59431499-59431499 on Build GRCh38
rs577615695 Chr.18: 59431263-59431263 on Build GRCh38
rs987502191 Chr.18: 59431361-59431361 on Build GRCh38
rs898981815 Chr.18: 59431265-59431265 on Build GRCh38
rs200966621 Chr.18: 59431486-59431486 on Build GRCh38
rs1050277276 Chr.18: 59431565-59431565 on Build GRCh38
rs986994563 Chr.18: 59431261-59431261 on Build GRCh38
rs139066911 Chr.18: 59431551-59431551 on Build GRCh38
rs201402743 Chr.18: 59431482-59431482 on Build GRCh38
rs956200001 Chr.18: 59431536-59431536 on Build GRCh38
rs139351765 Chr.18: 59431445-59431445 on Build GRCh38
rs182963415 Chr.18: 59431386-59431386 on Build GRCh38
rs890180714 Chr.18: 59431566-59431566 on Build GRCh38
rs1057434144 Chr.18: 59431506-59431506 on Build GRCh38
rs987878392 Chr.18: 59431541-59431541 on Build GRCh38
rs776172092 Chr.18: 59431533-59431533 on Build GRCh38
rs1056315583 Chr.18: 59431670-59431670 on Build GRCh38
rs997273874 Chr.18: 59431499-59431499 on Build GRCh38
rs749523231 Chr.18: 59431390-59431390 on Build GRCh38
rs750975475 Chr.18: 59431504-59431507 on Build GRCh38
rs886054040 Chr.18: 59431662-59431662 on Build GRCh38
rs774924698 Chr.18: 59431426-59431426 on Build GRCh38
rs536307825 Chr.18: 59431501-59431501 on Build GRCh38
rs1004817731 Chr.18: 59431231-59431231 on Build GRCh38
rs1017444878 Chr.18: 59431446-59431446 on Build GRCh38
rs980247091 Chr.18: 59431474-59431474 on Build GRCh38
rs956356791 Chr.18: 59431669-59431669 on Build GRCh38
rs975659622 Chr.18: 59431235-59431235 on Build GRCh38
rs556813935 Chr.18: 59431364-59431364 on Build GRCh38
rs980875689 Chr.18: 59431580-59431580 on Build GRCh38
rs201316273 Chr.18: 59431304-59431304 on Build GRCh38
rs1031197975 Chr.18: 59431620-59431620 on Build GRCh38
rs7230906 Chr.18: 59431404-59431404 on Build GRCh38
rs955640091 Chr.18: 59431513-59431513 on Build GRCh38
rs6567083 Chr.18: 59431510-59431510 on Build GRCh38
rs575123087 Chr.18: 59431395-59431395 on Build GRCh38
rs539838910 Chr.18: 59431389-59431389 on Build GRCh38
rs939310770 Chr.18: 59431658-59431658 on Build GRCh38
rs911634360 Chr.18: 59431300-59431300 on Build GRCh38
rs772516091 Chr.18: 59431521-59431521 on Build GRCh38
rs887190537 Chr.18: 59431431-59431431 on Build GRCh38
rs866996406 Chr.18: 59431626-59431626 on Build GRCh38
rs557695360 Chr.18: 59431258-59431258 on Build GRCh38
rs946386569 Chr.18: 59431380-59431380 on Build GRCh38
rs146222689 Chr.18: 59431410-59431410 on Build GRCh38
rs1052185867 Chr.18: 59431325-59431325 on Build GRCh38
rs935105000 Chr.18: 59431314-59431314 on Build GRCh38
rs200560154 Chr.18: 59431481-59431481 on Build GRCh38
rs543665085 Chr.18: 59431672-59431672 on Build GRCh38
rs914992048 Chr.18: 59431559-59431559 on Build GRCh38
rs905133459 Chr.18: 59431397-59431397 on Build GRCh38
rs946385733 Chr.18: 59431568-59431568 on Build GRCh38
rs544894423 Chr.18: 59431339-59431339 on Build GRCh38
rs557591805 Chr.18: 59431293-59431295 on Build GRCh38
rs553910582 Chr.18: 59431353-59431355 on Build GRCh38
rs940694626 Chr.18: 59431512-59431512 on Build GRCh38
rs66482821 Chr.18: 59431482-59431482 on Build GRCh38
rs962715385 Chr.18: 59431486-59431486 on Build GRCh38

View Full Product Details