Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40509650-40510102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXO5
Entrez Gene ID
64789
Gene Name
exonuclease 5
Gene Aliases
C1orf176, DEM1, Exo V, hExo5
UniGene
Hs.59584
Gene Chromosome Location
Chr.1: 40508719-40516556 on Build GRCh38
Transcripts
NM_022774.1 XM_005271125.3 XM_005271126.3 XM_005271128.3 XM_011541970.2 XM_017002099.1 XM_017002100.1 XM_017002101.1 XM_017002102.1 XM_017002103.1 XM_017002104.1

SNP Detail

SNP ID Chromosome Location
rs999916251 Chr.1: 40509725-40509725 on Build GRCh38
rs983520192 Chr.1: 40509883-40509883 on Build GRCh38
rs923464776 Chr.1: 40509688-40509688 on Build GRCh38
rs974265505 Chr.1: 40509847-40509847 on Build GRCh38
rs773967078 Chr.1: 40509759-40509759 on Build GRCh38
rs903798463 Chr.1: 40509724-40509724 on Build GRCh38
rs951683652 Chr.1: 40509865-40509866 on Build GRCh38
rs1031620711 Chr.1: 40509878-40509878 on Build GRCh38
rs942050358 Chr.1: 40509767-40509767 on Build GRCh38
rs1043590538 Chr.1: 40510070-40510070 on Build GRCh38
rs1044049062 Chr.1: 40509721-40509721 on Build GRCh38
rs999090091 Chr.1: 40509852-40509852 on Build GRCh38
rs1040580230 Chr.1: 40509678-40509678 on Build GRCh38
rs867270698 Chr.1: 40509997-40509997 on Build GRCh38
rs767129379 Chr.1: 40509908-40509908 on Build GRCh38
rs1031042872 Chr.1: 40509731-40509731 on Build GRCh38
rs929494454 Chr.1: 40509990-40509990 on Build GRCh38
rs1013594252 Chr.1: 40509797-40509797 on Build GRCh38
rs1023721008 Chr.1: 40509813-40509813 on Build GRCh38
rs921735906 Chr.1: 40510009-40510009 on Build GRCh38
rs960772269 Chr.1: 40509737-40509745 on Build GRCh38
rs765259946 Chr.1: 40509753-40509753 on Build GRCh38
rs773227832 Chr.1: 40509763-40509763 on Build GRCh38
rs1025000819 Chr.1: 40509810-40509810 on Build GRCh38
rs568389584 Chr.1: 40509886-40509886 on Build GRCh38
rs975856268 Chr.1: 40509950-40509950 on Build GRCh38
rs931791131 Chr.1: 40510048-40510048 on Build GRCh38
rs923013337 Chr.1: 40509965-40509965 on Build GRCh38
rs75760781 Chr.1: 40509784-40509784 on Build GRCh38
rs964363301 Chr.1: 40509935-40509935 on Build GRCh38
rs575787339 Chr.1: 40510048-40510048 on Build GRCh38
rs766049228 Chr.1: 40509909-40509909 on Build GRCh38
rs142703177 Chr.1: 40509843-40509843 on Build GRCh38
rs983992709 Chr.1: 40510020-40510020 on Build GRCh38
rs920157537 Chr.1: 40509859-40509859 on Build GRCh38
rs182834269 Chr.1: 40510076-40510076 on Build GRCh38
rs377408313 Chr.1: 40510059-40510059 on Build GRCh38
rs909469392 Chr.1: 40510030-40510030 on Build GRCh38
rs1055145566 Chr.1: 40510066-40510066 on Build GRCh38
rs943077694 Chr.1: 40509677-40509677 on Build GRCh38
rs888020065 Chr.1: 40509748-40509748 on Build GRCh38
rs942251127 Chr.1: 40510045-40510045 on Build GRCh38
rs760533903 Chr.1: 40509732-40509753 on Build GRCh38
rs147179262 Chr.1: 40509691-40509691 on Build GRCh38
rs1048015427 Chr.1: 40509746-40509746 on Build GRCh38
rs895252189 Chr.1: 40509795-40509795 on Build GRCh38

View Full Product Details