Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40983067-40983568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTPS1
Entrez Gene ID
1503
Gene Name
CTP synthase 1
Gene Aliases
CTPS, IMD24
UniGene
Hs.473087
Gene Chromosome Location
Chr.1: 40979299-41012565 on Build GRCh38
Transcripts
NM_001905.3 NR_125440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3419221 c.44G>A p.G15E Chr.1: 40983334-40983334 on Build GRCh38
4426419 c.50G>T p.G17V Chr.1: 40983340-40983340 on Build GRCh38
7121208 c.152C>A p.S51Y Chr.1: 40983442-40983442 on Build GRCh38
4426418 c.50G>T p.G17V Chr.1: 40983340-40983340 on Build GRCh38
3419222 c.44G>A p.G15E Chr.1: 40983334-40983334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768027664 Chr.1: 40983228-40983228 on Build GRCh38
rs1027585050 Chr.1: 40983141-40983141 on Build GRCh38
rs1034780349 Chr.1: 40983185-40983185 on Build GRCh38
rs1020690759 Chr.1: 40983355-40983355 on Build GRCh38
rs779248936 Chr.1: 40983446-40983446 on Build GRCh38
rs147898373 Chr.1: 40983219-40983219 on Build GRCh38
rs746497507 Chr.1: 40983244-40983244 on Build GRCh38
rs778922810 Chr.1: 40983476-40983478 on Build GRCh38
rs759914190 Chr.1: 40983303-40983303 on Build GRCh38
rs778065730 Chr.1: 40983234-40983234 on Build GRCh38
rs571425361 Chr.1: 40983119-40983119 on Build GRCh38
rs1021070908 Chr.1: 40983221-40983221 on Build GRCh38
rs750664691 Chr.1: 40983469-40983469 on Build GRCh38
rs541236235 Chr.1: 40983527-40983527 on Build GRCh38
rs367595948 Chr.1: 40983280-40983280 on Build GRCh38
rs780023479 Chr.1: 40983256-40983256 on Build GRCh38
rs753922614 Chr.1: 40983422-40983422 on Build GRCh38
rs757447163 Chr.1: 40983427-40983427 on Build GRCh38
rs982164330 Chr.1: 40983471-40983471 on Build GRCh38
rs772880925 Chr.1: 40983297-40983297 on Build GRCh38
rs940123927 Chr.1: 40983523-40983523 on Build GRCh38
rs144860512 Chr.1: 40983181-40983181 on Build GRCh38
rs11209315 Chr.1: 40983133-40983133 on Build GRCh38
rs370899922 Chr.1: 40983245-40983245 on Build GRCh38
rs764713994 Chr.1: 40983383-40983383 on Build GRCh38
rs756206074 Chr.1: 40983232-40983232 on Build GRCh38
rs767820852 Chr.1: 40983308-40983308 on Build GRCh38
rs753184274 Chr.1: 40983229-40983229 on Build GRCh38
rs959515210 Chr.1: 40983480-40983480 on Build GRCh38
rs868098496 Chr.1: 40983328-40983328 on Build GRCh38
rs757354207 Chr.1: 40983281-40983282 on Build GRCh38
rs941416147 Chr.1: 40983140-40983140 on Build GRCh38
rs898477692 Chr.1: 40983121-40983121 on Build GRCh38
rs199949603 Chr.1: 40983505-40983505 on Build GRCh38
rs967755684 Chr.1: 40983241-40983241 on Build GRCh38
rs1051113789 Chr.1: 40983195-40983195 on Build GRCh38
rs761020732 Chr.1: 40983380-40983380 on Build GRCh38
rs776453435 Chr.1: 40983521-40983521 on Build GRCh38
rs771807299 Chr.1: 40983251-40983251 on Build GRCh38
rs746973356 Chr.1: 40983257-40983257 on Build GRCh38
rs757498177 Chr.1: 40983242-40983242 on Build GRCh38
rs368206758 Chr.1: 40983485-40983485 on Build GRCh38
rs987001462 Chr.1: 40983494-40983494 on Build GRCh38
rs960475231 Chr.1: 40983218-40983218 on Build GRCh38
rs1037155483 Chr.1: 40983161-40983161 on Build GRCh38
rs533709230 Chr.1: 40983131-40983131 on Build GRCh38
rs781126937 Chr.1: 40983493-40983493 on Build GRCh38
rs769386291 Chr.1: 40983296-40983296 on Build GRCh38
rs753974219 Chr.1: 40983235-40983235 on Build GRCh38
rs941196893 Chr.1: 40983540-40983541 on Build GRCh38
rs545128484 Chr.1: 40983348-40983348 on Build GRCh38
rs750051352 Chr.1: 40983190-40983190 on Build GRCh38
rs576103457 Chr.1: 40983266-40983266 on Build GRCh38
rs756725880 Chr.1: 40983364-40983364 on Build GRCh38
rs556234590 Chr.1: 40983261-40983261 on Build GRCh38
rs374644871 Chr.1: 40983483-40983483 on Build GRCh38
rs745637693 Chr.1: 40983249-40983249 on Build GRCh38
rs905094757 Chr.1: 40983171-40983171 on Build GRCh38
rs779769756 Chr.1: 40983238-40983238 on Build GRCh38
rs141619431 Chr.1: 40983478-40983478 on Build GRCh38
rs1007151352 Chr.1: 40983226-40983226 on Build GRCh38

View Full Product Details