Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3885574-3886111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATCAY
Entrez Gene ID
85300
Gene Name
ATCAY, caytaxin
Gene Aliases
BNIP-H, CLAC
UniGene
Hs.418055
Gene Chromosome Location
Chr.19: 3880620-3928082 on Build GRCh38
Transcripts
NM_033064.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3388946 c.42C>T p.D14D Chr.19: 3885809-3885809 on Build GRCh38
5389639 c.74C>T p.P25L Chr.19: 3885841-3885841 on Build GRCh38
1183868 c.37G>A p.V13M Chr.19: 3885804-3885804 on Build GRCh38
1183869 c.37G>A p.V13M Chr.19: 3885804-3885804 on Build GRCh38
3202147 c.51G>C p.E17D Chr.19: 3885818-3885818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745720974 Chr.19: 3885804-3885804 on Build GRCh38
rs868017605 Chr.19: 3885780-3885780 on Build GRCh38
rs765457963 Chr.19: 3885877-3885877 on Build GRCh38
rs929859916 Chr.19: 3885984-3885984 on Build GRCh38
rs942528475 Chr.19: 3885741-3885741 on Build GRCh38
rs978144439 Chr.19: 3885643-3885643 on Build GRCh38
rs1030029547 Chr.19: 3885603-3885603 on Build GRCh38
rs529428286 Chr.19: 3885714-3885714 on Build GRCh38
rs955047270 Chr.19: 3885922-3885922 on Build GRCh38
rs141957025 Chr.19: 3885954-3885960 on Build GRCh38
rs978384357 Chr.19: 3885918-3885918 on Build GRCh38
rs564388390 Chr.19: 3885650-3885650 on Build GRCh38
rs978840838 Chr.19: 3886016-3886016 on Build GRCh38
rs541763903 Chr.19: 3886019-3886019 on Build GRCh38
rs1056520552 Chr.19: 3886047-3886047 on Build GRCh38
rs942475916 Chr.19: 3885958-3885958 on Build GRCh38
rs923642634 Chr.19: 3885669-3885669 on Build GRCh38
rs1057024264 Chr.19: 3885787-3885787 on Build GRCh38
rs996304067 Chr.19: 3885600-3885600 on Build GRCh38
rs991038545 Chr.19: 3885610-3885610 on Build GRCh38
rs937126056 Chr.19: 3885778-3885778 on Build GRCh38
rs775624223 Chr.19: 3885810-3885810 on Build GRCh38
rs772127259 Chr.19: 3885809-3885809 on Build GRCh38
rs73919386 Chr.19: 3885708-3885708 on Build GRCh38
rs146935412 Chr.19: 3885713-3885713 on Build GRCh38
rs939328152 Chr.19: 3886039-3886039 on Build GRCh38
rs768315218 Chr.19: 3885861-3885861 on Build GRCh38
rs1035163858 Chr.19: 3885603-3885614 on Build GRCh38
rs1020469076 Chr.19: 3885983-3885983 on Build GRCh38
rs916365288 Chr.19: 3885641-3885641 on Build GRCh38
rs74912608 Chr.19: 3885920-3885920 on Build GRCh38
rs968674495 Chr.19: 3885895-3885895 on Build GRCh38
rs375170455 Chr.19: 3885885-3885885 on Build GRCh38
rs777072643 Chr.19: 3885670-3885670 on Build GRCh38
rs556167354 Chr.19: 3885976-3885976 on Build GRCh38
rs959531075 Chr.19: 3885608-3885608 on Build GRCh38
rs990469123 Chr.19: 3885633-3885633 on Build GRCh38
rs968372230 Chr.19: 3886003-3886003 on Build GRCh38
rs957727712 Chr.19: 3885621-3885621 on Build GRCh38
rs56964691 Chr.19: 3885883-3885883 on Build GRCh38
rs968800488 Chr.19: 3885703-3885703 on Build GRCh38
rs533050723 Chr.19: 3885673-3885673 on Build GRCh38
rs1030481104 Chr.19: 3885942-3885942 on Build GRCh38
rs748431773 Chr.19: 3885788-3885788 on Build GRCh38
rs878925764 Chr.19: 3885631-3885631 on Build GRCh38
rs760237730 Chr.19: 3885857-3885857 on Build GRCh38
rs1048180602 Chr.19: 3885880-3885880 on Build GRCh38
rs374126058 Chr.19: 3885946-3885946 on Build GRCh38
rs534896329 Chr.19: 3885873-3885873 on Build GRCh38
rs915473307 Chr.19: 3885616-3885616 on Build GRCh38
rs912925598 Chr.19: 3885687-3885687 on Build GRCh38
rs374309679 Chr.19: 3885879-3885879 on Build GRCh38
rs368213837 Chr.19: 3885803-3885803 on Build GRCh38
rs575947379 Chr.19: 3886004-3886004 on Build GRCh38
rs992645354 Chr.19: 3886068-3886068 on Build GRCh38
rs761543482 Chr.19: 3885876-3885876 on Build GRCh38
rs1032251683 Chr.19: 3885615-3885615 on Build GRCh38
rs958652026 Chr.19: 3886046-3886046 on Build GRCh38
rs1037918542 Chr.19: 3885894-3885894 on Build GRCh38
rs112981952 Chr.19: 3885971-3885971 on Build GRCh38
rs770582696 Chr.19: 3885793-3885793 on Build GRCh38
rs927203522 Chr.19: 3885768-3885768 on Build GRCh38
rs541188234 Chr.19: 3885644-3885644 on Build GRCh38
rs996297438 Chr.19: 3885923-3885923 on Build GRCh38
rs565788691 Chr.19: 3885830-3885830 on Build GRCh38
rs986462846 Chr.19: 3885945-3885945 on Build GRCh38
rs1022486334 Chr.19: 3885886-3885886 on Build GRCh38

View Full Product Details