Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4795249-4795699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEM1A
Entrez Gene ID
55527
Gene Name
fem-1 homolog A
Gene Aliases
EPRAP
UniGene
Hs.515082
Gene Chromosome Location
Chr.19: 4791716-4795559 on Build GRCh38
Transcripts
NM_018708.2

SNP Detail

SNP ID Chromosome Location
rs774309064 Chr.19: 4795501-4795501 on Build GRCh38
rs567896158 Chr.19: 4795583-4795583 on Build GRCh38
rs866225481 Chr.19: 4795620-4795620 on Build GRCh38
rs201036592 Chr.19: 4795628-4795629 on Build GRCh38
rs373517352 Chr.19: 4795511-4795511 on Build GRCh38
rs558328197 Chr.19: 4795347-4795347 on Build GRCh38
rs867302346 Chr.19: 4795487-4795487 on Build GRCh38
rs551144881 Chr.19: 4795575-4795575 on Build GRCh38
rs1041396621 Chr.19: 4795444-4795444 on Build GRCh38
rs755765076 Chr.19: 4795507-4795507 on Build GRCh38
rs927108055 Chr.19: 4795326-4795326 on Build GRCh38
rs111468577 Chr.19: 4795378-4795379 on Build GRCh38
rs547287788 Chr.19: 4795603-4795603 on Build GRCh38
rs111790555 Chr.19: 4795584-4795584 on Build GRCh38
rs945856082 Chr.19: 4795441-4795441 on Build GRCh38
rs922961094 Chr.19: 4795486-4795486 on Build GRCh38
rs1051358956 Chr.19: 4795508-4795508 on Build GRCh38
rs73919324 Chr.19: 4795394-4795394 on Build GRCh38
rs752284109 Chr.19: 4795352-4795352 on Build GRCh38
rs143345083 Chr.19: 4795379-4795380 on Build GRCh38
rs989884619 Chr.19: 4795376-4795376 on Build GRCh38
rs113117276 Chr.19: 4795560-4795560 on Build GRCh38
rs8111933 Chr.19: 4795635-4795635 on Build GRCh38
rs34012693 Chr.19: 4795393-4795394 on Build GRCh38
rs1044386 Chr.19: 4795277-4795277 on Build GRCh38
rs564949216 Chr.19: 4795500-4795500 on Build GRCh38
rs1007472598 Chr.19: 4795524-4795524 on Build GRCh38
rs1009224825 Chr.19: 4795617-4795617 on Build GRCh38
rs1042090972 Chr.19: 4795625-4795625 on Build GRCh38
rs1044409 Chr.19: 4795330-4795330 on Build GRCh38
rs867653300 Chr.19: 4795505-4795505 on Build GRCh38
rs1025580567 Chr.19: 4795645-4795645 on Build GRCh38
rs567189833 Chr.19: 4795630-4795630 on Build GRCh38
rs3181004 Chr.19: 4795555-4795555 on Build GRCh38
rs11437 Chr.19: 4795333-4795333 on Build GRCh38
rs1025631446 Chr.19: 4795290-4795290 on Build GRCh38
rs1033898580 Chr.19: 4795666-4795666 on Build GRCh38
rs559244790 Chr.19: 4795271-4795271 on Build GRCh38
rs971273912 Chr.19: 4795298-4795298 on Build GRCh38
rs1000625232 Chr.19: 4795663-4795663 on Build GRCh38

View Full Product Details