Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18059429-18059944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12RB1
Entrez Gene ID
3594
Gene Name
interleukin 12 receptor subunit beta 1
Gene Aliases
CD212, IL-12R-BETA1, IL12RB, IMD30
UniGene
Hs.567294
Gene Chromosome Location
Chr.19: 18058994-18099027 on Build GRCh38
Transcripts
NM_001290023.1 NM_001290024.1 NM_005535.2 XM_006722741.3 XM_011527966.2 XM_011527967.2 XM_011527968.2 XM_011527969.2 XM_011527970.2 XM_011527971.2 XM_011527972.2 XM_011527973.2 XM_011527974.2 XM_011527975.2 XM_017026762.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3378659 c.1939G>A p.D647N Chr.19: 18059938-18059938 on Build GRCh38
3378658 c.1939G>A p.D647N Chr.19: 18059938-18059938 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1047190287 Chr.19: 18059735-18059735 on Build GRCh38
rs923778063 Chr.19: 18059643-18059643 on Build GRCh38
rs867674891 Chr.19: 18059665-18059665 on Build GRCh38
rs566576601 Chr.19: 18059905-18059905 on Build GRCh38
rs1017464476 Chr.19: 18059795-18059795 on Build GRCh38
rs761603497 Chr.19: 18059884-18059884 on Build GRCh38
rs752117474 Chr.19: 18059571-18059571 on Build GRCh38
rs3746190 Chr.19: 18059574-18059574 on Build GRCh38
rs777285303 Chr.19: 18059573-18059573 on Build GRCh38
rs560231272 Chr.19: 18059707-18059707 on Build GRCh38
rs12461312 Chr.19: 18059847-18059847 on Build GRCh38
rs775757288 Chr.19: 18059588-18059588 on Build GRCh38
rs17882555 Chr.19: 18059870-18059870 on Build GRCh38
rs372536968 Chr.19: 18059649-18059649 on Build GRCh38
rs760535885 Chr.19: 18059602-18059602 on Build GRCh38
rs760492840 Chr.19: 18059594-18059594 on Build GRCh38
rs998329896 Chr.19: 18059461-18059461 on Build GRCh38
rs933645266 Chr.19: 18059614-18059614 on Build GRCh38
rs1039912795 Chr.19: 18059686-18059686 on Build GRCh38
rs961525435 Chr.19: 18059750-18059750 on Build GRCh38
rs750881845 Chr.19: 18059913-18059913 on Build GRCh38
rs760387303 Chr.19: 18059555-18059555 on Build GRCh38
rs865848042 Chr.19: 18059921-18059921 on Build GRCh38
rs762302036 Chr.19: 18059646-18059646 on Build GRCh38
rs770668860 Chr.19: 18059645-18059645 on Build GRCh38
rs751046730 Chr.19: 18059879-18059879 on Build GRCh38
rs3833286 Chr.19: 18059455-18059456 on Build GRCh38
rs776582848 Chr.19: 18059611-18059611 on Build GRCh38
rs767132891 Chr.19: 18059515-18059515 on Build GRCh38
rs368249706 Chr.19: 18059647-18059647 on Build GRCh38
rs369239734 Chr.19: 18059712-18059712 on Build GRCh38
rs757085205 Chr.19: 18059504-18059504 on Build GRCh38
rs553163198 Chr.19: 18059875-18059875 on Build GRCh38
rs373710358 Chr.19: 18059591-18059591 on Build GRCh38
rs777955828 Chr.19: 18059843-18059843 on Build GRCh38
rs887314254 Chr.19: 18059759-18059759 on Build GRCh38
rs970804774 Chr.19: 18059524-18059524 on Build GRCh38
rs746333941 Chr.19: 18059580-18059580 on Build GRCh38
rs773114345 Chr.19: 18059554-18059554 on Build GRCh38
rs377287376 Chr.19: 18059626-18059626 on Build GRCh38
rs770391620 Chr.19: 18059584-18059584 on Build GRCh38
rs776991418 Chr.19: 18059854-18059854 on Build GRCh38
rs180780717 Chr.19: 18059749-18059749 on Build GRCh38
rs1032955548 Chr.19: 18059485-18059485 on Build GRCh38
rs779222722 Chr.19: 18059858-18059858 on Build GRCh38
rs759510863 Chr.19: 18059622-18059622 on Build GRCh38
rs748085048 Chr.19: 18059821-18059821 on Build GRCh38
rs754507050 Chr.19: 18059565-18059565 on Build GRCh38
rs934255662 Chr.19: 18059720-18059720 on Build GRCh38
rs955091939 Chr.19: 18059833-18059833 on Build GRCh38
rs536977912 Chr.19: 18059753-18059753 on Build GRCh38
rs1034171623 Chr.19: 18059552-18059552 on Build GRCh38
rs748543103 Chr.19: 18059834-18059834 on Build GRCh38
rs372358188 Chr.19: 18059608-18059608 on Build GRCh38
rs544880898 Chr.19: 18059575-18059575 on Build GRCh38
rs749311505 Chr.19: 18059590-18059590 on Build GRCh38
rs373982278 Chr.19: 18059572-18059572 on Build GRCh38
rs549618330 Chr.19: 18059566-18059566 on Build GRCh38
rs369210859 Chr.19: 18059656-18059656 on Build GRCh38
rs970598928 Chr.19: 18059842-18059842 on Build GRCh38
rs1005627741 Chr.19: 18059781-18059781 on Build GRCh38
rs376834832 Chr.19: 18059652-18059652 on Build GRCh38
rs982256803 Chr.19: 18059542-18059542 on Build GRCh38
rs970651176 Chr.19: 18059625-18059625 on Build GRCh38
rs868145882 Chr.19: 18059914-18059914 on Build GRCh38
rs376525105 Chr.19: 18059869-18059869 on Build GRCh38
rs368721970 Chr.19: 18059606-18059606 on Build GRCh38
rs2085457 Chr.19: 18059492-18059492 on Build GRCh38
rs775231157 Chr.19: 18059866-18059866 on Build GRCh38
rs753720796 Chr.19: 18059559-18059559 on Build GRCh38
rs935868069 Chr.19: 18059628-18059628 on Build GRCh38
rs188415514 Chr.19: 18059578-18059578 on Build GRCh38
rs894458142 Chr.19: 18059799-18059799 on Build GRCh38
rs549009800 Chr.19: 18059737-18059737 on Build GRCh38
rs138079162 Chr.19: 18059651-18059651 on Build GRCh38
rs901334647 Chr.19: 18059460-18059460 on Build GRCh38
rs942397342 Chr.19: 18059669-18059669 on Build GRCh38
rs866267146 Chr.19: 18059907-18059907 on Build GRCh38
rs759314886 Chr.19: 18059864-18059864 on Build GRCh38
rs374991461 Chr.19: 18059711-18059711 on Build GRCh38
rs750790051 Chr.19: 18059844-18059844 on Build GRCh38
rs2085456 Chr.19: 18059490-18059490 on Build GRCh38
rs370240272 Chr.19: 18059848-18059848 on Build GRCh38
rs1024323957 Chr.19: 18059840-18059840 on Build GRCh38
rs372962183 Chr.19: 18059694-18059694 on Build GRCh38
rs745932745 Chr.19: 18059882-18059882 on Build GRCh38
rs561622044 Chr.19: 18059553-18059553 on Build GRCh38
rs202106699 Chr.19: 18059917-18059917 on Build GRCh38
rs146989235 Chr.19: 18059482-18059482 on Build GRCh38
rs146334638 Chr.19: 18059864-18059864 on Build GRCh38
rs769726124 Chr.19: 18059865-18059865 on Build GRCh38
rs972995442 Chr.19: 18059785-18059785 on Build GRCh38
rs1003939184 Chr.19: 18059677-18059677 on Build GRCh38
rs1009584695 Chr.19: 18059507-18059507 on Build GRCh38
rs748330114 Chr.19: 18059862-18059862 on Build GRCh38
rs397756372 Chr.19: 18059460-18059461 on Build GRCh38
rs776461067 Chr.19: 18059863-18059863 on Build GRCh38
rs756740790 Chr.19: 18059845-18059845 on Build GRCh38
rs1039965132 Chr.19: 18059453-18059453 on Build GRCh38
rs758963566 Chr.19: 18059838-18059838 on Build GRCh38
rs1054291798 Chr.19: 18059859-18059860 on Build GRCh38

View Full Product Details