Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44782306-44782836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLC
Entrez Gene ID
23624
Gene Name
Cbl proto-oncogene C
Gene Aliases
CBL-3, CBL-SL, RNF57
UniGene
Hs.466907
Gene Chromosome Location
Chr.19: 44777855-44800646 on Build GRCh38
Transcripts
NM_001130852.1 NM_012116.3 XM_005258696.3 XM_011526688.2 XM_011526689.2 XM_011526690.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
998053 c.739G>C p.V247L Chr.19: 44782451-44782451 on Build GRCh38
5056758 c.741C>T p.V247V Chr.19: 44782453-44782453 on Build GRCh38
6620913 c.687G>A p.Q229Q Chr.19: 44782399-44782399 on Build GRCh38
392436 c.691_692insT p.A232fs*14 Chr.19: 44782403-44782404 on Build GRCh38
3892839 c.779+1G>C p.? Chr.19: 44782492-44782492 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934279686 Chr.19: 44782602-44782602 on Build GRCh38
rs765187360 Chr.19: 44782533-44782533 on Build GRCh38
rs953975785 Chr.19: 44782383-44782383 on Build GRCh38
rs1011407666 Chr.19: 44782342-44782342 on Build GRCh38
rs761236254 Chr.19: 44782503-44782503 on Build GRCh38
rs779208700 Chr.19: 44782446-44782446 on Build GRCh38
rs374937127 Chr.19: 44782762-44782762 on Build GRCh38
rs113678940 Chr.19: 44782744-44782744 on Build GRCh38
rs762581924 Chr.19: 44782356-44782356 on Build GRCh38
rs192645219 Chr.19: 44782672-44782672 on Build GRCh38
rs772749744 Chr.19: 44782349-44782349 on Build GRCh38
rs767920193 Chr.19: 44782487-44782487 on Build GRCh38
rs918634494 Chr.19: 44782498-44782498 on Build GRCh38
rs891540445 Chr.19: 44782610-44782610 on Build GRCh38
rs563940549 Chr.19: 44782761-44782761 on Build GRCh38
rs969928905 Chr.19: 44782745-44782745 on Build GRCh38
rs184303214 Chr.19: 44782686-44782686 on Build GRCh38
rs1006822543 Chr.19: 44782662-44782662 on Build GRCh38
rs537104035 Chr.19: 44782545-44782545 on Build GRCh38
rs111617296 Chr.19: 44782557-44782557 on Build GRCh38
rs759899304 Chr.19: 44782374-44782374 on Build GRCh38
rs1046605764 Chr.19: 44782554-44782554 on Build GRCh38
rs757478241 Chr.19: 44782424-44782424 on Build GRCh38
rs201543071 Chr.19: 44782338-44782338 on Build GRCh38
rs188745894 Chr.19: 44782737-44782737 on Build GRCh38
rs200548357 Chr.19: 44782339-44782339 on Build GRCh38
rs746806578 Chr.19: 44782332-44782332 on Build GRCh38
rs199634180 Chr.19: 44782548-44782548 on Build GRCh38
rs1016261156 Chr.19: 44782743-44782743 on Build GRCh38
rs771955392 Chr.19: 44782636-44782636 on Build GRCh38
rs757358367 Chr.19: 44782530-44782530 on Build GRCh38
rs10416628 Chr.19: 44782477-44782477 on Build GRCh38
rs779220373 Chr.19: 44782529-44782529 on Build GRCh38
rs140245762 Chr.19: 44782632-44782632 on Build GRCh38
rs773303776 Chr.19: 44782720-44782720 on Build GRCh38
rs916844484 Chr.19: 44782534-44782534 on Build GRCh38
rs200352104 Chr.19: 44782564-44782564 on Build GRCh38
rs376748601 Chr.19: 44782524-44782524 on Build GRCh38
rs892686305 Chr.19: 44782633-44782633 on Build GRCh38
rs747961173 Chr.19: 44782558-44782558 on Build GRCh38
rs200116296 Chr.19: 44782363-44782363 on Build GRCh38
rs148127553 Chr.19: 44782451-44782451 on Build GRCh38
rs769449007 Chr.19: 44782344-44782344 on Build GRCh38
rs753226393 Chr.19: 44782400-44782400 on Build GRCh38
rs770386913 Chr.19: 44782783-44782783 on Build GRCh38
rs746006448 Chr.19: 44782450-44782450 on Build GRCh38
rs776199814 Chr.19: 44782784-44782784 on Build GRCh38
rs767829298 Chr.19: 44782395-44782395 on Build GRCh38
rs889594928 Chr.19: 44782659-44782659 on Build GRCh38
rs750678344 Chr.19: 44782537-44782537 on Build GRCh38
rs147316311 Chr.19: 44782461-44782461 on Build GRCh38
rs768503313 Chr.19: 44782337-44782337 on Build GRCh38
rs759141229 Chr.19: 44782480-44782480 on Build GRCh38
rs373513195 Chr.19: 44782407-44782407 on Build GRCh38
rs138510701 Chr.19: 44782473-44782473 on Build GRCh38
rs753946761 Chr.19: 44782422-44782422 on Build GRCh38
rs764649009 Chr.19: 44782507-44782507 on Build GRCh38
rs748848345 Chr.19: 44782468-44782468 on Build GRCh38
rs952812482 Chr.19: 44782735-44782735 on Build GRCh38
rs539494394 Chr.19: 44782746-44782746 on Build GRCh38
rs781082374 Chr.19: 44782458-44782458 on Build GRCh38
rs1011854229 Chr.19: 44782622-44782622 on Build GRCh38
rs1052840655 Chr.19: 44782587-44782587 on Build GRCh38
rs775798790 Chr.19: 44782501-44782501 on Build GRCh38
rs141956736 Chr.19: 44782460-44782460 on Build GRCh38
rs574125970 Chr.19: 44782639-44782639 on Build GRCh38
rs908324880 Chr.19: 44782607-44782607 on Build GRCh38
rs755455903 Chr.19: 44782347-44782348 on Build GRCh38
rs938385982 Chr.19: 44782803-44782803 on Build GRCh38
rs987399210 Chr.19: 44782812-44782812 on Build GRCh38
rs994166391 Chr.19: 44782695-44782695 on Build GRCh38
rs764196556 Chr.19: 44782418-44782418 on Build GRCh38

View Full Product Details