Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48832559-48833070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B14
Entrez Gene ID
51171
Gene Name
hydroxysteroid 17-beta dehydrogenase 14
Gene Aliases
DHRS10, SDR47C1, retSDR3
UniGene
Hs.18788
Gene Chromosome Location
Chr.19: 48813014-48836688 on Build GRCh38
Transcripts
NM_016246.2 XM_005258969.3 XM_005258970.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7229674 c.233G>A p.R78H Chr.19: 48832710-48832710 on Build GRCh38
1395120 c.232C>T p.R78C Chr.19: 48832711-48832711 on Build GRCh38
4581274 c.244C>T p.R82C Chr.19: 48832699-48832699 on Build GRCh38
3960262 c.233G>C p.R78P Chr.19: 48832710-48832710 on Build GRCh38
4691265 c.246C>T p.R82R Chr.19: 48832697-48832697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770824713 Chr.19: 48832778-48832778 on Build GRCh38
rs937299266 Chr.19: 48833002-48833002 on Build GRCh38
rs372759566 Chr.19: 48832776-48832776 on Build GRCh38
rs895617546 Chr.19: 48833039-48833039 on Build GRCh38
rs551414990 Chr.19: 48832914-48832914 on Build GRCh38
rs933460526 Chr.19: 48832882-48832882 on Build GRCh38
rs549725283 Chr.19: 48832910-48832910 on Build GRCh38
rs768818583 Chr.19: 48832727-48832727 on Build GRCh38
rs139341223 Chr.19: 48832708-48832708 on Build GRCh38
rs775437835 Chr.19: 48832645-48832645 on Build GRCh38
rs542142249 Chr.19: 48832720-48832720 on Build GRCh38
rs184605823 Chr.19: 48832753-48832753 on Build GRCh38
rs768479422 Chr.19: 48832622-48832622 on Build GRCh38
rs983160864 Chr.19: 48832588-48832588 on Build GRCh38
rs1038362873 Chr.19: 48832945-48832945 on Build GRCh38
rs1054958840 Chr.19: 48833033-48833033 on Build GRCh38
rs777655600 Chr.19: 48832775-48832775 on Build GRCh38
rs756125735 Chr.19: 48832771-48832771 on Build GRCh38
rs374378406 Chr.19: 48832759-48832759 on Build GRCh38
rs372590687 Chr.19: 48832757-48832758 on Build GRCh38
rs559654067 Chr.19: 48832852-48832852 on Build GRCh38
rs895089280 Chr.19: 48833036-48833036 on Build GRCh38
rs767927839 Chr.19: 48832637-48832637 on Build GRCh38
rs780714536 Chr.19: 48832770-48832770 on Build GRCh38
rs765221767 Chr.19: 48832747-48832748 on Build GRCh38
rs572504759 Chr.19: 48832604-48832604 on Build GRCh38
rs763825996 Chr.19: 48832757-48832757 on Build GRCh38
rs563845406 Chr.19: 48832620-48832620 on Build GRCh38
rs200446358 Chr.19: 48832650-48832650 on Build GRCh38
rs982853020 Chr.19: 48832932-48832932 on Build GRCh38
rs751041317 Chr.19: 48832697-48832697 on Build GRCh38
rs569816782 Chr.19: 48833023-48833023 on Build GRCh38
rs758016296 Chr.19: 48832676-48832676 on Build GRCh38
rs767595849 Chr.19: 48832639-48832639 on Build GRCh38
rs779545107 Chr.19: 48832679-48832679 on Build GRCh38
rs780875813 Chr.19: 48832763-48832763 on Build GRCh38
rs751267728 Chr.19: 48832761-48832761 on Build GRCh38
rs759861638 Chr.19: 48832635-48832635 on Build GRCh38
rs758377129 Chr.19: 48833029-48833029 on Build GRCh38
rs999883919 Chr.19: 48832864-48832864 on Build GRCh38
rs189467300 Chr.19: 48832942-48832942 on Build GRCh38
rs568447417 Chr.19: 48832758-48832758 on Build GRCh38
rs762326427 Chr.19: 48832741-48832741 on Build GRCh38
rs144132021 Chr.19: 48832714-48832714 on Build GRCh38
rs745434256 Chr.19: 48832899-48832899 on Build GRCh38
rs920494964 Chr.19: 48832973-48832973 on Build GRCh38
rs368366441 Chr.19: 48832930-48832930 on Build GRCh38
rs1014508203 Chr.19: 48833040-48833040 on Build GRCh38
rs983493012 Chr.19: 48832788-48832789 on Build GRCh38
rs963923682 Chr.19: 48832846-48832846 on Build GRCh38
rs539969933 Chr.19: 48832623-48832623 on Build GRCh38
rs774695917 Chr.19: 48832630-48832630 on Build GRCh38
rs750788483 Chr.19: 48832754-48832754 on Build GRCh38
rs746020386 Chr.19: 48832710-48832710 on Build GRCh38
rs761160123 Chr.19: 48832816-48832816 on Build GRCh38
rs754586753 Chr.19: 48832698-48832698 on Build GRCh38
rs903743116 Chr.19: 48832715-48832715 on Build GRCh38
rs201026226 Chr.19: 48832707-48832707 on Build GRCh38
rs908622971 Chr.19: 48832937-48832937 on Build GRCh38
rs922832929 Chr.19: 48832912-48832912 on Build GRCh38
rs937378915 Chr.19: 48832946-48832946 on Build GRCh38
rs762132140 Chr.19: 48832661-48832661 on Build GRCh38
rs192095235 Chr.19: 48832652-48832652 on Build GRCh38
rs781248545 Chr.19: 48832699-48832699 on Build GRCh38
rs754428235 Chr.19: 48832762-48832762 on Build GRCh38
rs552717647 Chr.19: 48832863-48832863 on Build GRCh38
rs3760787 Chr.19: 48832898-48832898 on Build GRCh38
rs758952033 Chr.19: 48832630-48832635 on Build GRCh38
rs934112374 Chr.19: 48832688-48832688 on Build GRCh38
rs143254849 Chr.19: 48832666-48832666 on Build GRCh38
rs532759188 Chr.19: 48832849-48832849 on Build GRCh38
rs752563923 Chr.19: 48832753-48832754 on Build GRCh38
rs750026351 Chr.19: 48832673-48832673 on Build GRCh38
rs777098753 Chr.19: 48832731-48832731 on Build GRCh38
rs780106730 Chr.19: 48832779-48832779 on Build GRCh38
rs78970396 Chr.19: 48832954-48832954 on Build GRCh38
rs895553184 Chr.19: 48832848-48832848 on Build GRCh38
rs751759375 Chr.19: 48832758-48832759 on Build GRCh38
rs942310264 Chr.19: 48832610-48832610 on Build GRCh38
rs1015939234 Chr.19: 48832817-48832817 on Build GRCh38
rs373667121 Chr.19: 48832626-48832626 on Build GRCh38
rs747956807 Chr.19: 48832625-48832625 on Build GRCh38
rs765946620 Chr.19: 48832760-48832760 on Build GRCh38
rs966998322 Chr.19: 48832854-48832854 on Build GRCh38
rs752642413 Chr.19: 48832770-48832770 on Build GRCh38
rs987663144 Chr.19: 48832889-48832889 on Build GRCh38
rs756982120 Chr.19: 48832870-48832870 on Build GRCh38
rs772197700 Chr.19: 48832711-48832711 on Build GRCh38
rs568554789 Chr.19: 48832976-48832976 on Build GRCh38

View Full Product Details