Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49069245-49069751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNA7
Entrez Gene ID
3743
Gene Name
potassium voltage-gated channel subfamily A member 7
Gene Aliases
HAK6, KV1.7
UniGene
Hs.306973
Gene Chromosome Location
Chr.19: 49067418-49072941 on Build GRCh38
Transcripts
NM_031886.2

SNP Detail

SNP ID Chromosome Location
rs940935637 Chr.19: 49069409-49069409 on Build GRCh38
rs369281563 Chr.19: 49069374-49069374 on Build GRCh38
rs977008323 Chr.19: 49069622-49069622 on Build GRCh38
rs968637240 Chr.19: 49069303-49069303 on Build GRCh38
rs4802547 Chr.19: 49069290-49069290 on Build GRCh38
rs918408090 Chr.19: 49069711-49069711 on Build GRCh38
rs542117226 Chr.19: 49069498-49069498 on Build GRCh38
rs867110954 Chr.19: 49069400-49069400 on Build GRCh38
rs565989915 Chr.19: 49069698-49069698 on Build GRCh38
rs1022288325 Chr.19: 49069559-49069559 on Build GRCh38
rs985334205 Chr.19: 49069318-49069318 on Build GRCh38
rs115519627 Chr.19: 49069448-49069448 on Build GRCh38
rs908159584 Chr.19: 49069358-49069358 on Build GRCh38
rs116835540 Chr.19: 49069504-49069504 on Build GRCh38
rs1031513410 Chr.19: 49069274-49069274 on Build GRCh38
rs996322345 Chr.19: 49069438-49069438 on Build GRCh38
rs547019331 Chr.19: 49069604-49069604 on Build GRCh38
rs370138761 Chr.19: 49069423-49069423 on Build GRCh38
rs776779735 Chr.19: 49069474-49069474 on Build GRCh38
rs956786849 Chr.19: 49069284-49069284 on Build GRCh38
rs568706958 Chr.19: 49069289-49069289 on Build GRCh38
rs970501659 Chr.19: 49069611-49069611 on Build GRCh38
rs547731050 Chr.19: 49069317-49069317 on Build GRCh38
rs138664779 Chr.19: 49069413-49069413 on Build GRCh38
rs546202785 Chr.19: 49069546-49069546 on Build GRCh38
rs566475219 Chr.19: 49069410-49069410 on Build GRCh38
rs982210284 Chr.19: 49069614-49069614 on Build GRCh38
rs537692562 Chr.19: 49069443-49069443 on Build GRCh38
rs916295253 Chr.19: 49069299-49069299 on Build GRCh38
rs1032198296 Chr.19: 49069553-49069553 on Build GRCh38
rs574502199 Chr.19: 49069458-49069458 on Build GRCh38
rs940321246 Chr.19: 49069647-49069647 on Build GRCh38
rs907522509 Chr.19: 49069630-49069630 on Build GRCh38
rs990548730 Chr.19: 49069291-49069291 on Build GRCh38
rs189585441 Chr.19: 49069415-49069415 on Build GRCh38
rs531839018 Chr.19: 49069594-49069594 on Build GRCh38
rs999413664 Chr.19: 49069540-49069540 on Build GRCh38
rs572496187 Chr.19: 49069523-49069523 on Build GRCh38
rs973668333 Chr.19: 49069673-49069673 on Build GRCh38
rs3810185 Chr.19: 49069572-49069572 on Build GRCh38
rs538842150 Chr.19: 49069347-49069347 on Build GRCh38
rs867401026 Chr.19: 49069499-49069499 on Build GRCh38
rs888099108 Chr.19: 49069447-49069447 on Build GRCh38
rs567858467 Chr.19: 49069522-49069522 on Build GRCh38
rs911091583 Chr.19: 49069340-49069340 on Build GRCh38
rs763549119 Chr.19: 49069573-49069573 on Build GRCh38
rs943869861 Chr.19: 49069353-49069353 on Build GRCh38
rs1031146867 Chr.19: 49069508-49069508 on Build GRCh38
rs868832649 Chr.19: 49069533-49069533 on Build GRCh38
rs866253075 Chr.19: 49069595-49069595 on Build GRCh38
rs990581413 Chr.19: 49069589-49069589 on Build GRCh38

View Full Product Details