Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27326885-27327327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C2
Entrez Gene ID
2976
Gene Name
general transcription factor IIIC subunit 2
Gene Aliases
TFIIIC-BETA, TFIIIC110
UniGene
Hs.745168
Gene Chromosome Location
Chr.2: 27325849-27357034 on Build GRCh38
Transcripts
NM_001035521.2 NM_001318909.1 NM_001521.3 XM_005264272.4 XM_011532801.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1019639 c.2410-4_2419delCCAGGGTTCATTCC p.? Chr.2: 27327275-27327288 on Build GRCh38
6158124 c.2450G>T p.R817L Chr.2: 27327244-27327244 on Build GRCh38
1019638 c.2466G>T p.E822D Chr.2: 27327228-27327228 on Build GRCh38
5637947 c.2425C>T p.L809L Chr.2: 27327269-27327269 on Build GRCh38
3798906 c.2455C>A p.Q819K Chr.2: 27327239-27327239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770212777 Chr.2: 27327169-27327169 on Build GRCh38
rs766398661 Chr.2: 27327133-27327133 on Build GRCh38
rs761260088 Chr.2: 27327017-27327018 on Build GRCh38
rs756909209 Chr.2: 27327263-27327263 on Build GRCh38
rs756532324 Chr.2: 27327143-27327143 on Build GRCh38
rs113469548 Chr.2: 27326966-27326967 on Build GRCh38
rs773296313 Chr.2: 27327128-27327128 on Build GRCh38
rs771948035 Chr.2: 27326936-27326936 on Build GRCh38
rs767433437 Chr.2: 27326929-27326929 on Build GRCh38
rs374386568 Chr.2: 27327251-27327251 on Build GRCh38
rs75302569 Chr.2: 27327004-27327004 on Build GRCh38
rs946045245 Chr.2: 27327149-27327149 on Build GRCh38
rs772041729 Chr.2: 27327158-27327158 on Build GRCh38
rs377526579 Chr.2: 27327154-27327154 on Build GRCh38
rs369880736 Chr.2: 27327155-27327155 on Build GRCh38
rs1014524755 Chr.2: 27326973-27326973 on Build GRCh38
rs778140572 Chr.2: 27327144-27327144 on Build GRCh38
rs745360990 Chr.2: 27327157-27327157 on Build GRCh38
rs764398552 Chr.2: 27327255-27327255 on Build GRCh38
rs746976365 Chr.2: 27327280-27327280 on Build GRCh38
rs140411381 Chr.2: 27327066-27327068 on Build GRCh38
rs777015032 Chr.2: 27326915-27326916 on Build GRCh38
rs746866571 Chr.2: 27327185-27327185 on Build GRCh38
rs537451431 Chr.2: 27326969-27326969 on Build GRCh38
rs367564545 Chr.2: 27327287-27327287 on Build GRCh38
rs373604869 Chr.2: 27326922-27326922 on Build GRCh38
rs759683401 Chr.2: 27327241-27327241 on Build GRCh38
rs554231335 Chr.2: 27327068-27327069 on Build GRCh38
rs370776889 Chr.2: 27326914-27326914 on Build GRCh38
rs140952529 Chr.2: 27327238-27327238 on Build GRCh38
rs185853433 Chr.2: 27326968-27326968 on Build GRCh38
rs763103939 Chr.2: 27327222-27327222 on Build GRCh38
rs763165744 Chr.2: 27327132-27327132 on Build GRCh38
rs771956711 Chr.2: 27326945-27326945 on Build GRCh38
rs888315239 Chr.2: 27327045-27327045 on Build GRCh38
rs754875502 Chr.2: 27327167-27327168 on Build GRCh38
rs756902344 Chr.2: 27327145-27327146 on Build GRCh38
rs757892836 Chr.2: 27327271-27327271 on Build GRCh38
rs768352854 Chr.2: 27327198-27327198 on Build GRCh38
rs1041413538 Chr.2: 27327010-27327010 on Build GRCh38
rs35615796 Chr.2: 27326973-27326974 on Build GRCh38
rs913229262 Chr.2: 27327077-27327077 on Build GRCh38
rs181281564 Chr.2: 27326966-27326966 on Build GRCh38
rs1018821089 Chr.2: 27327031-27327031 on Build GRCh38
rs397868275 Chr.2: 27326978-27326978 on Build GRCh38
rs193107013 Chr.2: 27326939-27326939 on Build GRCh38
rs116010390 Chr.2: 27326931-27326931 on Build GRCh38
rs879247343 Chr.2: 27327091-27327091 on Build GRCh38
rs994563049 Chr.2: 27327002-27327002 on Build GRCh38
rs397871054 Chr.2: 27326978-27326979 on Build GRCh38
rs779967245 Chr.2: 27327164-27327164 on Build GRCh38
rs750705301 Chr.2: 27327257-27327262 on Build GRCh38
rs140259906 Chr.2: 27327052-27327052 on Build GRCh38
rs956380889 Chr.2: 27327107-27327107 on Build GRCh38
rs986382125 Chr.2: 27327030-27327030 on Build GRCh38
rs538518287 Chr.2: 27327012-27327012 on Build GRCh38
rs776444108 Chr.2: 27327201-27327201 on Build GRCh38
rs925442939 Chr.2: 27327245-27327245 on Build GRCh38
rs185237848 Chr.2: 27327073-27327073 on Build GRCh38
rs139405044 Chr.2: 27327273-27327273 on Build GRCh38
rs568205864 Chr.2: 27326914-27326915 on Build GRCh38
rs751605227 Chr.2: 27327134-27327134 on Build GRCh38
rs1055875732 Chr.2: 27327211-27327211 on Build GRCh38
rs768106441 Chr.2: 27327139-27327139 on Build GRCh38
rs574674464 Chr.2: 27327080-27327080 on Build GRCh38
rs753551344 Chr.2: 27327134-27327135 on Build GRCh38
rs760166169 Chr.2: 27327135-27327135 on Build GRCh38
rs761196731 Chr.2: 27327254-27327254 on Build GRCh38
rs542043449 Chr.2: 27327140-27327140 on Build GRCh38
rs764913410 Chr.2: 27327264-27327264 on Build GRCh38
rs897450884 Chr.2: 27327274-27327274 on Build GRCh38
rs1055946207 Chr.2: 27326959-27326959 on Build GRCh38
rs71441066 Chr.2: 27327011-27327011 on Build GRCh38
rs778420883 Chr.2: 27326930-27326930 on Build GRCh38
rs35568684 Chr.2: 27326967-27326967 on Build GRCh38
rs189095327 Chr.2: 27326977-27326977 on Build GRCh38
rs763681410 Chr.2: 27326914-27326917 on Build GRCh38
rs768196570 Chr.2: 27327244-27327244 on Build GRCh38
rs989507471 Chr.2: 27327121-27327121 on Build GRCh38
rs76943914 Chr.2: 27326967-27326967 on Build GRCh38
rs370941957 Chr.2: 27327148-27327148 on Build GRCh38
rs981815130 Chr.2: 27327074-27327074 on Build GRCh38
rs1031114362 Chr.2: 27327081-27327081 on Build GRCh38
rs754184411 Chr.2: 27327262-27327262 on Build GRCh38
rs1026898857 Chr.2: 27327036-27327036 on Build GRCh38
rs771157977 Chr.2: 27327232-27327232 on Build GRCh38

View Full Product Details