Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27443959-27444457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT172
Entrez Gene ID
26160
Gene Name
intraflagellar transport 172
Gene Aliases
BBS20, NPHP17, RP71, SLB, SRTD10, osm-1, wim
UniGene
Hs.127401
Gene Chromosome Location
Chr.2: 27444373-27489819 on Build GRCh38
Transcripts
NM_015662.2 XM_005264254.1 XM_006711986.3 XM_006711987.1 XM_011532757.2 XM_011532759.2 XM_011532760.2 XM_017003790.1 XM_017003791.1 XM_017003793.1 XM_017003794.1 XM_017003795.1 NM_001168364.1 NM_001321325.1 NM_173853.3

Gene Symbol
KRTCAP3
Entrez Gene ID
200634
Gene Name
keratinocyte associated protein 3
Gene Aliases
KCP3
UniGene
Hs.59509
Gene Chromosome Location
Chr.2: 27442366-27446481 on Build GRCh38
Transcripts
NM_015662.2 XM_005264254.1 XM_006711986.3 XM_006711987.1 XM_011532757.2 XM_011532759.2 XM_011532760.2 XM_017003790.1 XM_017003791.1 XM_017003793.1 XM_017003794.1 XM_017003795.1 NM_001168364.1 NM_001321325.1 NM_173853.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1019696 c.697C>T p.R233W Chr.2: 27444030-27444030 on Build GRCh38
4555926 c.672G>T p.Q224H Chr.2: 27444005-27444005 on Build GRCh38
3972203 c.716G>A p.W239* Chr.2: 27444049-27444049 on Build GRCh38
442747 c.5247G>T p.Q1749H Chr.2: 27444435-27444435 on Build GRCh38
3798917 c.627G>A p.M209I Chr.2: 27443960-27443960 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753906342 Chr.2: 27444102-27444102 on Build GRCh38
rs201844602 Chr.2: 27444014-27444014 on Build GRCh38
rs921327457 Chr.2: 27444249-27444249 on Build GRCh38
rs764167859 Chr.2: 27444252-27444252 on Build GRCh38
rs774639877 Chr.2: 27444045-27444045 on Build GRCh38
rs752693869 Chr.2: 27444178-27444178 on Build GRCh38
rs973701706 Chr.2: 27444006-27444006 on Build GRCh38
rs560284264 Chr.2: 27444278-27444278 on Build GRCh38
rs755372600 Chr.2: 27444002-27444002 on Build GRCh38
rs946602184 Chr.2: 27444150-27444150 on Build GRCh38
rs781247736 Chr.2: 27444033-27444034 on Build GRCh38
rs772224250 Chr.2: 27444420-27444420 on Build GRCh38
rs995333428 Chr.2: 27444358-27444360 on Build GRCh38
rs756880986 Chr.2: 27443992-27443992 on Build GRCh38
rs10208616 Chr.2: 27444241-27444241 on Build GRCh38
rs1054513596 Chr.2: 27444060-27444060 on Build GRCh38
rs778459750 Chr.2: 27444101-27444101 on Build GRCh38
rs939934759 Chr.2: 27444311-27444311 on Build GRCh38
rs764067258 Chr.2: 27444074-27444074 on Build GRCh38
rs533918778 Chr.2: 27444189-27444189 on Build GRCh38
rs911685067 Chr.2: 27444082-27444082 on Build GRCh38
rs745427537 Chr.2: 27444317-27444317 on Build GRCh38
rs746100848 Chr.2: 27444412-27444412 on Build GRCh38
rs544892369 Chr.2: 27444216-27444216 on Build GRCh38
rs766200814 Chr.2: 27444173-27444173 on Build GRCh38
rs4803 Chr.2: 27444430-27444430 on Build GRCh38
rs894890263 Chr.2: 27444418-27444418 on Build GRCh38
rs62131871 Chr.2: 27444317-27444317 on Build GRCh38
rs368365597 Chr.2: 27444000-27444000 on Build GRCh38
rs187699375 Chr.2: 27444031-27444031 on Build GRCh38
rs936156237 Chr.2: 27444217-27444218 on Build GRCh38
rs962262793 Chr.2: 27444336-27444336 on Build GRCh38
rs756644767 Chr.2: 27444100-27444100 on Build GRCh38
rs763147273 Chr.2: 27444203-27444203 on Build GRCh38
rs771889359 Chr.2: 27444058-27444058 on Build GRCh38
rs995486332 Chr.2: 27444356-27444356 on Build GRCh38
rs1033400323 Chr.2: 27444154-27444154 on Build GRCh38
rs1054911867 Chr.2: 27444380-27444380 on Build GRCh38
rs757428770 Chr.2: 27444382-27444382 on Build GRCh38
rs538979229 Chr.2: 27444251-27444251 on Build GRCh38
rs567002799 Chr.2: 27444097-27444097 on Build GRCh38
rs753039284 Chr.2: 27444098-27444098 on Build GRCh38
rs755653071 Chr.2: 27444390-27444391 on Build GRCh38
rs573966162 Chr.2: 27444211-27444211 on Build GRCh38
rs199933840 Chr.2: 27444087-27444087 on Build GRCh38
rs991812572 Chr.2: 27444168-27444168 on Build GRCh38
rs776552648 Chr.2: 27444435-27444435 on Build GRCh38
rs745593932 Chr.2: 27444047-27444047 on Build GRCh38
rs779876638 Chr.2: 27444428-27444428 on Build GRCh38
rs775216589 Chr.2: 27444070-27444070 on Build GRCh38
rs750101396 Chr.2: 27444090-27444090 on Build GRCh38
rs375251963 Chr.2: 27444383-27444383 on Build GRCh38
rs768540015 Chr.2: 27444434-27444434 on Build GRCh38
rs778920690 Chr.2: 27444391-27444391 on Build GRCh38
rs959050431 Chr.2: 27444161-27444161 on Build GRCh38
rs112481808 Chr.2: 27444206-27444206 on Build GRCh38
rs748672726 Chr.2: 27444013-27444013 on Build GRCh38
rs1015839474 Chr.2: 27444139-27444139 on Build GRCh38
rs776068769 Chr.2: 27444078-27444078 on Build GRCh38
rs891082706 Chr.2: 27444302-27444302 on Build GRCh38
rs527418791 Chr.2: 27444284-27444284 on Build GRCh38
rs972055313 Chr.2: 27444200-27444200 on Build GRCh38
rs781492553 Chr.2: 27444003-27444003 on Build GRCh38
rs748029831 Chr.2: 27444044-27444044 on Build GRCh38
rs545654643 Chr.2: 27444270-27444270 on Build GRCh38
rs200649406 Chr.2: 27444015-27444015 on Build GRCh38
rs758040970 Chr.2: 27444094-27444094 on Build GRCh38
rs898288095 Chr.2: 27444146-27444146 on Build GRCh38
rs749405253 Chr.2: 27444030-27444030 on Build GRCh38
rs932681648 Chr.2: 27444254-27444254 on Build GRCh38
rs764727063 Chr.2: 27444088-27444088 on Build GRCh38
rs760575802 Chr.2: 27444073-27444073 on Build GRCh38

View Full Product Details