Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 65241912-65242421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR2
Entrez Gene ID
10097
Gene Name
ARP2 actin related protein 2 homolog
Gene Aliases
ARP2
UniGene
Hs.643727
Gene Chromosome Location
Chr.2: 65227695-65271253 on Build GRCh38
Transcripts
NM_001005386.2 NM_005722.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1615034 c.174+2T>C p.? Chr.2: 65242065-65242065 on Build GRCh38
6257747 c.174+1G>T p.? Chr.2: 65242064-65242064 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs930025036 Chr.2: 65242263-65242263 on Build GRCh38
rs1011789711 Chr.2: 65242358-65242358 on Build GRCh38
rs111563286 Chr.2: 65242171-65242171 on Build GRCh38
rs1034084419 Chr.2: 65242375-65242375 on Build GRCh38
rs776164693 Chr.2: 65242081-65242081 on Build GRCh38
rs990722735 Chr.2: 65242224-65242224 on Build GRCh38
rs945683184 Chr.2: 65242246-65242246 on Build GRCh38
rs185150978 Chr.2: 65242376-65242376 on Build GRCh38
rs915468401 Chr.2: 65242245-65242245 on Build GRCh38
rs551860130 Chr.2: 65242154-65242154 on Build GRCh38
rs373657502 Chr.2: 65242256-65242256 on Build GRCh38
rs1004945978 Chr.2: 65242088-65242091 on Build GRCh38
rs560006273 Chr.2: 65242222-65242222 on Build GRCh38
rs765005829 Chr.2: 65242091-65242091 on Build GRCh38
rs572102502 Chr.2: 65242248-65242248 on Build GRCh38
rs1039113041 Chr.2: 65242269-65242269 on Build GRCh38
rs891111468 Chr.2: 65242330-65242330 on Build GRCh38
rs773989352 Chr.2: 65242027-65242027 on Build GRCh38
rs1048921250 Chr.2: 65242310-65242310 on Build GRCh38
rs556345476 Chr.2: 65242128-65242128 on Build GRCh38
rs756598675 Chr.2: 65242039-65242039 on Build GRCh38
rs564421492 Chr.2: 65242332-65242332 on Build GRCh38
rs758731353 Chr.2: 65242105-65242105 on Build GRCh38
rs142962959 Chr.2: 65242333-65242338 on Build GRCh38
rs532540976 Chr.2: 65242147-65242147 on Build GRCh38
rs748047170 Chr.2: 65242030-65242030 on Build GRCh38
rs541064940 Chr.2: 65242258-65242258 on Build GRCh38
rs559435475 Chr.2: 65242190-65242190 on Build GRCh38
rs539235105 Chr.2: 65242001-65242001 on Build GRCh38
rs893347692 Chr.2: 65242343-65242343 on Build GRCh38
rs759155219 Chr.2: 65242089-65242089 on Build GRCh38
rs752988253 Chr.2: 65242100-65242100 on Build GRCh38
rs558984012 Chr.2: 65242360-65242360 on Build GRCh38
rs563922867 Chr.2: 65242038-65242038 on Build GRCh38
rs974276316 Chr.2: 65242257-65242257 on Build GRCh38
rs1024014957 Chr.2: 65242137-65242137 on Build GRCh38
rs1020453026 Chr.2: 65241965-65241965 on Build GRCh38
rs930458852 Chr.2: 65242287-65242287 on Build GRCh38
rs747331188 Chr.2: 65241946-65241946 on Build GRCh38
rs900607761 Chr.2: 65242281-65242281 on Build GRCh38
rs375536973 Chr.2: 65242056-65242056 on Build GRCh38
rs749654955 Chr.2: 65242336-65242336 on Build GRCh38
rs977880699 Chr.2: 65242098-65242098 on Build GRCh38
rs978060738 Chr.2: 65242249-65242249 on Build GRCh38
rs372909860 Chr.2: 65242048-65242048 on Build GRCh38
rs746038665 Chr.2: 65241999-65241999 on Build GRCh38
rs35988716 Chr.2: 65242310-65242311 on Build GRCh38
rs770191316 Chr.2: 65242005-65242005 on Build GRCh38
rs1051208659 Chr.2: 65242266-65242266 on Build GRCh38
rs886350148 Chr.2: 65242320-65242320 on Build GRCh38
rs765822809 Chr.2: 65242075-65242075 on Build GRCh38
rs192140951 Chr.2: 65242242-65242242 on Build GRCh38
rs971094179 Chr.2: 65242153-65242154 on Build GRCh38

View Full Product Details