Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 67399080-67399591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETAA1
Entrez Gene ID
54465
Gene Name
Ewing tumor associated antigen 1
Gene Aliases
ETAA16
UniGene
Hs.353022
Gene Chromosome Location
Chr.2: 67397310-67410411 on Build GRCh38
Transcripts
NM_019002.3 XM_017004376.1 XM_017004377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4613092 c.362_363insA p.Q124fs*7 Chr.2: 67399559-67399560 on Build GRCh38
5465599 c.318T>G p.F106L Chr.2: 67399263-67399263 on Build GRCh38
5034826 c.259G>A p.D87N Chr.2: 67399204-67399204 on Build GRCh38
1022312 c.249G>A p.A83A Chr.2: 67399194-67399194 on Build GRCh38
1632082 c.242A>G p.K81R Chr.2: 67399187-67399187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867547092 Chr.2: 67399128-67399128 on Build GRCh38
rs774092806 Chr.2: 67399555-67399555 on Build GRCh38
rs573345443 Chr.2: 67399260-67399260 on Build GRCh38
rs779222859 Chr.2: 67399328-67399328 on Build GRCh38
rs1038418908 Chr.2: 67399348-67399348 on Build GRCh38
rs532275210 Chr.2: 67399193-67399193 on Build GRCh38
rs10209114 Chr.2: 67399206-67399206 on Build GRCh38
rs376605562 Chr.2: 67399250-67399250 on Build GRCh38
rs780875982 Chr.2: 67399229-67399231 on Build GRCh38
rs756170422 Chr.2: 67399300-67399300 on Build GRCh38
rs1056161260 Chr.2: 67399186-67399186 on Build GRCh38
rs373730082 Chr.2: 67399342-67399342 on Build GRCh38
rs968513872 Chr.2: 67399509-67399509 on Build GRCh38
rs145364981 Chr.2: 67399541-67399541 on Build GRCh38
rs758816435 Chr.2: 67399123-67399123 on Build GRCh38
rs996097515 Chr.2: 67399490-67399490 on Build GRCh38
rs200287867 Chr.2: 67399343-67399343 on Build GRCh38
rs199959789 Chr.2: 67399192-67399192 on Build GRCh38
rs756696193 Chr.2: 67399508-67399509 on Build GRCh38
rs754623782 Chr.2: 67399540-67399540 on Build GRCh38
rs570885483 Chr.2: 67399282-67399282 on Build GRCh38
rs778211775 Chr.2: 67399514-67399515 on Build GRCh38
rs1049672881 Chr.2: 67399366-67399366 on Build GRCh38
rs569998581 Chr.2: 67399506-67399506 on Build GRCh38
rs575547615 Chr.2: 67399483-67399483 on Build GRCh38
rs746062685 Chr.2: 67399539-67399539 on Build GRCh38
rs752437911 Chr.2: 67399122-67399122 on Build GRCh38
rs753200301 Chr.2: 67399314-67399315 on Build GRCh38
rs745860769 Chr.2: 67399525-67399525 on Build GRCh38
rs553516333 Chr.2: 67399256-67399256 on Build GRCh38
rs866253106 Chr.2: 67399266-67399266 on Build GRCh38
rs377043304 Chr.2: 67399522-67399522 on Build GRCh38
rs775269026 Chr.2: 67399231-67399231 on Build GRCh38
rs773954411 Chr.2: 67399122-67399131 on Build GRCh38
rs535036766 Chr.2: 67399323-67399323 on Build GRCh38
rs754502120 Chr.2: 67399187-67399188 on Build GRCh38
rs143819253 Chr.2: 67399258-67399258 on Build GRCh38
rs1051264015 Chr.2: 67399328-67399331 on Build GRCh38
rs745892449 Chr.2: 67399179-67399179 on Build GRCh38
rs925612701 Chr.2: 67399225-67399225 on Build GRCh38
rs757646215 Chr.2: 67399139-67399139 on Build GRCh38
rs368069429 Chr.2: 67399185-67399185 on Build GRCh38
rs201337200 Chr.2: 67399166-67399166 on Build GRCh38
rs781733766 Chr.2: 67399171-67399171 on Build GRCh38
rs921511921 Chr.2: 67399277-67399277 on Build GRCh38
rs772818206 Chr.2: 67399194-67399194 on Build GRCh38
rs189398507 Chr.2: 67399419-67399419 on Build GRCh38
rs1014074616 Chr.2: 67399440-67399441 on Build GRCh38
rs1018939297 Chr.2: 67399425-67399425 on Build GRCh38
rs369665535 Chr.2: 67399379-67399379 on Build GRCh38
rs886718980 Chr.2: 67399383-67399383 on Build GRCh38
rs750974206 Chr.2: 67399276-67399276 on Build GRCh38
rs762626955 Chr.2: 67399240-67399240 on Build GRCh38
rs898510263 Chr.2: 67399376-67399376 on Build GRCh38
rs746839075 Chr.2: 67399195-67399195 on Build GRCh38
rs764891696 Chr.2: 67399224-67399224 on Build GRCh38
rs113130006 Chr.2: 67399223-67399223 on Build GRCh38
rs769426341 Chr.2: 67399526-67399526 on Build GRCh38
rs780171493 Chr.2: 67399309-67399309 on Build GRCh38
rs749378760 Chr.2: 67399311-67399311 on Build GRCh38
rs778341383 Chr.2: 67399136-67399136 on Build GRCh38
rs755137984 Chr.2: 67399312-67399312 on Build GRCh38
rs763614211 Chr.2: 67399550-67399550 on Build GRCh38
rs201956399 Chr.2: 67399245-67399245 on Build GRCh38
rs1034247751 Chr.2: 67399127-67399127 on Build GRCh38

View Full Product Details