Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97537932-97538443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36B
Entrez Gene ID
57730
Gene Name
ankyrin repeat domain 36B
Gene Aliases
KIAA1641
UniGene
Hs.532921
Gene Chromosome Location
Chr.2: 97492663-97590245 on Build GRCh38
Transcripts
NM_025190.3 XM_005263995.4 XM_006712661.1 XM_011511555.1 XM_011511556.1 XM_011511557.2 XM_011511558.1 XM_017004578.1 XM_017004579.1 XM_017004580.1 XM_017004581.1 XM_017004582.1 XM_017004583.1 XM_017004584.1 XM_017004585.1 XM_017004586.1 XM_017004587.1 XM_017004588.1 XM_017004589.1 XM_017004590.1 XM_017004591.1 XM_017004592.1 XM_017004593.1 XM_017004594.1 XM_017004595.1

SNP Detail

SNP ID Chromosome Location
rs3925992 Chr.2: 97538088-97538088 on Build GRCh38
rs183246566 Chr.2: 97538153-97538153 on Build GRCh38
rs192471878 Chr.2: 97538143-97538143 on Build GRCh38
rs531233129 Chr.2: 97537959-97537959 on Build GRCh38
rs2258818 Chr.2: 97538160-97538160 on Build GRCh38
rs773526062 Chr.2: 97538152-97538152 on Build GRCh38
rs779488647 Chr.2: 97538389-97538389 on Build GRCh38
rs764454667 Chr.2: 97538187-97538187 on Build GRCh38
rs567702197 Chr.2: 97538029-97538029 on Build GRCh38
rs3925991 Chr.2: 97538115-97538115 on Build GRCh38
rs780982924 Chr.2: 97538305-97538305 on Build GRCh38
rs748879927 Chr.2: 97538131-97538131 on Build GRCh38
rs777261758 Chr.2: 97538158-97538158 on Build GRCh38
rs745749700 Chr.2: 97538344-97538344 on Build GRCh38
rs767610021 Chr.2: 97538151-97538152 on Build GRCh38
rs550990818 Chr.2: 97537960-97537960 on Build GRCh38
rs565501760 Chr.2: 97538156-97538156 on Build GRCh38
rs759966582 Chr.2: 97538159-97538159 on Build GRCh38
rs756057708 Chr.2: 97538345-97538345 on Build GRCh38
rs557674379 Chr.2: 97538213-97538213 on Build GRCh38
rs577471647 Chr.2: 97538252-97538252 on Build GRCh38
rs2463574 Chr.2: 97537958-97537958 on Build GRCh38
rs2442233 Chr.2: 97538420-97538420 on Build GRCh38
rs752848875 Chr.2: 97538186-97538186 on Build GRCh38
rs369666439 Chr.2: 97538191-97538191 on Build GRCh38

View Full Product Details