Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97551337-97551837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36B
Entrez Gene ID
57730
Gene Name
ankyrin repeat domain 36B
Gene Aliases
KIAA1641
UniGene
Hs.532921
Gene Chromosome Location
Chr.2: 97492663-97590245 on Build GRCh38
Transcripts
NM_025190.3 XM_005263995.4 XM_006712661.1 XM_011511555.1 XM_011511556.1 XM_011511557.2 XM_011511558.1 XM_017004578.1 XM_017004579.1 XM_017004580.1 XM_017004581.1 XM_017004582.1 XM_017004583.1 XM_017004584.1 XM_017004585.1 XM_017004586.1 XM_017004587.1 XM_017004588.1 XM_017004589.1 XM_017004590.1 XM_017004592.1 XM_017004593.1 XM_017004594.1 XM_017004595.1

SNP Detail

SNP ID Chromosome Location
rs930960975 Chr.2: 97551666-97551666 on Build GRCh38
rs370935310 Chr.2: 97551414-97551414 on Build GRCh38
rs763375366 Chr.2: 97551752-97551752 on Build GRCh38
rs764282744 Chr.2: 97551464-97551464 on Build GRCh38
rs775114867 Chr.2: 97551415-97551415 on Build GRCh38
rs777789109 Chr.2: 97551525-97551525 on Build GRCh38
rs539249832 Chr.2: 97551505-97551505 on Build GRCh38
rs770572561 Chr.2: 97551408-97551408 on Build GRCh38
rs1010276693 Chr.2: 97551713-97551713 on Build GRCh38
rs539162498 Chr.2: 97551434-97551434 on Build GRCh38
rs150582296 Chr.2: 97551372-97551372 on Build GRCh38
rs182907771 Chr.2: 97551712-97551712 on Build GRCh38
rs995777544 Chr.2: 97551495-97551495 on Build GRCh38
rs766605758 Chr.2: 97551438-97551438 on Build GRCh38
rs544277156 Chr.2: 97551533-97551533 on Build GRCh38
rs576064237 Chr.2: 97551629-97551629 on Build GRCh38
rs746457459 Chr.2: 97551667-97551667 on Build GRCh38
rs1049979448 Chr.2: 97551432-97551432 on Build GRCh38
rs373922937 Chr.2: 97551509-97551509 on Build GRCh38
rs376644275 Chr.2: 97551461-97551461 on Build GRCh38
rs192211646 Chr.2: 97551636-97551636 on Build GRCh38
rs774023237 Chr.2: 97551499-97551499 on Build GRCh38
rs367810469 Chr.2: 97551522-97551522 on Build GRCh38
rs777834431 Chr.2: 97551519-97551519 on Build GRCh38
rs544997880 Chr.2: 97551678-97551678 on Build GRCh38
rs755408504 Chr.2: 97551453-97551453 on Build GRCh38
rs574574681 Chr.2: 97551590-97551590 on Build GRCh38
rs755532881 Chr.2: 97551368-97551368 on Build GRCh38
rs750372671 Chr.2: 97551463-97551464 on Build GRCh38
rs370515955 Chr.2: 97551501-97551501 on Build GRCh38
rs895891176 Chr.2: 97551550-97551550 on Build GRCh38
rs567290576 Chr.2: 97551733-97551733 on Build GRCh38
rs762993076 Chr.2: 97551484-97551484 on Build GRCh38
rs536693016 Chr.2: 97551663-97551663 on Build GRCh38
rs758406268 Chr.2: 97551460-97551460 on Build GRCh38
rs768225416 Chr.2: 97551498-97551498 on Build GRCh38
rs539186577 Chr.2: 97551816-97551816 on Build GRCh38
rs749147829 Chr.2: 97551530-97551530 on Build GRCh38
rs373665293 Chr.2: 97551440-97551440 on Build GRCh38
rs555330358 Chr.2: 97551716-97551716 on Build GRCh38
rs943782725 Chr.2: 97551640-97551640 on Build GRCh38
rs751193025 Chr.2: 97551426-97551426 on Build GRCh38
rs556657078 Chr.2: 97551759-97551759 on Build GRCh38
rs560235954 Chr.2: 97551623-97551623 on Build GRCh38
rs747235926 Chr.2: 97551405-97551405 on Build GRCh38
rs972454195 Chr.2: 97551648-97551648 on Build GRCh38
rs937534516 Chr.2: 97551695-97551695 on Build GRCh38
rs1032698821 Chr.2: 97551789-97551790 on Build GRCh38
rs553591553 Chr.2: 97551458-97551458 on Build GRCh38
rs771212967 Chr.2: 97551465-97551465 on Build GRCh38
rs769602226 Chr.2: 97551477-97551477 on Build GRCh38
rs575894568 Chr.2: 97551513-97551513 on Build GRCh38
rs954055088 Chr.2: 97551624-97551624 on Build GRCh38
rs780815435 Chr.2: 97551466-97551466 on Build GRCh38
rs573991694 Chr.2: 97551751-97551751 on Build GRCh38
rs551562226 Chr.2: 97551680-97551680 on Build GRCh38
rs748268338 Chr.2: 97551383-97551383 on Build GRCh38
rs752999936 Chr.2: 97551414-97551415 on Build GRCh38
rs768407425 Chr.2: 97551720-97551720 on Build GRCh38
rs1972985 Chr.2: 97551592-97551592 on Build GRCh38
rs779524384 Chr.2: 97551454-97551454 on Build GRCh38
rs775638992 Chr.2: 97551483-97551483 on Build GRCh38
rs139975232 Chr.2: 97551586-97551586 on Build GRCh38
rs193279706 Chr.2: 97551753-97551753 on Build GRCh38
rs757112688 Chr.2: 97551570-97551570 on Build GRCh38
rs772260570 Chr.2: 97551388-97551388 on Build GRCh38
rs913849242 Chr.2: 97551379-97551379 on Build GRCh38
rs1001707605 Chr.2: 97551523-97551523 on Build GRCh38
rs763839070 Chr.2: 97551420-97551420 on Build GRCh38
rs745721155 Chr.2: 97551471-97551471 on Build GRCh38
rs765589914 Chr.2: 97551514-97551514 on Build GRCh38
rs1020258617 Chr.2: 97551572-97551572 on Build GRCh38
rs773489467 Chr.2: 97551400-97551400 on Build GRCh38
rs566579099 Chr.2: 97551796-97551796 on Build GRCh38
rs377458299 Chr.2: 97551412-97551412 on Build GRCh38
rs188707949 Chr.2: 97551732-97551732 on Build GRCh38
rs749703011 Chr.2: 97551362-97551362 on Build GRCh38
rs760771087 Chr.2: 97551438-97551438 on Build GRCh38

View Full Product Details