Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 227532047-227532550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGFG1
Entrez Gene ID
3267
Gene Name
ArfGAP with FG repeats 1
Gene Aliases
HRB, RAB, RIP
UniGene
Hs.352962
Gene Chromosome Location
Chr.2: 227472169-227561222 on Build GRCh38
Transcripts
NM_001135187.1 NM_001135188.1 NM_001135189.1 NM_004504.4 XM_005246516.3 XM_006712478.3 XM_006712479.3 XM_006712480.3 XM_006712481.3 XM_017003954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5502626 c.815-9G>A p.? Chr.2: 227532129-227532129 on Build GRCh38
6534888 c.872T>G p.L291R Chr.2: 227532195-227532195 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774277017 Chr.2: 227532265-227532266 on Build GRCh38
rs750543397 Chr.2: 227532099-227532099 on Build GRCh38
rs756164230 Chr.2: 227532101-227532101 on Build GRCh38
rs545298387 Chr.2: 227532228-227532228 on Build GRCh38
rs560767631 Chr.2: 227532287-227532287 on Build GRCh38
rs182869936 Chr.2: 227532239-227532239 on Build GRCh38
rs971882983 Chr.2: 227532222-227532222 on Build GRCh38
rs1028968203 Chr.2: 227532133-227532133 on Build GRCh38
rs569187282 Chr.2: 227532484-227532484 on Build GRCh38
rs905359481 Chr.2: 227532336-227532336 on Build GRCh38
rs543514448 Chr.2: 227532427-227532427 on Build GRCh38
rs1020592867 Chr.2: 227532461-227532461 on Build GRCh38
rs368982901 Chr.2: 227532168-227532168 on Build GRCh38
rs536595893 Chr.2: 227532503-227532503 on Build GRCh38
rs1031146852 Chr.2: 227532369-227532369 on Build GRCh38
rs986730714 Chr.2: 227532279-227532279 on Build GRCh38
rs867832943 Chr.2: 227532198-227532198 on Build GRCh38
rs752530796 Chr.2: 227532353-227532358 on Build GRCh38
rs530858857 Chr.2: 227532241-227532241 on Build GRCh38
rs920440768 Chr.2: 227532226-227532226 on Build GRCh38
rs745716512 Chr.2: 227532180-227532180 on Build GRCh38
rs930712729 Chr.2: 227532327-227532327 on Build GRCh38
rs967766387 Chr.2: 227532172-227532172 on Build GRCh38
rs1009255638 Chr.2: 227532423-227532423 on Build GRCh38
rs187036374 Chr.2: 227532333-227532333 on Build GRCh38
rs760396002 Chr.2: 227532464-227532464 on Build GRCh38
rs577521685 Chr.2: 227532325-227532325 on Build GRCh38
rs372968413 Chr.2: 227532182-227532182 on Build GRCh38
rs938288040 Chr.2: 227532340-227532340 on Build GRCh38
rs758972194 Chr.2: 227532235-227532235 on Build GRCh38
rs550827001 Chr.2: 227532419-227532419 on Build GRCh38
rs748410091 Chr.2: 227532234-227532234 on Build GRCh38
rs538868469 Chr.2: 227532383-227532383 on Build GRCh38
rs767251540 Chr.2: 227532388-227532391 on Build GRCh38
rs528177919 Chr.2: 227532300-227532300 on Build GRCh38
rs920427170 Chr.2: 227532506-227532509 on Build GRCh38
rs989992258 Chr.2: 227532108-227532108 on Build GRCh38
rs775420408 Chr.2: 227532495-227532495 on Build GRCh38
rs767411964 Chr.2: 227532270-227532270 on Build GRCh38
rs571459466 Chr.2: 227532372-227532372 on Build GRCh38
rs953387823 Chr.2: 227532159-227532159 on Build GRCh38
rs1018958485 Chr.2: 227532465-227532465 on Build GRCh38
rs755087442 Chr.2: 227532220-227532220 on Build GRCh38
rs779347436 Chr.2: 227532221-227532221 on Build GRCh38
rs548938768 Chr.2: 227532285-227532285 on Build GRCh38
rs1056718314 Chr.2: 227532368-227532368 on Build GRCh38
rs1009348940 Chr.2: 227532074-227532074 on Build GRCh38
rs940499094 Chr.2: 227532194-227532194 on Build GRCh38
rs139412422 Chr.2: 227532520-227532520 on Build GRCh38
rs972018248 Chr.2: 227532306-227532306 on Build GRCh38
rs979677589 Chr.2: 227532184-227532184 on Build GRCh38

View Full Product Details