Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 488491-488988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK2A1
Entrez Gene ID
1457
Gene Name
casein kinase 2 alpha 1
Gene Aliases
CK2A1, CKII, CSNK2A3
UniGene
Hs.644056
Gene Chromosome Location
Chr.20: 482694-543838 on Build GRCh38
Transcripts
NM_001895.3 NM_177559.2 NM_177560.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
419358 c.816C>G p.I272M Chr.20: 488686-488686 on Build GRCh38
6430900 c.767A>G p.D256G Chr.20: 488735-488735 on Build GRCh38
1732302 c.724-2A>G p.? Chr.20: 488780-488780 on Build GRCh38
4390387 c.809A>G p.N270S Chr.20: 488693-488693 on Build GRCh38
7165655 c.799C>G p.P267A Chr.20: 488703-488703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757090146 Chr.20: 488657-488657 on Build GRCh38
rs754970504 Chr.20: 488873-488873 on Build GRCh38
rs531239124 Chr.20: 488892-488892 on Build GRCh38
rs760403708 Chr.20: 488560-488560 on Build GRCh38
rs770999955 Chr.20: 488874-488874 on Build GRCh38
rs755426312 Chr.20: 488727-488727 on Build GRCh38
rs1054207603 Chr.20: 488609-488609 on Build GRCh38
rs749790755 Chr.20: 488716-488716 on Build GRCh38
rs372072408 Chr.20: 488805-488805 on Build GRCh38
rs150398280 Chr.20: 488685-488685 on Build GRCh38
rs869312846 Chr.20: 488676-488676 on Build GRCh38
rs375633788 Chr.20: 488693-488693 on Build GRCh38
rs551703352 Chr.20: 488817-488817 on Build GRCh38
rs755588741 Chr.20: 488687-488687 on Build GRCh38
rs752761226 Chr.20: 488910-488910 on Build GRCh38
rs1000651242 Chr.20: 488515-488515 on Build GRCh38
rs764170352 Chr.20: 488821-488821 on Build GRCh38
rs1042686979 Chr.20: 488953-488953 on Build GRCh38
rs753774206 Chr.20: 488864-488864 on Build GRCh38
rs764007004 Chr.20: 488646-488646 on Build GRCh38
rs773320713 Chr.20: 488782-488782 on Build GRCh38
rs747188536 Chr.20: 488802-488802 on Build GRCh38
rs941516483 Chr.20: 488777-488777 on Build GRCh38
rs551283577 Chr.20: 488906-488906 on Build GRCh38
rs185032703 Chr.20: 488593-488593 on Build GRCh38
rs982977815 Chr.20: 488674-488674 on Build GRCh38
rs180695439 Chr.20: 488549-488549 on Build GRCh38
rs368630997 Chr.20: 488749-488749 on Build GRCh38
rs936649158 Chr.20: 488595-488595 on Build GRCh38
rs1026390056 Chr.20: 488890-488890 on Build GRCh38
rs751452414 Chr.20: 488651-488651 on Build GRCh38
rs754145602 Chr.20: 488541-488541 on Build GRCh38
rs144288682 Chr.20: 488535-488535 on Build GRCh38
rs917198515 Chr.20: 488931-488931 on Build GRCh38
rs565429343 Chr.20: 488859-488859 on Build GRCh38
rs766240990 Chr.20: 488617-488617 on Build GRCh38
rs767343481 Chr.20: 488660-488660 on Build GRCh38
rs779368725 Chr.20: 488731-488731 on Build GRCh38
rs932687727 Chr.20: 488849-488849 on Build GRCh38
rs61742776 Chr.20: 488760-488760 on Build GRCh38
rs1045128070 Chr.20: 488558-488558 on Build GRCh38
rs963460433 Chr.20: 488833-488833 on Build GRCh38

View Full Product Details