Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24035361-24035864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABIN1
Entrez Gene ID
23523
Gene Name
calcineurin binding protein 1
Gene Aliases
CAIN, KB-318B8.7, PPP3IN
UniGene
Hs.517478
Gene Chromosome Location
Chr.22: 24011192-24178628 on Build GRCh38
Transcripts
NM_001199281.1 NM_001201429.1 NM_012295.3 XM_005261415.2 XM_005261416.2 XM_005261417.2 XM_005261419.2 XM_006724179.1 XM_006724180.1 XM_006724181.1 XM_011530026.1 XM_011530029.2 XM_011530030.2 XM_017028680.1 XM_017028681.1 XM_017028683.1

SNP Detail

SNP ID Chromosome Location
rs116201419 Chr.22: 24035633-24035633 on Build GRCh38
rs894508236 Chr.22: 24035460-24035460 on Build GRCh38
rs919751853 Chr.22: 24035814-24035814 on Build GRCh38
rs778711037 Chr.22: 24035509-24035510 on Build GRCh38
rs749848388 Chr.22: 24035565-24035565 on Build GRCh38
rs754840857 Chr.22: 24035513-24035513 on Build GRCh38
rs551230052 Chr.22: 24035704-24035704 on Build GRCh38
rs956934232 Chr.22: 24035692-24035692 on Build GRCh38
rs375515565 Chr.22: 24035512-24035512 on Build GRCh38
rs910902581 Chr.22: 24035765-24035765 on Build GRCh38
rs757505434 Chr.22: 24035537-24035537 on Build GRCh38
rs979547974 Chr.22: 24035386-24035386 on Build GRCh38
rs938768933 Chr.22: 24035395-24035395 on Build GRCh38
rs896469696 Chr.22: 24035580-24035580 on Build GRCh38
rs747896382 Chr.22: 24035513-24035513 on Build GRCh38
rs775798611 Chr.22: 24035562-24035562 on Build GRCh38
rs753788122 Chr.22: 24035504-24035504 on Build GRCh38
rs760707363 Chr.22: 24035456-24035456 on Build GRCh38
rs781331053 Chr.22: 24035543-24035543 on Build GRCh38
rs973062972 Chr.22: 24035609-24035609 on Build GRCh38
rs568111322 Chr.22: 24035716-24035716 on Build GRCh38
rs747054795 Chr.22: 24035536-24035536 on Build GRCh38
rs929860227 Chr.22: 24035835-24035835 on Build GRCh38
rs983350249 Chr.22: 24035651-24035651 on Build GRCh38
rs751497544 Chr.22: 24035585-24035585 on Build GRCh38
rs948702560 Chr.22: 24035474-24035474 on Build GRCh38
rs973872177 Chr.22: 24035807-24035807 on Build GRCh38
rs368328330 Chr.22: 24035547-24035547 on Build GRCh38
rs1043326842 Chr.22: 24035487-24035487 on Build GRCh38
rs535523839 Chr.22: 24035725-24035725 on Build GRCh38
rs890604159 Chr.22: 24035533-24035533 on Build GRCh38
rs781322406 Chr.22: 24035697-24035697 on Build GRCh38
rs1023611105 Chr.22: 24035617-24035617 on Build GRCh38
rs1047340339 Chr.22: 24035839-24035839 on Build GRCh38
rs200563699 Chr.22: 24035499-24035499 on Build GRCh38
rs942412637 Chr.22: 24035767-24035767 on Build GRCh38
rs777958662 Chr.22: 24035514-24035514 on Build GRCh38
rs986440414 Chr.22: 24035753-24035753 on Build GRCh38
rs763698581 Chr.22: 24035478-24035478 on Build GRCh38
rs927806818 Chr.22: 24035703-24035703 on Build GRCh38
rs750252323 Chr.22: 24035496-24035496 on Build GRCh38
rs1000934649 Chr.22: 24035638-24035638 on Build GRCh38
rs925488810 Chr.22: 24035411-24035411 on Build GRCh38
rs950702808 Chr.22: 24035642-24035642 on Build GRCh38
rs1047013516 Chr.22: 24035551-24035551 on Build GRCh38
rs533378270 Chr.22: 24035586-24035586 on Build GRCh38
rs1013502259 Chr.22: 24035582-24035582 on Build GRCh38
rs544346168 Chr.22: 24035570-24035570 on Build GRCh38
rs112382602 Chr.22: 24035824-24035824 on Build GRCh38
rs933708095 Chr.22: 24035469-24035469 on Build GRCh38
rs912889673 Chr.22: 24035467-24035467 on Build GRCh38
rs557126182 Chr.22: 24035816-24035816 on Build GRCh38
rs200432141 Chr.22: 24035492-24035492 on Build GRCh38
rs562727604 Chr.22: 24035583-24035583 on Build GRCh38
rs1032908092 Chr.22: 24035660-24035660 on Build GRCh38
rs988436717 Chr.22: 24035461-24035461 on Build GRCh38
rs1057187708 Chr.22: 24035431-24035431 on Build GRCh38
rs552022718 Chr.22: 24035610-24035610 on Build GRCh38
rs528050652 Chr.22: 24035644-24035644 on Build GRCh38
rs867408130 Chr.22: 24035440-24035440 on Build GRCh38
rs773707402 Chr.22: 24035481-24035481 on Build GRCh38
rs189271579 Chr.22: 24035493-24035493 on Build GRCh38
rs755364786 Chr.22: 24035710-24035710 on Build GRCh38

View Full Product Details