Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 25674145-25674670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK3
Entrez Gene ID
157
Gene Name
G protein-coupled receptor kinase 3
Gene Aliases
ADRBK2, BARK2
UniGene
Hs.657494
Gene Chromosome Location
Chr.22: 25564894-25729294 on Build GRCh38
Transcripts
NM_005160.3 XM_011529975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1032808 c.634G>C p.D212H Chr.22: 25674515-25674515 on Build GRCh38
4654847 c.595C>T p.R199* Chr.22: 25674476-25674476 on Build GRCh38
6595586 c.563T>C p.M188T Chr.22: 25674444-25674444 on Build GRCh38
5587147 c.584G>A p.R195K Chr.22: 25674465-25674465 on Build GRCh38
4555084 c.640G>A p.G214R Chr.22: 25674521-25674521 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750531032 Chr.22: 25674427-25674427 on Build GRCh38
rs766371590 Chr.22: 25674389-25674389 on Build GRCh38
rs746558457 Chr.22: 25674490-25674490 on Build GRCh38
rs778442347 Chr.22: 25674280-25674280 on Build GRCh38
rs1047187544 Chr.22: 25674610-25674610 on Build GRCh38
rs930101249 Chr.22: 25674604-25674604 on Build GRCh38
rs374198066 Chr.22: 25674456-25674456 on Build GRCh38
rs756157357 Chr.22: 25674572-25674572 on Build GRCh38
rs147019446 Chr.22: 25674222-25674222 on Build GRCh38
rs759329352 Chr.22: 25674402-25674402 on Build GRCh38
rs894848001 Chr.22: 25674510-25674510 on Build GRCh38
rs989866260 Chr.22: 25674354-25674354 on Build GRCh38
rs1024230350 Chr.22: 25674191-25674191 on Build GRCh38
rs563297548 Chr.22: 25674177-25674177 on Build GRCh38
rs886085726 Chr.22: 25674207-25674207 on Build GRCh38
rs764259673 Chr.22: 25674406-25674406 on Build GRCh38
rs138180634 Chr.22: 25674352-25674355 on Build GRCh38
rs372621335 Chr.22: 25674408-25674408 on Build GRCh38
rs896532468 Chr.22: 25674611-25674611 on Build GRCh38
rs568094636 Chr.22: 25674365-25674365 on Build GRCh38
rs780724604 Chr.22: 25674452-25674452 on Build GRCh38
rs970038953 Chr.22: 25674217-25674217 on Build GRCh38
rs1024753755 Chr.22: 25674575-25674575 on Build GRCh38
rs778662378 Chr.22: 25674470-25674470 on Build GRCh38
rs754515438 Chr.22: 25674187-25674187 on Build GRCh38
rs370546452 Chr.22: 25674336-25674336 on Build GRCh38
rs994277384 Chr.22: 25674168-25674168 on Build GRCh38
rs959086334 Chr.22: 25674242-25674242 on Build GRCh38
rs759276190 Chr.22: 25674540-25674540 on Build GRCh38
rs764992709 Chr.22: 25674543-25674543 on Build GRCh38
rs568634390 Chr.22: 25674306-25674306 on Build GRCh38
rs978350090 Chr.22: 25674308-25674308 on Build GRCh38
rs923103516 Chr.22: 25674451-25674451 on Build GRCh38
rs757467902 Chr.22: 25674420-25674420 on Build GRCh38
rs755429053 Chr.22: 25674466-25674466 on Build GRCh38
rs770788642 Chr.22: 25674506-25674506 on Build GRCh38
rs762088342 Chr.22: 25674547-25674547 on Build GRCh38
rs1037330019 Chr.22: 25674504-25674504 on Build GRCh38
rs368573348 Chr.22: 25674386-25674386 on Build GRCh38
rs988537401 Chr.22: 25674314-25674314 on Build GRCh38
rs138092452 Chr.22: 25674305-25674305 on Build GRCh38
rs776580964 Chr.22: 25674516-25674516 on Build GRCh38
rs191078747 Chr.22: 25674549-25674549 on Build GRCh38
rs751800954 Chr.22: 25674411-25674411 on Build GRCh38
rs781297701 Chr.22: 25674423-25674423 on Build GRCh38
rs986470685 Chr.22: 25674380-25674380 on Build GRCh38
rs1025140688 Chr.22: 25674294-25674294 on Build GRCh38
rs772775988 Chr.22: 25674488-25674488 on Build GRCh38
rs942996669 Chr.22: 25674555-25674555 on Build GRCh38
rs898681885 Chr.22: 25674584-25674584 on Build GRCh38
rs200278215 Chr.22: 25674541-25674541 on Build GRCh38
rs755592496 Chr.22: 25674578-25674578 on Build GRCh38
rs775186156 Chr.22: 25674545-25674545 on Build GRCh38
rs535157417 Chr.22: 25674391-25674391 on Build GRCh38
rs572939587 Chr.22: 25674175-25674175 on Build GRCh38
rs534954440 Chr.22: 25674315-25674315 on Build GRCh38
rs756626950 Chr.22: 25674442-25674442 on Build GRCh38
rs767686315 Chr.22: 25674548-25674548 on Build GRCh38
rs766955414 Chr.22: 25674573-25674573 on Build GRCh38
rs1047627188 Chr.22: 25674463-25674463 on Build GRCh38
rs754422442 Chr.22: 25674574-25674574 on Build GRCh38
rs771664171 Chr.22: 25674487-25674487 on Build GRCh38
rs747815978 Chr.22: 25674477-25674477 on Build GRCh38
rs552473926 Chr.22: 25674224-25674224 on Build GRCh38

View Full Product Details