Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 28689029-28689534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHEK2
Entrez Gene ID
11200
Gene Name
checkpoint kinase 2
Gene Aliases
CDS1, CHK2, HuCds1, LFS2, PP1425, RAD53, hCds1
UniGene
Hs.291363
Gene Chromosome Location
Chr.22: 28687743-28741866 on Build GRCh38
Transcripts
NM_001005735.1 NM_001257387.1 NM_007194.3 NM_145862.2 XM_006724114.3 XM_006724116.2 XM_011529839.2 XM_011529840.2 XM_011529841.1 XM_011529842.2 XM_011529843.1 XM_011529845.2 XM_017028560.1 XM_017028561.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6056969 c.1525C>T p.P509S Chr.22: 28689152-28689152 on Build GRCh38
1033061 c.1542+1G>T p.? Chr.22: 28689134-28689134 on Build GRCh38
4298198 c.357G>C p.L119L Chr.22: 28689180-28689180 on Build GRCh38
6305186 c.1616A>C p.Q539P Chr.22: 28689190-28689190 on Build GRCh38
6956828 c.1530G>T p.Q510H Chr.22: 28689147-28689147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78525381 Chr.22: 28689072-28689072 on Build GRCh38
rs17881716 Chr.22: 28689124-28689124 on Build GRCh38
rs876658673 Chr.22: 28689218-28689218 on Build GRCh38
rs753168652 Chr.22: 28689314-28689314 on Build GRCh38
rs747797219 Chr.22: 28689135-28689135 on Build GRCh38
rs184381247 Chr.22: 28689374-28689374 on Build GRCh38
rs187789144 Chr.22: 28689352-28689352 on Build GRCh38
rs898342452 Chr.22: 28689201-28689201 on Build GRCh38
rs747767986 Chr.22: 28689235-28689235 on Build GRCh38
rs587782489 Chr.22: 28689167-28689167 on Build GRCh38
rs786201149 Chr.22: 28689178-28689178 on Build GRCh38
rs28909981 Chr.22: 28689178-28689178 on Build GRCh38
rs869312552 Chr.22: 28689396-28689396 on Build GRCh38
rs750983184 Chr.22: 28689089-28689089 on Build GRCh38
rs754339938 Chr.22: 28689197-28689197 on Build GRCh38
rs754321880 Chr.22: 28689298-28689298 on Build GRCh38
rs1029269380 Chr.22: 28689375-28689375 on Build GRCh38
rs760968095 Chr.22: 28689405-28689405 on Build GRCh38
rs201208235 Chr.22: 28689312-28689313 on Build GRCh38
rs762041905 Chr.22: 28689186-28689186 on Build GRCh38
rs587782575 Chr.22: 28689217-28689217 on Build GRCh38
rs746578090 Chr.22: 28689247-28689247 on Build GRCh38
rs968907192 Chr.22: 28689355-28689355 on Build GRCh38
rs767043399 Chr.22: 28689195-28689195 on Build GRCh38
rs761750423 Chr.22: 28689181-28689181 on Build GRCh38
rs773547462 Chr.22: 28689284-28689284 on Build GRCh38
rs587780890 Chr.22: 28689180-28689180 on Build GRCh38
rs4035546 Chr.22: 28689087-28689087 on Build GRCh38
rs4035547 Chr.22: 28689128-28689128 on Build GRCh38
rs886041173 Chr.22: 28689215-28689215 on Build GRCh38
rs753299053 Chr.22: 28689333-28689333 on Build GRCh38
rs143965148 Chr.22: 28689188-28689188 on Build GRCh38
rs587781960 Chr.22: 28689164-28689164 on Build GRCh38
rs777394761 Chr.22: 28689246-28689246 on Build GRCh38
rs755454399 Chr.22: 28689299-28689299 on Build GRCh38
rs528610668 Chr.22: 28689436-28689436 on Build GRCh38
rs931170322 Chr.22: 28689483-28689483 on Build GRCh38
rs749728970 Chr.22: 28689119-28689119 on Build GRCh38
rs876660087 Chr.22: 28689157-28689157 on Build GRCh38
rs1002573308 Chr.22: 28689337-28689337 on Build GRCh38
rs905509335 Chr.22: 28689319-28689319 on Build GRCh38
rs1052262892 Chr.22: 28689159-28689159 on Build GRCh38
rs17882942 Chr.22: 28689143-28689143 on Build GRCh38
rs532203040 Chr.22: 28689291-28689291 on Build GRCh38
rs730881707 Chr.22: 28689222-28689222 on Build GRCh38
rs876660898 Chr.22: 28689155-28689155 on Build GRCh38
rs17883429 Chr.22: 28689354-28689354 on Build GRCh38
rs770402705 Chr.22: 28689259-28689259 on Build GRCh38
rs587782707 Chr.22: 28689173-28689174 on Build GRCh38
rs587782541 Chr.22: 28689151-28689151 on Build GRCh38
rs1035779067 Chr.22: 28689340-28689340 on Build GRCh38
rs766525226 Chr.22: 28689295-28689295 on Build GRCh38
rs769315091 Chr.22: 28689398-28689399 on Build GRCh38
rs757998634 Chr.22: 28689237-28689237 on Build GRCh38
rs549945536 Chr.22: 28689187-28689187 on Build GRCh38
rs756250205 Chr.22: 28689191-28689191 on Build GRCh38
rs762653025 Chr.22: 28689280-28689280 on Build GRCh38
rs886039512 Chr.22: 28689149-28689149 on Build GRCh38
rs769493672 Chr.22: 28689276-28689276 on Build GRCh38
rs587780178 Chr.22: 28689207-28689207 on Build GRCh38
rs772228947 Chr.22: 28689289-28689289 on Build GRCh38
rs774175654 Chr.22: 28689188-28689188 on Build GRCh38
rs17883172 Chr.22: 28689176-28689176 on Build GRCh38
rs765523546 Chr.22: 28689312-28689312 on Build GRCh38
rs776585043 Chr.22: 28689264-28689264 on Build GRCh38
rs775292304 Chr.22: 28689279-28689279 on Build GRCh38
rs569879751 Chr.22: 28689498-28689498 on Build GRCh38
rs554654938 Chr.22: 28689390-28689390 on Build GRCh38
rs919757475 Chr.22: 28689476-28689476 on Build GRCh38
rs17886970 Chr.22: 28689426-28689426 on Build GRCh38
rs375281470 Chr.22: 28689269-28689269 on Build GRCh38
rs587780179 Chr.22: 28689152-28689152 on Build GRCh38
rs767910233 Chr.22: 28689281-28689281 on Build GRCh38
rs756624380 Chr.22: 28689100-28689100 on Build GRCh38
rs17883639 Chr.22: 28689413-28689413 on Build GRCh38
rs780704259 Chr.22: 28689102-28689102 on Build GRCh38
rs531931623 Chr.22: 28689183-28689183 on Build GRCh38

View Full Product Details