Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89258736-89259274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP5
Entrez Gene ID
115362
Gene Name
guanylate binding protein 5
Gene Aliases
GBP-5
UniGene
Hs.513726
Gene Chromosome Location
Chr.1: 89258950-89272861 on Build GRCh38
Transcripts
NM_001134486.2 NM_052942.3

SNP Detail

SNP ID Chromosome Location
rs1007882679 Chr.1: 89258875-89258875 on Build GRCh38
rs1031574661 Chr.1: 89259033-89259033 on Build GRCh38
rs889079352 Chr.1: 89258858-89258858 on Build GRCh38
rs182837779 Chr.1: 89258909-89258909 on Build GRCh38
rs569772931 Chr.1: 89258834-89258834 on Build GRCh38
rs957249706 Chr.1: 89258779-89258780 on Build GRCh38
rs519086 Chr.1: 89258962-89258962 on Build GRCh38
rs115248047 Chr.1: 89259107-89259107 on Build GRCh38
rs1031630028 Chr.1: 89258767-89258767 on Build GRCh38
rs192088474 Chr.1: 89258883-89258883 on Build GRCh38
rs878987288 Chr.1: 89258772-89258772 on Build GRCh38
rs538863014 Chr.1: 89258932-89258932 on Build GRCh38
rs924627252 Chr.1: 89258769-89258769 on Build GRCh38
rs34953942 Chr.1: 89258831-89258832 on Build GRCh38
rs958992204 Chr.1: 89259084-89259084 on Build GRCh38
rs374061852 Chr.1: 89258828-89258828 on Build GRCh38
rs1017893232 Chr.1: 89258987-89258987 on Build GRCh38
rs540088008 Chr.1: 89258819-89258819 on Build GRCh38
rs989856978 Chr.1: 89258783-89258783 on Build GRCh38
rs1017237355 Chr.1: 89259069-89259069 on Build GRCh38
rs987524761 Chr.1: 89259250-89259250 on Build GRCh38
rs924720899 Chr.1: 89259161-89259161 on Build GRCh38
rs1011921294 Chr.1: 89259052-89259052 on Build GRCh38
rs1055107126 Chr.1: 89258913-89258913 on Build GRCh38
rs1021812924 Chr.1: 89259110-89259110 on Build GRCh38
rs977545681 Chr.1: 89259157-89259157 on Build GRCh38
rs778185277 Chr.1: 89258933-89258936 on Build GRCh38
rs964465628 Chr.1: 89258799-89258800 on Build GRCh38
rs949459712 Chr.1: 89258934-89258934 on Build GRCh38
rs933351865 Chr.1: 89258797-89258797 on Build GRCh38
rs149248585 Chr.1: 89259050-89259050 on Build GRCh38
rs749759325 Chr.1: 89258937-89258937 on Build GRCh38
rs1037549526 Chr.1: 89258877-89258877 on Build GRCh38
rs933405906 Chr.1: 89259224-89259224 on Build GRCh38
rs975811565 Chr.1: 89258799-89258799 on Build GRCh38
rs1046559511 Chr.1: 89258947-89258947 on Build GRCh38
rs968829023 Chr.1: 89259130-89259130 on Build GRCh38
rs1026042020 Chr.1: 89258938-89258938 on Build GRCh38
rs906270497 Chr.1: 89258948-89258948 on Build GRCh38
rs1061953 Chr.1: 89259235-89259235 on Build GRCh38

View Full Product Details