Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89266230-89266739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP5
Entrez Gene ID
115362
Gene Name
guanylate binding protein 5
Gene Aliases
GBP-5
UniGene
Hs.513726
Gene Chromosome Location
Chr.1: 89258950-89272861 on Build GRCh38
Transcripts
NM_001134486.2 NM_052942.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3369862 c.681C>G p.F227L Chr.1: 89266533-89266533 on Build GRCh38
3751442 c.660C>T p.P220P Chr.1: 89266554-89266554 on Build GRCh38
913017 c.626-2A>C p.? Chr.1: 89266590-89266590 on Build GRCh38
1561042 c.868+1G>A p.? Chr.1: 89266345-89266345 on Build GRCh38
6064588 c.768A>G p.L256L Chr.1: 89266446-89266446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772178189 Chr.1: 89266465-89266465 on Build GRCh38
rs1021699416 Chr.1: 89266298-89266298 on Build GRCh38
rs267598757 Chr.1: 89266386-89266386 on Build GRCh38
rs147424374 Chr.1: 89266302-89266302 on Build GRCh38
rs201216517 Chr.1: 89266388-89266388 on Build GRCh38
rs745501766 Chr.1: 89266329-89266329 on Build GRCh38
rs894724260 Chr.1: 89266550-89266550 on Build GRCh38
rs569908396 Chr.1: 89266307-89266307 on Build GRCh38
rs778827233 Chr.1: 89266448-89266448 on Build GRCh38
rs377547796 Chr.1: 89266553-89266553 on Build GRCh38
rs920380389 Chr.1: 89266489-89266489 on Build GRCh38
rs756041388 Chr.1: 89266539-89266540 on Build GRCh38
rs767007824 Chr.1: 89266395-89266395 on Build GRCh38
rs938998949 Chr.1: 89266521-89266521 on Build GRCh38
rs1030510306 Chr.1: 89266441-89266441 on Build GRCh38
rs777811088 Chr.1: 89266561-89266561 on Build GRCh38
rs907086997 Chr.1: 89266268-89266268 on Build GRCh38
rs369440866 Chr.1: 89266426-89266426 on Build GRCh38
rs34148688 Chr.1: 89266489-89266489 on Build GRCh38
rs774554369 Chr.1: 89266609-89266609 on Build GRCh38
rs12729209 Chr.1: 89266276-89266276 on Build GRCh38
rs767245260 Chr.1: 89266522-89266522 on Build GRCh38
rs958890588 Chr.1: 89266287-89266287 on Build GRCh38
rs544359636 Chr.1: 89266404-89266404 on Build GRCh38
rs559067004 Chr.1: 89266470-89266470 on Build GRCh38
rs776917882 Chr.1: 89266370-89266370 on Build GRCh38
rs749445485 Chr.1: 89266673-89266673 on Build GRCh38
rs887358170 Chr.1: 89266263-89266263 on Build GRCh38
rs758289601 Chr.1: 89266555-89266555 on Build GRCh38
rs780691495 Chr.1: 89266305-89266305 on Build GRCh38
rs752690565 Chr.1: 89266530-89266530 on Build GRCh38
rs765930716 Chr.1: 89266541-89266541 on Build GRCh38
rs757815053 Chr.1: 89266393-89266393 on Build GRCh38
rs751949710 Chr.1: 89266389-89266389 on Build GRCh38
rs768959736 Chr.1: 89266604-89266604 on Build GRCh38
rs761825347 Chr.1: 89266361-89266361 on Build GRCh38
rs779805988 Chr.1: 89266346-89266346 on Build GRCh38
rs201259247 Chr.1: 89266420-89266420 on Build GRCh38
rs748992424 Chr.1: 89266349-89266349 on Build GRCh38
rs201141768 Chr.1: 89266591-89266591 on Build GRCh38
rs954756909 Chr.1: 89266641-89266641 on Build GRCh38
rs983247154 Chr.1: 89266712-89266712 on Build GRCh38
rs769936809 Chr.1: 89266585-89266585 on Build GRCh38
rs140405391 Chr.1: 89266345-89266345 on Build GRCh38
rs762274852 Chr.1: 89266517-89266517 on Build GRCh38
rs1053163490 Chr.1: 89266282-89266282 on Build GRCh38
rs754727393 Chr.1: 89266552-89266552 on Build GRCh38
rs764349155 Chr.1: 89266512-89266512 on Build GRCh38
rs964619279 Chr.1: 89266676-89266676 on Build GRCh38
rs1045588848 Chr.1: 89266265-89266265 on Build GRCh38
rs774093264 Chr.1: 89266358-89266358 on Build GRCh38
rs771123564 Chr.1: 89266366-89266366 on Build GRCh38
rs759802738 Chr.1: 89266373-89266373 on Build GRCh38
rs745891638 Chr.1: 89266476-89266476 on Build GRCh38
rs61729628 Chr.1: 89266554-89266554 on Build GRCh38
rs749973130 Chr.1: 89266402-89266402 on Build GRCh38
rs998922219 Chr.1: 89266556-89266556 on Build GRCh38
rs771204823 Chr.1: 89266707-89266707 on Build GRCh38
rs770024893 Chr.1: 89266482-89266482 on Build GRCh38
rs768530163 Chr.1: 89266354-89266354 on Build GRCh38
rs987455309 Chr.1: 89266652-89266652 on Build GRCh38
rs781079393 Chr.1: 89266669-89266669 on Build GRCh38
rs746947192 Chr.1: 89266574-89266574 on Build GRCh38
rs61730934 Chr.1: 89266511-89266511 on Build GRCh38
rs759335217 Chr.1: 89266483-89266483 on Build GRCh38
rs148476506 Chr.1: 89266678-89266678 on Build GRCh38
rs765603813 Chr.1: 89266376-89266376 on Build GRCh38
rs748033916 Chr.1: 89266449-89266449 on Build GRCh38
rs775507526 Chr.1: 89266488-89266488 on Build GRCh38
rs754673294 Chr.1: 89266447-89266447 on Build GRCh38
rs750962076 Chr.1: 89266536-89266536 on Build GRCh38
rs749463583 Chr.1: 89266594-89266594 on Build GRCh38
rs551144729 Chr.1: 89266692-89266692 on Build GRCh38
rs1003091204 Chr.1: 89266270-89266270 on Build GRCh38
rs776829331 Chr.1: 89266472-89266472 on Build GRCh38
rs199813318 Chr.1: 89266516-89266516 on Build GRCh38
rs145007481 Chr.1: 89266427-89266427 on Build GRCh38
rs200485655 Chr.1: 89266383-89266383 on Build GRCh38
rs567349374 Chr.1: 89266706-89266706 on Build GRCh38
rs751175245 Chr.1: 89266340-89266343 on Build GRCh38
rs58111009 Chr.1: 89266295-89266295 on Build GRCh38
rs755558615 Chr.1: 89266539-89266539 on Build GRCh38

View Full Product Details