Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93904432-93904886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCLM
Entrez Gene ID
2730
Gene Name
glutamate-cysteine ligase modifier subunit
Gene Aliases
GLCLR
UniGene
Hs.315562
Gene Chromosome Location
Chr.1: 93885199-93909598 on Build GRCh38
Transcripts
NM_001308253.1 NM_002061.3 XM_011541261.2 XM_017001056.1 XM_017001057.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
465114 c.171A>G p.Q57Q Chr.1: 93904544-93904544 on Build GRCh38
6670026 c.133G>T p.D45Y Chr.1: 93904582-93904582 on Build GRCh38
7422730 c.172A>G p.I58V Chr.1: 93904543-93904543 on Build GRCh38
6487957 c.131A>T p.H44L Chr.1: 93904584-93904584 on Build GRCh38
5164882 c.148_149insA p.T50fs*4 Chr.1: 93904566-93904567 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750118004 Chr.1: 93904638-93904638 on Build GRCh38
rs1009036152 Chr.1: 93904756-93904756 on Build GRCh38
rs533969445 Chr.1: 93904843-93904843 on Build GRCh38
rs187554918 Chr.1: 93904725-93904725 on Build GRCh38
rs760798197 Chr.1: 93904493-93904493 on Build GRCh38
rs970239515 Chr.1: 93904465-93904465 on Build GRCh38
rs1014677782 Chr.1: 93904767-93904767 on Build GRCh38
rs1014455963 Chr.1: 93904858-93904858 on Build GRCh38
rs746214937 Chr.1: 93904814-93904814 on Build GRCh38
rs371631298 Chr.1: 93904491-93904491 on Build GRCh38
rs768556157 Chr.1: 93904479-93904479 on Build GRCh38
rs763595719 Chr.1: 93904580-93904580 on Build GRCh38
rs771288344 Chr.1: 93904753-93904753 on Build GRCh38
rs533326619 Chr.1: 93904776-93904776 on Build GRCh38
rs981038420 Chr.1: 93904817-93904817 on Build GRCh38
rs780642722 Chr.1: 93904717-93904717 on Build GRCh38
rs550965492 Chr.1: 93904690-93904690 on Build GRCh38
rs980241972 Chr.1: 93904469-93904473 on Build GRCh38
rs759637489 Chr.1: 93904556-93904556 on Build GRCh38
rs764691816 Chr.1: 93904584-93904584 on Build GRCh38
rs529591274 Chr.1: 93904682-93904682 on Build GRCh38
rs202041507 Chr.1: 93904679-93904680 on Build GRCh38
rs192600610 Chr.1: 93904802-93904802 on Build GRCh38
rs770862823 Chr.1: 93904501-93904501 on Build GRCh38
rs776920229 Chr.1: 93904512-93904512 on Build GRCh38
rs368031229 Chr.1: 93904565-93904565 on Build GRCh38
rs756804007 Chr.1: 93904629-93904629 on Build GRCh38
rs780500496 Chr.1: 93904630-93904630 on Build GRCh38
rs944633954 Chr.1: 93904674-93904674 on Build GRCh38
rs1040356854 Chr.1: 93904680-93904680 on Build GRCh38
rs150334675 Chr.1: 93904779-93904779 on Build GRCh38
rs907554349 Chr.1: 93904671-93904671 on Build GRCh38
rs1031658551 Chr.1: 93904816-93904816 on Build GRCh38
rs995787797 Chr.1: 93904693-93904693 on Build GRCh38
rs751170589 Chr.1: 93904598-93904598 on Build GRCh38
rs765656488 Chr.1: 93904562-93904562 on Build GRCh38
rs2234729 Chr.1: 93904660-93904660 on Build GRCh38
rs375362956 Chr.1: 93904492-93904492 on Build GRCh38

View Full Product Details