Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37063074-37063574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD17
Entrez Gene ID
79734
Gene Name
potassium channel tetramerization domain containing 17
Gene Aliases
-
UniGene
Hs.517597
Gene Chromosome Location
Chr.22: 37051725-37063390 on Build GRCh38
Transcripts
NM_001282684.1 NM_001282685.1 NM_001282686.1 NM_024681.3 XM_005261741.3 XM_005261744.2 XM_011530374.2 XM_011530377.2

SNP Detail

SNP ID Chromosome Location
rs1022520264 Chr.22: 37063448-37063448 on Build GRCh38
rs965238301 Chr.22: 37063112-37063112 on Build GRCh38
rs61214014 Chr.22: 37063419-37063419 on Build GRCh38
rs934370595 Chr.22: 37063251-37063251 on Build GRCh38
rs947067589 Chr.22: 37063211-37063211 on Build GRCh38
rs979281046 Chr.22: 37063391-37063391 on Build GRCh38
rs968230757 Chr.22: 37063458-37063458 on Build GRCh38
rs978403794 Chr.22: 37063465-37063465 on Build GRCh38
rs180708556 Chr.22: 37063178-37063178 on Build GRCh38
rs564644935 Chr.22: 37063270-37063270 on Build GRCh38
rs1030210643 Chr.22: 37063128-37063128 on Build GRCh38
rs572534859 Chr.22: 37063214-37063214 on Build GRCh38
rs8138791 Chr.22: 37063499-37063499 on Build GRCh38
rs988655472 Chr.22: 37063353-37063353 on Build GRCh38
rs753280846 Chr.22: 37063115-37063115 on Build GRCh38
rs948559958 Chr.22: 37063284-37063284 on Build GRCh38
rs976950601 Chr.22: 37063116-37063116 on Build GRCh38
rs993007964 Chr.22: 37063498-37063498 on Build GRCh38
rs937509425 Chr.22: 37063208-37063208 on Build GRCh38
rs1048507024 Chr.22: 37063472-37063472 on Build GRCh38
rs947467131 Chr.22: 37063157-37063157 on Build GRCh38
rs373025155 Chr.22: 37063292-37063293 on Build GRCh38
rs150924179 Chr.22: 37063552-37063552 on Build GRCh38
rs373925933 Chr.22: 37063247-37063247 on Build GRCh38
rs73886911 Chr.22: 37063310-37063310 on Build GRCh38
rs554299766 Chr.22: 37063199-37063199 on Build GRCh38
rs1006557121 Chr.22: 37063474-37063474 on Build GRCh38
rs529688110 Chr.22: 37063431-37063431 on Build GRCh38
rs913197480 Chr.22: 37063145-37063145 on Build GRCh38
rs767493300 Chr.22: 37063323-37063323 on Build GRCh38
rs968748312 Chr.22: 37063373-37063373 on Build GRCh38
rs765511833 Chr.22: 37063098-37063098 on Build GRCh38
rs115646797 Chr.22: 37063491-37063491 on Build GRCh38
rs978372810 Chr.22: 37063212-37063212 on Build GRCh38
rs889561748 Chr.22: 37063473-37063473 on Build GRCh38
rs56381800 Chr.22: 37063432-37063432 on Build GRCh38
rs142525657 Chr.22: 37063471-37063471 on Build GRCh38
rs1014357334 Chr.22: 37063332-37063332 on Build GRCh38
rs954230481 Chr.22: 37063191-37063191 on Build GRCh38
rs1010150586 Chr.22: 37063542-37063543 on Build GRCh38
rs11542757 Chr.22: 37063272-37063272 on Build GRCh38
rs573293088 Chr.22: 37063189-37063200 on Build GRCh38
rs376480859 Chr.22: 37063434-37063434 on Build GRCh38
rs558841460 Chr.22: 37063215-37063215 on Build GRCh38
rs1020278216 Chr.22: 37063370-37063370 on Build GRCh38
rs755708041 Chr.22: 37063424-37063424 on Build GRCh38
rs145848141 Chr.22: 37063288-37063289 on Build GRCh38
rs915480546 Chr.22: 37063209-37063209 on Build GRCh38
rs531824792 Chr.22: 37063289-37063289 on Build GRCh38
rs948550375 Chr.22: 37063548-37063548 on Build GRCh38
rs759192503 Chr.22: 37063271-37063271 on Build GRCh38
rs1015565123 Chr.22: 37063324-37063324 on Build GRCh38
rs889647264 Chr.22: 37063304-37063304 on Build GRCh38
rs985715512 Chr.22: 37063198-37063198 on Build GRCh38

View Full Product Details